Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56994445-56994706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2B
Entrez Gene ID
5431
Gene Name
RNA polymerase II subunit B
Gene Aliases
POL2RB, RPB2, hRPB140
UniGene
Hs.602757
Gene Chromosome Location
Chr.4: 56978640-57031162 on Build GRCh38
Transcripts
NM_000938.2 NM_001303268.1 NM_001303269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1310161 c.317C>T p.S106L Chr.4: 56994477-56994477 on Build GRCh38
5995547 c.397A>G p.I133V Chr.4: 56994687-56994687 on Build GRCh38
4922221 c.400A>G p.K134E Chr.4: 56994690-56994690 on Build GRCh38
4417857 c.406G>T p.G136C Chr.4: 56994696-56994696 on Build GRCh38
4125327 c.314C>A p.P105H Chr.4: 56994474-56994474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755585636 Chr.4: 56994616-56994616 on Build GRCh38
rs781004445 Chr.4: 56994670-56994670 on Build GRCh38
rs372042978 Chr.4: 56994543-56994543 on Build GRCh38
rs746874643 Chr.4: 56994479-56994479 on Build GRCh38
rs774340642 Chr.4: 56994534-56994534 on Build GRCh38
rs560279188 Chr.4: 56994542-56994542 on Build GRCh38
rs375114461 Chr.4: 56994659-56994659 on Build GRCh38
rs746756090 Chr.4: 56994657-56994657 on Build GRCh38
rs534908426 Chr.4: 56994549-56994549 on Build GRCh38
rs899217148 Chr.4: 56994593-56994597 on Build GRCh38
rs374386426 Chr.4: 56994525-56994525 on Build GRCh38
rs746229704 Chr.4: 56994623-56994624 on Build GRCh38
rs758067808 Chr.4: 56994636-56994636 on Build GRCh38
rs762078653 Chr.4: 56994537-56994537 on Build GRCh38
rs745781732 Chr.4: 56994671-56994671 on Build GRCh38
rs769673916 Chr.4: 56994673-56994673 on Build GRCh38
rs762979906 Chr.4: 56994519-56994519 on Build GRCh38
rs777297320 Chr.4: 56994642-56994642 on Build GRCh38
rs532686013 Chr.4: 56994567-56994567 on Build GRCh38
rs752207344 Chr.4: 56994627-56994627 on Build GRCh38
rs770511423 Chr.4: 56994516-56994516 on Build GRCh38
rs1055621830 Chr.4: 56994575-56994575 on Build GRCh38
rs926968230 Chr.4: 56994596-56994596 on Build GRCh38
rs770796818 Chr.4: 56994492-56994492 on Build GRCh38
rs146736128 Chr.4: 56994540-56994540 on Build GRCh38
rs78320117 Chr.4: 56994676-56994676 on Build GRCh38
rs938012550 Chr.4: 56994561-56994561 on Build GRCh38
rs989634466 Chr.4: 56994517-56994517 on Build GRCh38
rs1008851563 Chr.4: 56994656-56994656 on Build GRCh38
rs979728652 Chr.4: 56994594-56994594 on Build GRCh38
rs766947033 Chr.4: 56994555-56994555 on Build GRCh38
rs776570740 Chr.4: 56994515-56994515 on Build GRCh38
rs778465434 Chr.4: 56994620-56994620 on Build GRCh38
rs201940173 Chr.4: 56994622-56994622 on Build GRCh38

View Full Product Details