Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99401154-99401401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REV1
Entrez Gene ID
51455
Gene Name
REV1, DNA directed polymerase
Gene Aliases
AIBP80, REV1L
UniGene
Hs.443077
Gene Chromosome Location
Chr.2: 99400475-99490123 on Build GRCh38
Transcripts
NM_001037872.2 NM_001321454.1 NM_001321455.1 NM_001321458.1 NM_001321459.1 NM_001321460.1 NM_016316.3 NR_135649.1 NR_135650.1 NR_135651.1 NR_135652.1 NR_135653.1 XM_011511341.2 XM_017004309.1 XM_017004310.1 XM_017004311.1 XM_017004312.1 XM_017004313.1 XM_017004314.1 XM_017004315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904158 c.3682G>A p.A1228T Chr.2: 99401315-99401315 on Build GRCh38
7173065 c.3700G>C p.D1234H Chr.2: 99401297-99401297 on Build GRCh38
6765107 c.3652C>T p.Q1218* Chr.2: 99401345-99401345 on Build GRCh38
5904159 c.3681G>A p.M1227I Chr.2: 99401316-99401316 on Build GRCh38
1004396 c.3669G>A p.S1223S Chr.2: 99401328-99401328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746250159 Chr.2: 99401218-99401218 on Build GRCh38
rs377140119 Chr.2: 99401201-99401201 on Build GRCh38
rs761996646 Chr.2: 99401190-99401190 on Build GRCh38
rs561233948 Chr.2: 99401210-99401210 on Build GRCh38
rs931198045 Chr.2: 99401265-99401265 on Build GRCh38
rs372932712 Chr.2: 99401364-99401364 on Build GRCh38
rs767683665 Chr.2: 99401195-99401196 on Build GRCh38
rs960365081 Chr.2: 99401347-99401347 on Build GRCh38
rs762086858 Chr.2: 99401262-99401262 on Build GRCh38
rs760496438 Chr.2: 99401286-99401286 on Build GRCh38
rs763467666 Chr.2: 99401232-99401232 on Build GRCh38
rs928374501 Chr.2: 99401182-99401182 on Build GRCh38
rs146377515 Chr.2: 99401292-99401292 on Build GRCh38
rs770127623 Chr.2: 99401226-99401226 on Build GRCh38
rs199687154 Chr.2: 99401222-99401222 on Build GRCh38
rs767710888 Chr.2: 99401266-99401266 on Build GRCh38
rs750211082 Chr.2: 99401195-99401195 on Build GRCh38
rs752243571 Chr.2: 99401359-99401359 on Build GRCh38
rs756100486 Chr.2: 99401197-99401197 on Build GRCh38
rs757925939 Chr.2: 99401362-99401362 on Build GRCh38
rs1055385900 Chr.2: 99401219-99401219 on Build GRCh38
rs747843772 Chr.2: 99401207-99401207 on Build GRCh38
rs373211992 Chr.2: 99401370-99401370 on Build GRCh38
rs777569976 Chr.2: 99401366-99401366 on Build GRCh38
rs553247448 Chr.2: 99401340-99401340 on Build GRCh38
rs753772487 Chr.2: 99401317-99401317 on Build GRCh38
rs781728560 Chr.2: 99401217-99401217 on Build GRCh38
rs370092323 Chr.2: 99401206-99401206 on Build GRCh38
rs1026930672 Chr.2: 99401216-99401216 on Build GRCh38
rs919847060 Chr.2: 99401223-99401223 on Build GRCh38
rs747974845 Chr.2: 99401358-99401358 on Build GRCh38
rs768852923 Chr.2: 99401211-99401211 on Build GRCh38
rs748435976 Chr.2: 99401364-99401365 on Build GRCh38
rs142886583 Chr.2: 99401337-99401337 on Build GRCh38
rs774379931 Chr.2: 99401248-99401248 on Build GRCh38
rs757176462 Chr.2: 99401214-99401215 on Build GRCh38
rs767337091 Chr.2: 99401191-99401191 on Build GRCh38
rs369323449 Chr.2: 99401268-99401268 on Build GRCh38
rs754473039 Chr.2: 99401204-99401204 on Build GRCh38
rs371779609 Chr.2: 99401225-99401225 on Build GRCh38
rs768667488 Chr.2: 99401234-99401234 on Build GRCh38
rs752742712 Chr.2: 99401199-99401200 on Build GRCh38
rs539823384 Chr.2: 99401228-99401228 on Build GRCh38
rs750584264 Chr.2: 99401283-99401283 on Build GRCh38

View Full Product Details