Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12391020-12391267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF799
Entrez Gene ID
90576
Gene Name
zinc finger protein 799
Gene Aliases
HIT-40, ZNF842
UniGene
Hs.546637
Gene Chromosome Location
Chr.19: 12390014-12401274 on Build GRCh38
Transcripts
NM_001080821.2 NM_001322497.1 NM_001322498.1 XM_017027452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187273 c.622T>G p.Y208D Chr.19: 12391137-12391137 on Build GRCh38
7448035 c.1226A>G p.Q409R Chr.19: 12391172-12391172 on Build GRCh38
991267 c.710A>C p.K237T Chr.19: 12391049-12391049 on Build GRCh38
4745002 c.634C>G p.Q212E Chr.19: 12391125-12391125 on Build GRCh38
5464398 c.1239G>T p.K413N Chr.19: 12391159-12391159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776694616 Chr.19: 12391115-12391115 on Build GRCh38
rs375319755 Chr.19: 12391205-12391205 on Build GRCh38
rs767075260 Chr.19: 12391160-12391160 on Build GRCh38
rs752930102 Chr.19: 12391071-12391071 on Build GRCh38
rs769859128 Chr.19: 12391122-12391122 on Build GRCh38
rs765180227 Chr.19: 12391055-12391055 on Build GRCh38
rs567790989 Chr.19: 12391166-12391166 on Build GRCh38
rs747072794 Chr.19: 12391105-12391105 on Build GRCh38
rs778219417 Chr.19: 12391082-12391082 on Build GRCh38
rs183068429 Chr.19: 12391185-12391185 on Build GRCh38
rs891944936 Chr.19: 12391165-12391165 on Build GRCh38
rs746000060 Chr.19: 12391215-12391216 on Build GRCh38
rs773050618 Chr.19: 12391234-12391234 on Build GRCh38
rs754475061 Chr.19: 12391078-12391078 on Build GRCh38
rs781354071 Chr.19: 12391219-12391219 on Build GRCh38
rs750751850 Chr.19: 12391176-12391176 on Build GRCh38
rs201389705 Chr.19: 12391230-12391230 on Build GRCh38
rs749075683 Chr.19: 12391236-12391236 on Build GRCh38
rs796172009 Chr.19: 12391215-12391216 on Build GRCh38
rs757608439 Chr.19: 12391088-12391088 on Build GRCh38
rs758077310 Chr.19: 12391053-12391053 on Build GRCh38
rs759022575 Chr.19: 12391149-12391149 on Build GRCh38
rs780292679 Chr.19: 12391204-12391204 on Build GRCh38
rs556226523 Chr.19: 12391093-12391093 on Build GRCh38
rs751028377 Chr.19: 12391075-12391075 on Build GRCh38
rs761908060 Chr.19: 12391118-12391118 on Build GRCh38
rs758567703 Chr.19: 12391209-12391209 on Build GRCh38
rs199693720 Chr.19: 12391123-12391123 on Build GRCh38
rs759253785 Chr.19: 12391244-12391244 on Build GRCh38
rs781171891 Chr.19: 12391195-12391195 on Build GRCh38
rs762673934 Chr.19: 12391132-12391132 on Build GRCh38
rs200836165 Chr.19: 12391168-12391168 on Build GRCh38
rs146942808 Chr.19: 12391173-12391173 on Build GRCh38
rs754341434 Chr.19: 12391167-12391167 on Build GRCh38
rs750039130 Chr.19: 12391066-12391066 on Build GRCh38
rs549869787 Chr.19: 12391129-12391129 on Build GRCh38
rs770783358 Chr.19: 12391151-12391152 on Build GRCh38
rs746755456 Chr.19: 12391101-12391101 on Build GRCh38
rs770678785 Chr.19: 12391242-12391242 on Build GRCh38
rs866933924 Chr.19: 12391197-12391197 on Build GRCh38
rs372293608 Chr.19: 12391143-12391143 on Build GRCh38
rs748251265 Chr.19: 12391226-12391226 on Build GRCh38
rs779319756 Chr.19: 12391089-12391089 on Build GRCh38
rs780162300 Chr.19: 12391099-12391099 on Build GRCh38
rs1044878292 Chr.19: 12391108-12391108 on Build GRCh38
rs553762868 Chr.19: 12391212-12391212 on Build GRCh38
rs762599652 Chr.19: 12391068-12391068 on Build GRCh38
rs777745732 Chr.19: 12391081-12391081 on Build GRCh38
rs754394394 Chr.19: 12391086-12391086 on Build GRCh38
rs772279828 Chr.19: 12391098-12391098 on Build GRCh38
rs768899109 Chr.19: 12391107-12391107 on Build GRCh38

View Full Product Details