Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67203973-67204222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3361497 c.754C>T p.L252F Chr.15: 67204110-67204110 on Build GRCh38
1374225 c.782A>G p.N261S Chr.15: 67204082-67204082 on Build GRCh38
2257433 c.744A>T p.E248D Chr.15: 67204120-67204120 on Build GRCh38
6432454 c.727G>T p.D243Y Chr.15: 67204137-67204137 on Build GRCh38
1749287 c.780G>A p.E260E Chr.15: 67204084-67204084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756589595 Chr.15: 67204066-67204066 on Build GRCh38
rs1010715115 Chr.15: 67204174-67204174 on Build GRCh38
rs749583839 Chr.15: 67204081-67204081 on Build GRCh38
rs939736095 Chr.15: 67204039-67204039 on Build GRCh38
rs867797999 Chr.15: 67204146-67204146 on Build GRCh38
rs767464285 Chr.15: 67204065-67204066 on Build GRCh38
rs774644135 Chr.15: 67204195-67204195 on Build GRCh38
rs755480469 Chr.15: 67204027-67204027 on Build GRCh38
rs764806559 Chr.15: 67204134-67204134 on Build GRCh38
rs919809911 Chr.15: 67204097-67204097 on Build GRCh38
rs367555610 Chr.15: 67204050-67204050 on Build GRCh38
rs915968056 Chr.15: 67204003-67204003 on Build GRCh38
rs777838692 Chr.15: 67204067-67204067 on Build GRCh38
rs765401006 Chr.15: 67204104-67204105 on Build GRCh38
rs375444996 Chr.15: 67204137-67204137 on Build GRCh38
rs371685408 Chr.15: 67204089-67204089 on Build GRCh38
rs141177071 Chr.15: 67204180-67204180 on Build GRCh38
rs1046128547 Chr.15: 67204045-67204045 on Build GRCh38
rs760748779 Chr.15: 67204092-67204094 on Build GRCh38
rs369410834 Chr.15: 67204116-67204116 on Build GRCh38
rs372325047 Chr.15: 67204120-67204120 on Build GRCh38
rs756536786 Chr.15: 67204155-67204155 on Build GRCh38
rs768110809 Chr.15: 67204151-67204151 on Build GRCh38
rs752656407 Chr.15: 67204065-67204065 on Build GRCh38
rs753310459 Chr.15: 67204154-67204154 on Build GRCh38
rs146169842 Chr.15: 67204190-67204190 on Build GRCh38
rs375593998 Chr.15: 67204049-67204049 on Build GRCh38
rs761214742 Chr.15: 67204009-67204009 on Build GRCh38
rs754158336 Chr.15: 67204176-67204176 on Build GRCh38
rs746782588 Chr.15: 67204112-67204112 on Build GRCh38
rs368693236 Chr.15: 67204142-67204142 on Build GRCh38
rs778984821 Chr.15: 67204094-67204094 on Build GRCh38
rs770852267 Chr.15: 67204025-67204026 on Build GRCh38
rs376820468 Chr.15: 67204107-67204107 on Build GRCh38
rs764587719 Chr.15: 67204173-67204173 on Build GRCh38
rs764866327 Chr.15: 67204023-67204023 on Build GRCh38
rs768157646 Chr.15: 67204038-67204038 on Build GRCh38
rs745960635 Chr.15: 67204103-67204103 on Build GRCh38
rs1044858345 Chr.15: 67204096-67204096 on Build GRCh38
rs775445355 Chr.15: 67204110-67204110 on Build GRCh38
rs931214910 Chr.15: 67204098-67204098 on Build GRCh38
rs761459547 Chr.15: 67204123-67204123 on Build GRCh38
rs867395597 Chr.15: 67204125-67204125 on Build GRCh38
rs749904131 Chr.15: 67204026-67204026 on Build GRCh38
rs1046940979 Chr.15: 67204100-67204100 on Build GRCh38

View Full Product Details