Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 33829691-33829936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC3
Entrez Gene ID
5983
Gene Name
replication factor C subunit 3
Gene Aliases
RFC38
UniGene
Hs.115474
Gene Chromosome Location
Chr.13: 33818049-33973946 on Build GRCh38
Transcripts
NM_002915.3 NM_181558.2 XM_011535172.2 XM_011535173.2 XM_011535174.2 XM_011535175.1 XM_017020680.1 XM_017020681.1 XM_017020682.1 XM_017020683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6765236 c.446G>A p.R149Q Chr.13: 33829890-33829890 on Build GRCh38
5349894 c.413A>T p.D138V Chr.13: 33829857-33829857 on Build GRCh38
1366560 c.423C>A p.T141T Chr.13: 33829867-33829867 on Build GRCh38
4047162 c.449G>A p.R150K Chr.13: 33829893-33829893 on Build GRCh38
3955547 c.396A>G p.V132V Chr.13: 33829840-33829840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960762368 Chr.13: 33829810-33829810 on Build GRCh38
rs757642535 Chr.13: 33829854-33829854 on Build GRCh38
rs749859448 Chr.13: 33829864-33829864 on Build GRCh38
rs751954699 Chr.13: 33829849-33829849 on Build GRCh38
rs779586709 Chr.13: 33829869-33829869 on Build GRCh38
rs755589016 Chr.13: 33829867-33829867 on Build GRCh38
rs1043371901 Chr.13: 33829746-33829746 on Build GRCh38
rs1052759992 Chr.13: 33829751-33829751 on Build GRCh38
rs369364314 Chr.13: 33829803-33829803 on Build GRCh38
rs768543538 Chr.13: 33829733-33829735 on Build GRCh38
rs113168994 Chr.13: 33829757-33829757 on Build GRCh38
rs764546364 Chr.13: 33829844-33829844 on Build GRCh38
rs778506103 Chr.13: 33829898-33829898 on Build GRCh38
rs748928589 Chr.13: 33829873-33829873 on Build GRCh38
rs748032032 Chr.13: 33829788-33829788 on Build GRCh38
rs763321786 Chr.13: 33829835-33829835 on Build GRCh38
rs1002333308 Chr.13: 33829807-33829807 on Build GRCh38
rs199939532 Chr.13: 33829876-33829876 on Build GRCh38
rs1053318530 Chr.13: 33829723-33829723 on Build GRCh38
rs766615656 Chr.13: 33829893-33829893 on Build GRCh38
rs775826688 Chr.13: 33829819-33829819 on Build GRCh38
rs370103377 Chr.13: 33829825-33829825 on Build GRCh38
rs5001163 Chr.13: 33829840-33829840 on Build GRCh38
rs1004001007 Chr.13: 33829833-33829833 on Build GRCh38
rs888729206 Chr.13: 33829842-33829842 on Build GRCh38
rs749240594 Chr.13: 33829824-33829824 on Build GRCh38
rs759687839 Chr.13: 33829805-33829805 on Build GRCh38
rs200751265 Chr.13: 33829861-33829861 on Build GRCh38
rs533458165 Chr.13: 33829780-33829784 on Build GRCh38
rs770365679 Chr.13: 33829906-33829907 on Build GRCh38
rs1011843061 Chr.13: 33829745-33829745 on Build GRCh38
rs754693463 Chr.13: 33829886-33829886 on Build GRCh38
rs548117002 Chr.13: 33829729-33829729 on Build GRCh38
rs772051205 Chr.13: 33829791-33829791 on Build GRCh38
rs777824953 Chr.13: 33829811-33829815 on Build GRCh38
rs768090866 Chr.13: 33829860-33829860 on Build GRCh38
rs1805374 Chr.13: 33829809-33829809 on Build GRCh38

View Full Product Details