Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45690956-45691229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM69
Entrez Gene ID
51249
Gene Name
transmembrane protein 69
Gene Aliases
C1orf154
UniGene
Hs.699548
Gene Chromosome Location
Chr.1: 45688175-45694436 on Build GRCh38
Transcripts
NM_016486.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
464690 c.12C>T p.F4F Chr.1: 45691080-45691080 on Build GRCh38
3805417 c.13A>C p.I5L Chr.1: 45691081-45691081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950932836 Chr.1: 45691128-45691128 on Build GRCh38
rs749365242 Chr.1: 45691161-45691161 on Build GRCh38
rs957767313 Chr.1: 45691149-45691149 on Build GRCh38
rs971476838 Chr.1: 45691176-45691176 on Build GRCh38
rs780582875 Chr.1: 45691111-45691111 on Build GRCh38
rs909046749 Chr.1: 45690983-45690983 on Build GRCh38
rs560399038 Chr.1: 45691130-45691130 on Build GRCh38
rs773205108 Chr.1: 45691058-45691058 on Build GRCh38
rs895922862 Chr.1: 45690990-45690990 on Build GRCh38
rs762825469 Chr.1: 45691065-45691065 on Build GRCh38
rs773298836 Chr.1: 45691159-45691159 on Build GRCh38
rs376918735 Chr.1: 45691085-45691085 on Build GRCh38
rs753340227 Chr.1: 45691096-45691096 on Build GRCh38
rs370166809 Chr.1: 45691127-45691127 on Build GRCh38
rs781475881 Chr.1: 45691131-45691131 on Build GRCh38
rs369476853 Chr.1: 45691042-45691042 on Build GRCh38
rs529838228 Chr.1: 45691075-45691075 on Build GRCh38
rs150511863 Chr.1: 45691020-45691020 on Build GRCh38
rs758882971 Chr.1: 45691110-45691110 on Build GRCh38
rs1048858007 Chr.1: 45690981-45690981 on Build GRCh38
rs747381423 Chr.1: 45691021-45691021 on Build GRCh38
rs1014312558 Chr.1: 45691008-45691008 on Build GRCh38
rs752413276 Chr.1: 45691083-45691083 on Build GRCh38
rs774062687 Chr.1: 45691067-45691067 on Build GRCh38
rs974101759 Chr.1: 45691048-45691048 on Build GRCh38
rs193298114 Chr.1: 45691205-45691205 on Build GRCh38
rs776832374 Chr.1: 45691047-45691047 on Build GRCh38
rs768804465 Chr.1: 45691030-45691030 on Build GRCh38
rs766291319 Chr.1: 45691066-45691066 on Build GRCh38
rs770813752 Chr.1: 45691162-45691162 on Build GRCh38
rs761451942 Chr.1: 45691094-45691094 on Build GRCh38
rs756577805 Chr.1: 45691096-45691099 on Build GRCh38
rs199639691 Chr.1: 45691076-45691076 on Build GRCh38
rs769960536 Chr.1: 45691151-45691151 on Build GRCh38
rs769873085 Chr.1: 45691057-45691057 on Build GRCh38
rs890016943 Chr.1: 45691117-45691117 on Build GRCh38
rs1020311136 Chr.1: 45691165-45691165 on Build GRCh38
rs761935527 Chr.1: 45691055-45691055 on Build GRCh38
rs527525927 Chr.1: 45691134-45691134 on Build GRCh38
rs115547462 Chr.1: 45691203-45691203 on Build GRCh38
rs940574142 Chr.1: 45690996-45690996 on Build GRCh38

View Full Product Details