Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19224606-19224879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT17
Entrez Gene ID
51760
Gene Name
synaptotagmin 17
Gene Aliases
sytXVII
UniGene
Hs.258326
Gene Chromosome Location
Chr.16: 19167834-19268334 on Build GRCh38
Transcripts
NM_001308157.1 NM_016524.3 XM_005255362.3 XM_011545865.1 XM_011545866.2 XM_011545868.1 XM_011545869.2 XM_011545870.2 XM_011545871.2 XM_011545872.2 XM_017023280.1 XM_017023281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230316 c.1204G>A p.E402K Chr.16: 19224814-19224814 on Build GRCh38
6893754 c.1133C>T p.S378F Chr.16: 19224743-19224743 on Build GRCh38
1678868 c.1130C>T p.T377M Chr.16: 19224740-19224740 on Build GRCh38
702598 c.1163A>G p.Y388C Chr.16: 19224773-19224773 on Build GRCh38
3506667 c.1089C>T p.I363I Chr.16: 19224699-19224699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868397125 Chr.16: 19224821-19224821 on Build GRCh38
rs749640525 Chr.16: 19224843-19224843 on Build GRCh38
rs774549627 Chr.16: 19224724-19224724 on Build GRCh38
rs745935226 Chr.16: 19224732-19224732 on Build GRCh38
rs778072355 Chr.16: 19224705-19224705 on Build GRCh38
rs753234680 Chr.16: 19224805-19224805 on Build GRCh38
rs974757188 Chr.16: 19224739-19224739 on Build GRCh38
rs867094576 Chr.16: 19224848-19224848 on Build GRCh38
rs767002815 Chr.16: 19224796-19224796 on Build GRCh38
rs1014581003 Chr.16: 19224721-19224721 on Build GRCh38
rs373690785 Chr.16: 19224711-19224711 on Build GRCh38
rs2072076 Chr.16: 19224859-19224859 on Build GRCh38
rs778123484 Chr.16: 19224828-19224828 on Build GRCh38
rs758054805 Chr.16: 19224670-19224670 on Build GRCh38
rs781519434 Chr.16: 19224684-19224684 on Build GRCh38
rs748558908 Chr.16: 19224686-19224686 on Build GRCh38
rs765942529 Chr.16: 19224781-19224781 on Build GRCh38
rs577729100 Chr.16: 19224631-19224631 on Build GRCh38
rs765034825 Chr.16: 19224657-19224657 on Build GRCh38
rs761672917 Chr.16: 19224649-19224649 on Build GRCh38
rs761799665 Chr.16: 19224740-19224740 on Build GRCh38
rs961588058 Chr.16: 19224738-19224738 on Build GRCh38
rs959811634 Chr.16: 19224748-19224748 on Build GRCh38
rs749518133 Chr.16: 19224706-19224706 on Build GRCh38
rs776690349 Chr.16: 19224736-19224736 on Build GRCh38
rs756491245 Chr.16: 19224687-19224687 on Build GRCh38
rs371413320 Chr.16: 19224682-19224682 on Build GRCh38
rs879243389 Chr.16: 19224673-19224673 on Build GRCh38
rs746311546 Chr.16: 19224653-19224653 on Build GRCh38
rs757569403 Chr.16: 19224854-19224854 on Build GRCh38
rs765922702 Chr.16: 19224672-19224672 on Build GRCh38
rs995797638 Chr.16: 19224756-19224756 on Build GRCh38
rs762720358 Chr.16: 19224761-19224761 on Build GRCh38
rs768683487 Chr.16: 19224735-19224735 on Build GRCh38
rs773865114 Chr.16: 19224760-19224760 on Build GRCh38
rs570336679 Chr.16: 19224630-19224630 on Build GRCh38
rs118182524 Chr.16: 19224741-19224741 on Build GRCh38
rs751120526 Chr.16: 19224782-19224782 on Build GRCh38
rs376465552 Chr.16: 19224679-19224679 on Build GRCh38
rs754422083 Chr.16: 19224786-19224786 on Build GRCh38
rs199781941 Chr.16: 19224665-19224665 on Build GRCh38
rs756548346 Chr.16: 19224822-19224822 on Build GRCh38

View Full Product Details