Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4859705-4859956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLIN3
Entrez Gene ID
10226
Gene Name
perilipin 3
Gene Aliases
M6PRBP1, PP17, TIP47
UniGene
Hs.140452
Gene Chromosome Location
Chr.19: 4838334-4867768 on Build GRCh38
Transcripts
NM_001164189.1 NM_001164194.1 NM_005817.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304847 c.181G>A p.D61N Chr.19: 4859910-4859910 on Build GRCh38
367203 c.199G>T p.V67L Chr.19: 4859892-4859892 on Build GRCh38
6276269 c.165C>A p.H55Q Chr.19: 4859926-4859926 on Build GRCh38
3960245 c.256G>C p.E86Q Chr.19: 4859835-4859835 on Build GRCh38
6351349 c.166A>G p.I56V Chr.19: 4859925-4859925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984952523 Chr.19: 4859908-4859908 on Build GRCh38
rs774224377 Chr.19: 4859929-4859929 on Build GRCh38
rs768641790 Chr.19: 4859848-4859848 on Build GRCh38
rs933736006 Chr.19: 4859830-4859830 on Build GRCh38
rs374429923 Chr.19: 4859886-4859886 on Build GRCh38
rs1007398252 Chr.19: 4859802-4859802 on Build GRCh38
rs370728215 Chr.19: 4859777-4859777 on Build GRCh38
rs765819103 Chr.19: 4859863-4859863 on Build GRCh38
rs779604772 Chr.19: 4859776-4859776 on Build GRCh38
rs762394082 Chr.19: 4859862-4859862 on Build GRCh38
rs748210637 Chr.19: 4859745-4859745 on Build GRCh38
rs368003339 Chr.19: 4859905-4859905 on Build GRCh38
rs764459098 Chr.19: 4859930-4859930 on Build GRCh38
rs772404378 Chr.19: 4859894-4859894 on Build GRCh38
rs2292150 Chr.19: 4859801-4859801 on Build GRCh38
rs769584712 Chr.19: 4859899-4859899 on Build GRCh38
rs755409894 Chr.19: 4859735-4859735 on Build GRCh38
rs8289 Chr.19: 4859925-4859925 on Build GRCh38
rs771000325 Chr.19: 4859816-4859816 on Build GRCh38
rs769845235 Chr.19: 4859926-4859926 on Build GRCh38
rs761403657 Chr.19: 4859811-4859811 on Build GRCh38
rs774734665 Chr.19: 4859824-4859824 on Build GRCh38
rs770186633 Chr.19: 4859794-4859794 on Build GRCh38
rs2292149 Chr.19: 4859876-4859876 on Build GRCh38
rs1011121844 Chr.19: 4859844-4859844 on Build GRCh38
rs190599490 Chr.19: 4859762-4859762 on Build GRCh38
rs141196203 Chr.19: 4859875-4859875 on Build GRCh38
rs892227902 Chr.19: 4859842-4859842 on Build GRCh38
rs147933215 Chr.19: 4859852-4859852 on Build GRCh38
rs960810299 Chr.19: 4859759-4859759 on Build GRCh38
rs757295388 Chr.19: 4859884-4859884 on Build GRCh38
rs751165075 Chr.19: 4859788-4859788 on Build GRCh38
rs202240827 Chr.19: 4859910-4859910 on Build GRCh38
rs773161639 Chr.19: 4859859-4859859 on Build GRCh38
rs778170698 Chr.19: 4859879-4859879 on Build GRCh38
rs560198401 Chr.19: 4859760-4859760 on Build GRCh38
rs749292936 Chr.19: 4859810-4859810 on Build GRCh38
rs947646583 Chr.19: 4859911-4859911 on Build GRCh38
rs938913879 Chr.19: 4859755-4859755 on Build GRCh38
rs1002107126 Chr.19: 4859860-4859860 on Build GRCh38
rs960969907 Chr.19: 4859888-4859888 on Build GRCh38
rs568505257 Chr.19: 4859882-4859882 on Build GRCh38
rs765938093 Chr.19: 4859907-4859907 on Build GRCh38
rs753038078 Chr.19: 4859877-4859877 on Build GRCh38
rs756680594 Chr.19: 4859878-4859878 on Build GRCh38
rs746153640 Chr.19: 4859891-4859891 on Build GRCh38
rs773038339 Chr.19: 4859902-4859902 on Build GRCh38
rs747927169 Chr.19: 4859895-4859895 on Build GRCh38
rs375256286 Chr.19: 4859793-4859793 on Build GRCh38
rs769954922 Chr.19: 4859791-4859791 on Build GRCh38
rs370874870 Chr.19: 4859851-4859851 on Build GRCh38
rs868074798 Chr.19: 4859898-4859898 on Build GRCh38
rs754372917 Chr.19: 4859935-4859935 on Build GRCh38
rs532074339 Chr.19: 4859833-4859833 on Build GRCh38
rs45488495 Chr.19: 4859737-4859737 on Build GRCh38
rs767335755 Chr.19: 4859912-4859915 on Build GRCh38
rs930779505 Chr.19: 4859763-4859763 on Build GRCh38
rs757829695 Chr.19: 4859936-4859936 on Build GRCh38
rs984967457 Chr.19: 4859772-4859772 on Build GRCh38
rs11552103 Chr.19: 4859920-4859920 on Build GRCh38

View Full Product Details