Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134137946-134138219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF7
Entrez Gene ID
6932
Gene Name
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases
TCF-1
UniGene
Hs.573153
Gene Chromosome Location
Chr.5: 134114703-134148229 on Build GRCh38
Transcripts
NM_001134851.2 NM_003202.3 NM_201632.3 NM_201634.3 NM_213648.3 NR_033449.1 XM_006714678.3 XM_006714679.3 XM_006714680.3 XM_006714682.2 XM_006714684.2 XM_006714685.3 XM_006714686.3 XM_006714687.3 XM_011543604.2 XM_011543606.1 XM_011543607.2 XM_011543608.2 XM_011543609.2 XM_011543610.2 XM_011543611.2 XM_011543612.2 XM_011543613.2 XM_017009789.1 XM_017009790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275173 c.547+7G>T p.? Chr.5: 134138171-134138171 on Build GRCh38
4337807 c.111_112insC p.H40fs*37 Chr.5: 134138073-134138074 on Build GRCh38
6275170 c.202+7G>T p.? Chr.5: 134138171-134138171 on Build GRCh38
6168647 c.447G>T p.K149N Chr.5: 134138064-134138064 on Build GRCh38
6168644 c.102G>T p.K34N Chr.5: 134138064-134138064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759259896 Chr.5: 134138105-134138105 on Build GRCh38
rs923712839 Chr.5: 134137970-134137970 on Build GRCh38
rs756616925 Chr.5: 134138080-134138080 on Build GRCh38
rs1039666827 Chr.5: 134138069-134138069 on Build GRCh38
rs751079657 Chr.5: 134138124-134138124 on Build GRCh38
rs765049839 Chr.5: 134138106-134138106 on Build GRCh38
rs547059055 Chr.5: 134138136-134138136 on Build GRCh38
rs749714755 Chr.5: 134138031-134138031 on Build GRCh38
rs748400515 Chr.5: 134138083-134138083 on Build GRCh38
rs373409914 Chr.5: 134138145-134138145 on Build GRCh38
rs557713669 Chr.5: 134138056-134138056 on Build GRCh38
rs142489265 Chr.5: 134138118-134138118 on Build GRCh38
rs775236301 Chr.5: 134138186-134138186 on Build GRCh38
rs748561799 Chr.5: 134138144-134138144 on Build GRCh38
rs144072187 Chr.5: 134138075-134138075 on Build GRCh38
rs766265869 Chr.5: 134138067-134138067 on Build GRCh38
rs768166196 Chr.5: 134138029-134138032 on Build GRCh38
rs55718907 Chr.5: 134138173-134138173 on Build GRCh38
rs770887816 Chr.5: 134138098-134138098 on Build GRCh38
rs777914631 Chr.5: 134138147-134138147 on Build GRCh38
rs375582645 Chr.5: 134138065-134138065 on Build GRCh38
rs779396127 Chr.5: 134138143-134138143 on Build GRCh38
rs747735893 Chr.5: 134138047-134138049 on Build GRCh38
rs933729012 Chr.5: 134137973-134137973 on Build GRCh38
rs73790177 Chr.5: 134137992-134137992 on Build GRCh38
rs747052436 Chr.5: 134138149-134138149 on Build GRCh38
rs775076567 Chr.5: 134138107-134138107 on Build GRCh38
rs769773246 Chr.5: 134138174-134138174 on Build GRCh38
rs369357652 Chr.5: 134138026-134138026 on Build GRCh38
rs755343167 Chr.5: 134138140-134138140 on Build GRCh38
rs908867552 Chr.5: 134138070-134138070 on Build GRCh38
rs756780122 Chr.5: 134138133-134138133 on Build GRCh38
rs768999327 Chr.5: 134138035-134138035 on Build GRCh38
rs772307103 Chr.5: 134138049-134138049 on Build GRCh38
rs774198453 Chr.5: 134138025-134138025 on Build GRCh38
rs148407013 Chr.5: 134138096-134138096 on Build GRCh38
rs372641597 Chr.5: 134138043-134138043 on Build GRCh38
rs766839118 Chr.5: 134138134-134138134 on Build GRCh38
rs770857766 Chr.5: 134138095-134138100 on Build GRCh38
rs762574704 Chr.5: 134138111-134138111 on Build GRCh38
rs770920272 Chr.5: 134138155-134138155 on Build GRCh38
rs977923140 Chr.5: 134138166-134138166 on Build GRCh38
rs249610 Chr.5: 134138006-134138006 on Build GRCh38
rs773062085 Chr.5: 134138095-134138098 on Build GRCh38
rs771602751 Chr.5: 134138073-134138074 on Build GRCh38
rs561867522 Chr.5: 134138082-134138082 on Build GRCh38
rs774724579 Chr.5: 134138042-134138042 on Build GRCh38
rs1035031553 Chr.5: 134138091-134138091 on Build GRCh38
rs776512352 Chr.5: 134138169-134138169 on Build GRCh38
rs965675124 Chr.5: 134137979-134137979 on Build GRCh38
rs990520777 Chr.5: 134138076-134138076 on Build GRCh38
rs375338781 Chr.5: 134138024-134138024 on Build GRCh38
rs757866191 Chr.5: 134138078-134138078 on Build GRCh38
rs752351497 Chr.5: 134138077-134138077 on Build GRCh38
rs778319819 Chr.5: 134138044-134138046 on Build GRCh38
rs747124527 Chr.5: 134138097-134138097 on Build GRCh38
rs1020308659 Chr.5: 134137966-134137966 on Build GRCh38
rs147318706 Chr.5: 134138079-134138079 on Build GRCh38
rs973278726 Chr.5: 134137989-134137989 on Build GRCh38
rs760546259 Chr.5: 134138095-134138096 on Build GRCh38

View Full Product Details