Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35809333-35809594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPR2
Entrez Gene ID
4882
Gene Name
natriuretic peptide receptor 2
Gene Aliases
AMDM, ANPRB, ANPb, ECDM, GUC2B, GUCY2B, NPRB, NPRBi, SNSK
UniGene
Hs.78518
Gene Chromosome Location
Chr.9: 35752945-35809731 on Build GRCh38
Transcripts
NM_003995.3 XM_005251478.3 XM_005251479.4 XM_011517890.2 XM_011517891.2 XM_011517892.2 XM_011517893.2 XM_011517895.2 XM_017014745.1 XM_017014746.1 XM_017014747.1 XM_017014748.1 XM_017014749.1 XM_017014750.1 NM_172312.1

Gene Symbol
SPAG8
Entrez Gene ID
26206
Gene Name
sperm associated antigen 8
Gene Aliases
BS-84, CILD28, CT142, HSD-1, SMP1, SPAG3, hSMP-1
UniGene
Hs.256747
Gene Chromosome Location
Chr.9: 35807785-35812262 on Build GRCh38
Transcripts
NM_003995.3 XM_005251478.3 XM_005251479.4 XM_011517890.2 XM_011517891.2 XM_011517892.2 XM_011517893.2 XM_011517895.2 XM_017014745.1 XM_017014746.1 XM_017014747.1 XM_017014748.1 XM_017014749.1 XM_017014750.1 NM_172312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
254868 c.1360C>G p.L454V Chr.9: 35809416-35809416 on Build GRCh38
7142704 c.1372+8C>A p.? Chr.9: 35809396-35809396 on Build GRCh38
7413743 c.3125G>A p.G1042E Chr.9: 35809426-35809426 on Build GRCh38
381676 c.3119G>T p.R1040L Chr.9: 35809420-35809420 on Build GRCh38
231179 c.1326_1327GG>AA p.W442>? Chr.9: 35809449-35809450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768756676 Chr.9: 35809467-35809467 on Build GRCh38
rs985223475 Chr.9: 35809403-35809403 on Build GRCh38
rs763697758 Chr.9: 35809545-35809545 on Build GRCh38
rs776228913 Chr.9: 35809367-35809367 on Build GRCh38
rs758121945 Chr.9: 35809376-35809376 on Build GRCh38
rs779860186 Chr.9: 35809564-35809564 on Build GRCh38
rs756198197 Chr.9: 35809561-35809562 on Build GRCh38
rs970886339 Chr.9: 35809528-35809528 on Build GRCh38
rs368245199 Chr.9: 35809374-35809374 on Build GRCh38
rs746901282 Chr.9: 35809463-35809463 on Build GRCh38
rs778618798 Chr.9: 35809447-35809447 on Build GRCh38
rs760078276 Chr.9: 35809544-35809544 on Build GRCh38
rs138010251 Chr.9: 35809554-35809555 on Build GRCh38
rs752131387 Chr.9: 35809520-35809520 on Build GRCh38
rs756826864 Chr.9: 35809550-35809550 on Build GRCh38
rs766860402 Chr.9: 35809405-35809405 on Build GRCh38
rs771871411 Chr.9: 35809456-35809456 on Build GRCh38
rs138254005 Chr.9: 35809388-35809388 on Build GRCh38
rs572542145 Chr.9: 35809518-35809518 on Build GRCh38
rs923892675 Chr.9: 35809446-35809446 on Build GRCh38
rs776626025 Chr.9: 35809488-35809488 on Build GRCh38
rs756900261 Chr.9: 35809433-35809433 on Build GRCh38
rs762037268 Chr.9: 35809492-35809492 on Build GRCh38
rs556064802 Chr.9: 35809462-35809462 on Build GRCh38
rs139173465 Chr.9: 35809498-35809498 on Build GRCh38
rs752073130 Chr.9: 35809414-35809414 on Build GRCh38
rs758153314 Chr.9: 35809561-35809561 on Build GRCh38
rs183710056 Chr.9: 35809455-35809455 on Build GRCh38
rs777719336 Chr.9: 35809548-35809548 on Build GRCh38
rs750070790 Chr.9: 35809554-35809554 on Build GRCh38
rs753336898 Chr.9: 35809548-35809548 on Build GRCh38
rs763264983 Chr.9: 35809516-35809516 on Build GRCh38
rs755731628 Chr.9: 35809419-35809419 on Build GRCh38
rs773478755 Chr.9: 35809511-35809511 on Build GRCh38
rs778475167 Chr.9: 35809553-35809553 on Build GRCh38
rs146546770 Chr.9: 35809420-35809420 on Build GRCh38
rs143876283 Chr.9: 35809396-35809396 on Build GRCh38
rs765639995 Chr.9: 35809382-35809382 on Build GRCh38

View Full Product Details