Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102424897-102425170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100630923
Entrez Gene ID
100630923
Gene Name
LOC100289561-PRKRIP1 readthrough
Gene Aliases
-
UniGene
Hs.406395
Gene Chromosome Location
Chr.7: 102363861-102426682 on Build GRCh38
Transcripts
NR_038967.1 NM_024653.3

Gene Symbol
PRKRIP1
Entrez Gene ID
79706
Gene Name
PRKR interacting protein 1 (IL11 inducible)
Gene Aliases
C114, KRBOX3
UniGene
Hs.722104
Gene Chromosome Location
Chr.7: 102396357-102426682 on Build GRCh38
Transcripts
NR_038967.1 NM_024653.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4496296 c.471C>T p.P157P Chr.7: 102425027-102425027 on Build GRCh38
6175347 c.496G>T p.A166S Chr.7: 102425052-102425052 on Build GRCh38
5032245 c.517G>C p.E173Q Chr.7: 102425073-102425073 on Build GRCh38
3394428 c.551G>A p.R184Q Chr.7: 102425107-102425107 on Build GRCh38
3876720 c.543C>A p.T181T Chr.7: 102425099-102425099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782062616 Chr.7: 102425078-102425078 on Build GRCh38
rs782674418 Chr.7: 102425133-102425133 on Build GRCh38
rs200275795 Chr.7: 102424993-102424993 on Build GRCh38
rs782765412 Chr.7: 102425123-102425123 on Build GRCh38
rs782008491 Chr.7: 102424940-102424940 on Build GRCh38
rs563458955 Chr.7: 102425019-102425019 on Build GRCh38
rs781964538 Chr.7: 102425082-102425082 on Build GRCh38
rs782339015 Chr.7: 102425081-102425081 on Build GRCh38
rs781814373 Chr.7: 102425114-102425114 on Build GRCh38
rs553217491 Chr.7: 102425058-102425058 on Build GRCh38
rs986656481 Chr.7: 102425090-102425090 on Build GRCh38
rs782223011 Chr.7: 102425148-102425148 on Build GRCh38
rs142831999 Chr.7: 102425013-102425013 on Build GRCh38
rs1040129115 Chr.7: 102424938-102424938 on Build GRCh38
rs147851001 Chr.7: 102425107-102425107 on Build GRCh38
rs912376960 Chr.7: 102425093-102425093 on Build GRCh38
rs782104266 Chr.7: 102425099-102425100 on Build GRCh38
rs782690144 Chr.7: 102425006-102425006 on Build GRCh38
rs782757106 Chr.7: 102425021-102425021 on Build GRCh38
rs782468737 Chr.7: 102425115-102425115 on Build GRCh38
rs142271788 Chr.7: 102425087-102425087 on Build GRCh38
rs759502260 Chr.7: 102425062-102425062 on Build GRCh38
rs782115408 Chr.7: 102425084-102425084 on Build GRCh38
rs782812255 Chr.7: 102425112-102425113 on Build GRCh38
rs781908084 Chr.7: 102425045-102425045 on Build GRCh38
rs782402810 Chr.7: 102424969-102424969 on Build GRCh38
rs557218595 Chr.7: 102424955-102424955 on Build GRCh38
rs537188823 Chr.7: 102424978-102424978 on Build GRCh38
rs782766480 Chr.7: 102425038-102425038 on Build GRCh38
rs541814613 Chr.7: 102424956-102424956 on Build GRCh38
rs782364208 Chr.7: 102425059-102425059 on Build GRCh38
rs781924023 Chr.7: 102425087-102425088 on Build GRCh38
rs782013735 Chr.7: 102425001-102425001 on Build GRCh38
rs112698409 Chr.7: 102424985-102424985 on Build GRCh38
rs782698508 Chr.7: 102424973-102424973 on Build GRCh38
rs782669770 Chr.7: 102425031-102425031 on Build GRCh38
rs782327774 Chr.7: 102425069-102425070 on Build GRCh38
rs570116589 Chr.7: 102424966-102424966 on Build GRCh38
rs372824265 Chr.7: 102424968-102424968 on Build GRCh38
rs781865486 Chr.7: 102425126-102425126 on Build GRCh38
rs782416361 Chr.7: 102425072-102425072 on Build GRCh38
rs887190447 Chr.7: 102424983-102424983 on Build GRCh38
rs942626197 Chr.7: 102425104-102425104 on Build GRCh38
rs782018191 Chr.7: 102425106-102425106 on Build GRCh38
rs782708307 Chr.7: 102425112-102425112 on Build GRCh38
rs574816468 Chr.7: 102425076-102425076 on Build GRCh38
rs781863415 Chr.7: 102425126-102425126 on Build GRCh38
rs140002963 Chr.7: 102425053-102425053 on Build GRCh38
rs782093435 Chr.7: 102424936-102424936 on Build GRCh38
rs782176976 Chr.7: 102425144-102425144 on Build GRCh38
rs376084022 Chr.7: 102424974-102424974 on Build GRCh38
rs146092398 Chr.7: 102425018-102425018 on Build GRCh38
rs782540550 Chr.7: 102425128-102425128 on Build GRCh38
rs781862435 Chr.7: 102425023-102425023 on Build GRCh38
rs782113976 Chr.7: 102424988-102424988 on Build GRCh38
rs377032265 Chr.7: 102425026-102425026 on Build GRCh38
rs781961773 Chr.7: 102424986-102424986 on Build GRCh38
rs782646178 Chr.7: 102425061-102425061 on Build GRCh38
rs931369482 Chr.7: 102425110-102425110 on Build GRCh38

View Full Product Details