Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75053521-75053774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
UniGene
Hs.370480
Gene Chromosome Location
Chr.X: 75053172-75156340 on Build GRCh38
Transcripts
NM_001271696.1 NM_001271697.1 NM_001271698.1 NM_001271699.1 NM_004299.4

SNP Detail

SNP ID Chromosome Location
rs771613456 Chr.X: 75053611-75053611 on Build GRCh38
rs187151229 Chr.X: 75053666-75053666 on Build GRCh38
rs1051322210 Chr.X: 75053613-75053613 on Build GRCh38
rs897449802 Chr.X: 75053570-75053570 on Build GRCh38
rs797044558 Chr.X: 75053585-75053585 on Build GRCh38
rs1008541321 Chr.X: 75053669-75053669 on Build GRCh38
rs778556688 Chr.X: 75053593-75053593 on Build GRCh38
rs749045558 Chr.X: 75053675-75053675 on Build GRCh38
rs746271411 Chr.X: 75053559-75053559 on Build GRCh38
rs936800740 Chr.X: 75053574-75053574 on Build GRCh38
rs201312449 Chr.X: 75053573-75053573 on Build GRCh38
rs142425608 Chr.X: 75053609-75053609 on Build GRCh38
rs377712763 Chr.X: 75053612-75053612 on Build GRCh38
rs781524899 Chr.X: 75053556-75053556 on Build GRCh38
rs964445745 Chr.X: 75053685-75053685 on Build GRCh38
rs758369796 Chr.X: 75053741-75053741 on Build GRCh38
rs1017380220 Chr.X: 75053676-75053676 on Build GRCh38
rs767503863 Chr.X: 75053748-75053748 on Build GRCh38
rs769374153 Chr.X: 75053619-75053619 on Build GRCh38
rs768257406 Chr.X: 75053563-75053563 on Build GRCh38
rs771037745 Chr.X: 75053628-75053628 on Build GRCh38

View Full Product Details