Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5969152-5969425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWF
Entrez Gene ID
7450
Gene Name
von Willebrand factor
Gene Aliases
F8VWF, VWD
UniGene
Hs.440848
Gene Chromosome Location
Chr.12: 5948874-6124675 on Build GRCh38
Transcripts
NM_000552.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4740700 c.7721G>A p.C2574Y Chr.12: 5969219-5969219 on Build GRCh38
3872219 c.7662C>T p.V2554V Chr.12: 5969278-5969278 on Build GRCh38
694881 c.7676C>T p.S2559L Chr.12: 5969264-5969264 on Build GRCh38
1232473 c.7679G>A p.G2560D Chr.12: 5969261-5969261 on Build GRCh38
4044015 c.7685A>C p.Q2562P Chr.12: 5969255-5969255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61751297 Chr.12: 5969310-5969310 on Build GRCh38
rs368810263 Chr.12: 5969226-5969226 on Build GRCh38
rs926673879 Chr.12: 5969235-5969235 on Build GRCh38
rs771222658 Chr.12: 5969394-5969394 on Build GRCh38
rs772773846 Chr.12: 5969400-5969400 on Build GRCh38
rs752093211 Chr.12: 5969234-5969234 on Build GRCh38
rs779932835 Chr.12: 5969362-5969362 on Build GRCh38
rs758376829 Chr.12: 5969361-5969361 on Build GRCh38
rs942107186 Chr.12: 5969239-5969239 on Build GRCh38
rs948600047 Chr.12: 5969290-5969290 on Build GRCh38
rs16932285 Chr.12: 5969242-5969242 on Build GRCh38
rs137987906 Chr.12: 5969336-5969336 on Build GRCh38
rs760277526 Chr.12: 5969210-5969210 on Build GRCh38
rs763283384 Chr.12: 5969301-5969301 on Build GRCh38
rs767664790 Chr.12: 5969250-5969250 on Build GRCh38
rs373158962 Chr.12: 5969267-5969267 on Build GRCh38
rs771516504 Chr.12: 5969199-5969199 on Build GRCh38
rs376366370 Chr.12: 5969291-5969291 on Build GRCh38
rs546185888 Chr.12: 5969263-5969263 on Build GRCh38
rs143235468 Chr.12: 5969233-5969233 on Build GRCh38
rs562305031 Chr.12: 5969368-5969368 on Build GRCh38
rs780588032 Chr.12: 5969302-5969302 on Build GRCh38
rs757011174 Chr.12: 5969348-5969348 on Build GRCh38
rs568696523 Chr.12: 5969193-5969193 on Build GRCh38
rs61751294 Chr.12: 5969381-5969381 on Build GRCh38
rs890420023 Chr.12: 5969281-5969281 on Build GRCh38
rs267607364 Chr.12: 5969275-5969276 on Build GRCh38
rs1009015592 Chr.12: 5969370-5969370 on Build GRCh38
rs373321657 Chr.12: 5969216-5969216 on Build GRCh38
rs61751293 Chr.12: 5969388-5969388 on Build GRCh38
rs918057829 Chr.12: 5969206-5969206 on Build GRCh38
rs1025645485 Chr.12: 5969376-5969376 on Build GRCh38
rs202068340 Chr.12: 5969268-5969268 on Build GRCh38
rs61751300 Chr.12: 5969257-5969257 on Build GRCh38
rs546231736 Chr.12: 5969177-5969177 on Build GRCh38
rs749106300 Chr.12: 5969262-5969262 on Build GRCh38
rs774828008 Chr.12: 5969395-5969395 on Build GRCh38
rs199941643 Chr.12: 5969178-5969178 on Build GRCh38
rs192262780 Chr.12: 5969221-5969221 on Build GRCh38
rs150778949 Chr.12: 5969321-5969321 on Build GRCh38
rs61751296 Chr.12: 5969337-5969337 on Build GRCh38
rs369734683 Chr.12: 5969212-5969212 on Build GRCh38
rs759949578 Chr.12: 5969397-5969397 on Build GRCh38
rs369878547 Chr.12: 5969350-5969350 on Build GRCh38
rs372582651 Chr.12: 5969274-5969274 on Build GRCh38
rs61751298 Chr.12: 5969304-5969304 on Build GRCh38
rs374690023 Chr.12: 5969369-5969369 on Build GRCh38
rs752991654 Chr.12: 5969257-5969257 on Build GRCh38
rs61751295 Chr.12: 5969341-5969341 on Build GRCh38
rs1018153988 Chr.12: 5969371-5969371 on Build GRCh38
rs758747918 Chr.12: 5969260-5969260 on Build GRCh38
rs754037205 Chr.12: 5969340-5969340 on Build GRCh38
rs375991463 Chr.12: 5969217-5969217 on Build GRCh38
rs535167710 Chr.12: 5969179-5969179 on Build GRCh38
rs201213029 Chr.12: 5969315-5969315 on Build GRCh38
rs915754316 Chr.12: 5969276-5969276 on Build GRCh38
rs267607365 Chr.12: 5969265-5969266 on Build GRCh38
rs774929265 Chr.12: 5969270-5969270 on Build GRCh38
rs532963814 Chr.12: 5969316-5969316 on Build GRCh38
rs752865231 Chr.12: 5969324-5969324 on Build GRCh38
rs61751301 Chr.12: 5969204-5969204 on Build GRCh38
rs774724224 Chr.12: 5969311-5969311 on Build GRCh38
rs971026321 Chr.12: 5969387-5969387 on Build GRCh38
rs555564965 Chr.12: 5969184-5969184 on Build GRCh38
rs868760790 Chr.12: 5969320-5969320 on Build GRCh38
rs778732929 Chr.12: 5969264-5969264 on Build GRCh38
rs964948564 Chr.12: 5969201-5969201 on Build GRCh38
rs35335161 Chr.12: 5969258-5969258 on Build GRCh38
rs775123891 Chr.12: 5969203-5969203 on Build GRCh38
rs768335080 Chr.12: 5969284-5969284 on Build GRCh38
rs776340794 Chr.12: 5969289-5969289 on Build GRCh38
rs145955543 Chr.12: 5969389-5969389 on Build GRCh38

View Full Product Details