Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15989623-15989896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROM1
Entrez Gene ID
8842
Gene Name
prominin 1
Gene Aliases
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
UniGene
Hs.614734
Gene Chromosome Location
Chr.4: 15968226-16084100 on Build GRCh38
Transcripts
NM_001145847.1 NM_001145848.1 NM_001145849.1 NM_001145850.1 NM_001145851.1 NM_001145852.1 NM_006017.2 XM_005248195.4 XM_005248196.4 XM_006713974.3 XM_011513890.1 XM_011513892.2 XM_011513893.2 XM_011513894.2 XM_011513895.2 XM_011513896.2 XM_011513897.2 XM_011513898.2 XM_011513899.2 XM_011513900.2 XM_011513902.2 XM_011513903.2 XM_017008799.1 XM_017008800.1 XM_017008801.1 XM_017008802.1 XM_017008803.1 XM_017008804.1 XM_017008805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4815045 c.2032C>G p.P678A Chr.4: 15989749-15989749 on Build GRCh38
4946857 c.2016C>A p.H672Q Chr.4: 15989765-15989765 on Build GRCh38
5400726 c.1990G>A p.G664R Chr.4: 15989818-15989818 on Build GRCh38
4533801 c.1990G>A p.D664N Chr.4: 15989791-15989791 on Build GRCh38
4533800 c.2017G>A p.D673N Chr.4: 15989791-15989791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030358975 Chr.4: 15989876-15989876 on Build GRCh38
rs568290297 Chr.4: 15989728-15989728 on Build GRCh38
rs938627050 Chr.4: 15989807-15989807 on Build GRCh38
rs374219790 Chr.4: 15989716-15989716 on Build GRCh38
rs903299693 Chr.4: 15989851-15989851 on Build GRCh38
rs373680665 Chr.4: 15989825-15989825 on Build GRCh38
rs774758302 Chr.4: 15989645-15989646 on Build GRCh38
rs1047895041 Chr.4: 15989794-15989794 on Build GRCh38
rs919646653 Chr.4: 15989669-15989669 on Build GRCh38
rs974247212 Chr.4: 15989656-15989656 on Build GRCh38
rs753006062 Chr.4: 15989765-15989765 on Build GRCh38
rs763243562 Chr.4: 15989783-15989783 on Build GRCh38
rs894328432 Chr.4: 15989822-15989822 on Build GRCh38
rs771612815 Chr.4: 15989721-15989721 on Build GRCh38
rs545789716 Chr.4: 15989691-15989691 on Build GRCh38
rs559085602 Chr.4: 15989698-15989698 on Build GRCh38
rs867949539 Chr.4: 15989679-15989679 on Build GRCh38
rs760554830 Chr.4: 15989746-15989746 on Build GRCh38
rs757730805 Chr.4: 15989713-15989713 on Build GRCh38
rs770861575 Chr.4: 15989757-15989757 on Build GRCh38
rs548197663 Chr.4: 15989717-15989717 on Build GRCh38
rs377712580 Chr.4: 15989792-15989792 on Build GRCh38
rs371616315 Chr.4: 15989819-15989819 on Build GRCh38
rs999340623 Chr.4: 15989874-15989874 on Build GRCh38
rs781679763 Chr.4: 15989802-15989802 on Build GRCh38
rs759432430 Chr.4: 15989759-15989759 on Build GRCh38
rs909386255 Chr.4: 15989836-15989836 on Build GRCh38
rs781437553 Chr.4: 15989715-15989715 on Build GRCh38
rs750885395 Chr.4: 15989813-15989813 on Build GRCh38
rs377581515 Chr.4: 15989702-15989702 on Build GRCh38
rs550762397 Chr.4: 15989857-15989857 on Build GRCh38
rs765118929 Chr.4: 15989763-15989763 on Build GRCh38
rs763990524 Chr.4: 15989806-15989806 on Build GRCh38
rs762880254 Chr.4: 15989798-15989798 on Build GRCh38
rs777181097 Chr.4: 15989847-15989847 on Build GRCh38
rs775321230 Chr.4: 15989788-15989788 on Build GRCh38
rs528429658 Chr.4: 15989719-15989719 on Build GRCh38
rs774648298 Chr.4: 15989705-15989705 on Build GRCh38
rs200356026 Chr.4: 15989720-15989720 on Build GRCh38
rs2240685 Chr.4: 15989652-15989652 on Build GRCh38
rs780368576 Chr.4: 15989821-15989821 on Build GRCh38
rs369480629 Chr.4: 15989703-15989703 on Build GRCh38
rs530749007 Chr.4: 15989758-15989758 on Build GRCh38
rs373247019 Chr.4: 15989683-15989683 on Build GRCh38
rs772757789 Chr.4: 15989730-15989730 on Build GRCh38
rs757998973 Chr.4: 15989838-15989838 on Build GRCh38
rs929639559 Chr.4: 15989672-15989672 on Build GRCh38

View Full Product Details