Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19975378-19975639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB5A
Entrez Gene ID
5868
Gene Name
RAB5A, member RAS oncogene family
Gene Aliases
RAB5
UniGene
Hs.475663
Gene Chromosome Location
Chr.3: 19947080-19985175 on Build GRCh38
Transcripts
NM_001292048.1 NM_004162.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5939149 c.176C>T p.T59I Chr.3: 19975613-19975613 on Build GRCh38
5665202 c.188G>A p.C63Y Chr.3: 19975625-19975625 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943719686 Chr.3: 19975431-19975435 on Build GRCh38
rs927439828 Chr.3: 19975478-19975478 on Build GRCh38
rs938861648 Chr.3: 19975488-19975488 on Build GRCh38
rs556175633 Chr.3: 19975564-19975564 on Build GRCh38
rs751242785 Chr.3: 19975481-19975481 on Build GRCh38
rs183956916 Chr.3: 19975413-19975413 on Build GRCh38
rs956667019 Chr.3: 19975417-19975417 on Build GRCh38
rs773988797 Chr.3: 19975563-19975563 on Build GRCh38
rs899201015 Chr.3: 19975523-19975523 on Build GRCh38
rs758402113 Chr.3: 19975594-19975594 on Build GRCh38
rs537255253 Chr.3: 19975542-19975542 on Build GRCh38
rs767141189 Chr.3: 19975565-19975565 on Build GRCh38
rs374443689 Chr.3: 19975567-19975567 on Build GRCh38
rs918281574 Chr.3: 19975535-19975535 on Build GRCh38
rs756870845 Chr.3: 19975495-19975495 on Build GRCh38
rs779489579 Chr.3: 19975531-19975531 on Build GRCh38
rs554887382 Chr.3: 19975468-19975468 on Build GRCh38
rs752507655 Chr.3: 19975438-19975438 on Build GRCh38
rs116665507 Chr.3: 19975444-19975444 on Build GRCh38
rs747201801 Chr.3: 19975609-19975609 on Build GRCh38
rs755514870 Chr.3: 19975529-19975529 on Build GRCh38
rs770844601 Chr.3: 19975551-19975551 on Build GRCh38
rs368852137 Chr.3: 19975581-19975581 on Build GRCh38
rs765790302 Chr.3: 19975587-19975587 on Build GRCh38
rs70937042 Chr.3: 19975558-19975558 on Build GRCh38
rs530787211 Chr.3: 19975447-19975447 on Build GRCh38
rs569903967 Chr.3: 19975522-19975522 on Build GRCh38
rs760268912 Chr.3: 19975570-19975570 on Build GRCh38
rs1038065037 Chr.3: 19975508-19975508 on Build GRCh38
rs201173280 Chr.3: 19975435-19975436 on Build GRCh38
rs780738647 Chr.3: 19975496-19975496 on Build GRCh38
rs1021970036 Chr.3: 19975442-19975442 on Build GRCh38
rs1035949780 Chr.3: 19975487-19975487 on Build GRCh38
rs764495920 Chr.3: 19975580-19975580 on Build GRCh38
rs780136538 Chr.3: 19975598-19975598 on Build GRCh38
rs750938095 Chr.3: 19975592-19975592 on Build GRCh38
rs763745331 Chr.3: 19975463-19975463 on Build GRCh38
rs896928908 Chr.3: 19975436-19975436 on Build GRCh38
rs772932206 Chr.3: 19975537-19975537 on Build GRCh38
rs11917387 Chr.3: 19975424-19975424 on Build GRCh38
rs940958549 Chr.3: 19975499-19975499 on Build GRCh38
rs757777830 Chr.3: 19975585-19975585 on Build GRCh38
rs1002109452 Chr.3: 19975444-19975445 on Build GRCh38
rs968685556 Chr.3: 19975504-19975505 on Build GRCh38
rs749930489 Chr.3: 19975520-19975520 on Build GRCh38
rs748557365 Chr.3: 19975532-19975532 on Build GRCh38

View Full Product Details