Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14443431-14443664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKN1
Entrez Gene ID
5585
Gene Name
protein kinase N1
Gene Aliases
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1
UniGene
Hs.466044
Gene Chromosome Location
Chr.19: 14433354-14471867 on Build GRCh38
Transcripts
NM_002741.3 NM_213560.1 XM_011528128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4074806 c.382G>A p.V128M Chr.19: 14443529-14443529 on Build GRCh38
4074808 c.429G>A p.Q143Q Chr.19: 14443576-14443576 on Build GRCh38
991882 c.434C>T p.A145V Chr.19: 14443581-14443581 on Build GRCh38
5092904 c.344C>T p.A115V Chr.19: 14443491-14443491 on Build GRCh38
4630418 c.383C>T p.A128V Chr.19: 14443512-14443512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553669739 Chr.19: 14443634-14443634 on Build GRCh38
rs772254067 Chr.19: 14443558-14443558 on Build GRCh38
rs374744301 Chr.19: 14443529-14443529 on Build GRCh38
rs760992040 Chr.19: 14443509-14443509 on Build GRCh38
rs768919457 Chr.19: 14443483-14443483 on Build GRCh38
rs770604828 Chr.19: 14443455-14443455 on Build GRCh38
rs111523593 Chr.19: 14443623-14443623 on Build GRCh38
rs2161287 Chr.19: 14443534-14443534 on Build GRCh38
rs146198925 Chr.19: 14443492-14443492 on Build GRCh38
rs760067248 Chr.19: 14443589-14443589 on Build GRCh38
rs190031884 Chr.19: 14443461-14443461 on Build GRCh38
rs776987786 Chr.19: 14443588-14443588 on Build GRCh38
rs769391410 Chr.19: 14443581-14443581 on Build GRCh38
rs887718207 Chr.19: 14443462-14443462 on Build GRCh38
rs368601143 Chr.19: 14443510-14443510 on Build GRCh38
rs112824233 Chr.19: 14443528-14443528 on Build GRCh38
rs368909367 Chr.19: 14443533-14443533 on Build GRCh38
rs113604339 Chr.19: 14443624-14443624 on Build GRCh38
rs55814382 Chr.19: 14443582-14443582 on Build GRCh38
rs780875275 Chr.19: 14443526-14443526 on Build GRCh38
rs748187817 Chr.19: 14443553-14443553 on Build GRCh38
rs756926174 Chr.19: 14443520-14443520 on Build GRCh38
rs775956072 Chr.19: 14443603-14443603 on Build GRCh38
rs767012283 Chr.19: 14443631-14443631 on Build GRCh38
rs763599094 Chr.19: 14443514-14443514 on Build GRCh38
rs771257533 Chr.19: 14443584-14443584 on Build GRCh38
rs372099148 Chr.19: 14443527-14443527 on Build GRCh38
rs756836242 Chr.19: 14443457-14443457 on Build GRCh38
rs761354959 Chr.19: 14443618-14443618 on Build GRCh38
rs375594823 Chr.19: 14443466-14443466 on Build GRCh38
rs765685832 Chr.19: 14443600-14443600 on Build GRCh38
rs774510114 Chr.19: 14443485-14443485 on Build GRCh38
rs771387594 Chr.19: 14443477-14443477 on Build GRCh38
rs368926924 Chr.19: 14443633-14443633 on Build GRCh38
rs751226464 Chr.19: 14443450-14443450 on Build GRCh38
rs760030924 Chr.19: 14443513-14443513 on Build GRCh38
rs371928270 Chr.19: 14443491-14443491 on Build GRCh38
rs751031917 Chr.19: 14443518-14443518 on Build GRCh38
rs992081434 Chr.19: 14443561-14443561 on Build GRCh38
rs769613338 Chr.19: 14443470-14443470 on Build GRCh38
rs758042106 Chr.19: 14443644-14443644 on Build GRCh38
rs749247875 Chr.19: 14443482-14443482 on Build GRCh38

View Full Product Details