Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5063511-5063774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBAK
Entrez Gene ID
57786
Gene Name
RB associated KRAB zinc finger
Gene Aliases
ZNF769
UniGene
Hs.396178
Gene Chromosome Location
Chr.7: 5045821-5069488 on Build GRCh38
Transcripts
NM_001204456.1 NM_021163.3 NM_001204513.1

Gene Symbol
RBAK-RBAKDN
Entrez Gene ID
100533952
Gene Name
RBAK-RBAKDN readthrough
Gene Aliases
RBAK-LOC389458
UniGene
Hs.396178
Gene Chromosome Location
Chr.7: 5045821-5073223 on Build GRCh38
Transcripts
NM_001204456.1 NM_021163.3 NM_001204513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1090671 c.269G>T p.R90I Chr.7: 5063725-5063725 on Build GRCh38
1451356 c.295T>G p.S99A Chr.7: 5063751-5063751 on Build GRCh38
746556 c.302A>T p.Q101L Chr.7: 5063758-5063758 on Build GRCh38
80428 c.270A>G p.R90R Chr.7: 5063726-5063726 on Build GRCh38
3431678 c.283G>A p.E95K Chr.7: 5063739-5063739 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747889306 Chr.7: 5063688-5063688 on Build GRCh38
rs991660725 Chr.7: 5063571-5063571 on Build GRCh38
rs776761866 Chr.7: 5063704-5063704 on Build GRCh38
rs994722941 Chr.7: 5063611-5063611 on Build GRCh38
rs767733077 Chr.7: 5063746-5063746 on Build GRCh38
rs909356837 Chr.7: 5063586-5063586 on Build GRCh38
rs779488743 Chr.7: 5063674-5063674 on Build GRCh38
rs771703038 Chr.7: 5063691-5063691 on Build GRCh38
rs972546750 Chr.7: 5063615-5063615 on Build GRCh38
rs778529872 Chr.7: 5063685-5063685 on Build GRCh38
rs574149729 Chr.7: 5063561-5063561 on Build GRCh38
rs760493837 Chr.7: 5063702-5063702 on Build GRCh38
rs1008962656 Chr.7: 5063578-5063578 on Build GRCh38
rs980528728 Chr.7: 5063687-5063687 on Build GRCh38
rs187814441 Chr.7: 5063564-5063564 on Build GRCh38
rs1005002406 Chr.7: 5063646-5063646 on Build GRCh38
rs773067555 Chr.7: 5063692-5063692 on Build GRCh38
rs371950301 Chr.7: 5063738-5063738 on Build GRCh38
rs377226599 Chr.7: 5063681-5063681 on Build GRCh38
rs559882097 Chr.7: 5063630-5063630 on Build GRCh38
rs900435591 Chr.7: 5063607-5063607 on Build GRCh38
rs771017316 Chr.7: 5063703-5063703 on Build GRCh38
rs762109169 Chr.7: 5063744-5063744 on Build GRCh38
rs369934466 Chr.7: 5063613-5063613 on Build GRCh38
rs764218744 Chr.7: 5063676-5063676 on Build GRCh38
rs367915306 Chr.7: 5063633-5063633 on Build GRCh38
rs748771611 Chr.7: 5063680-5063680 on Build GRCh38
rs764338049 Chr.7: 5063723-5063723 on Build GRCh38

View Full Product Details