Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33279645-33279884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR46
Entrez Gene ID
9277
Gene Name
WD repeat domain 46
Gene Aliases
BING4, C6orf11, FP221, UTP7
UniGene
Hs.520063
Gene Chromosome Location
Chr.6: 33278850-33289527 on Build GRCh38
Transcripts
NM_001164267.1 NM_005452.5 XM_017011484.1 XM_017011485.1 XM_017011486.1 XM_017011487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
451297 c.1581C>T p.I527I Chr.6: 33279803-33279803 on Build GRCh38
6847810 c.1579A>G p.I527V Chr.6: 33279805-33279805 on Build GRCh38
395334 c.1620+2T>G p.? Chr.6: 33279762-33279762 on Build GRCh38
1196844 c.1556G>A p.R519Q Chr.6: 33279828-33279828 on Build GRCh38
3873134 c.1588G>A p.E530K Chr.6: 33279796-33279796 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200631390 Chr.6: 33279849-33279849 on Build GRCh38
rs776842044 Chr.6: 33279762-33279762 on Build GRCh38
rs762766693 Chr.6: 33279739-33279739 on Build GRCh38
rs376184361 Chr.6: 33279732-33279732 on Build GRCh38
rs879276643 Chr.6: 33279725-33279725 on Build GRCh38
rs758701039 Chr.6: 33279789-33279789 on Build GRCh38
rs766439702 Chr.6: 33279760-33279760 on Build GRCh38
rs1021118733 Chr.6: 33279748-33279748 on Build GRCh38
rs748713597 Chr.6: 33279823-33279823 on Build GRCh38
rs370710495 Chr.6: 33279751-33279751 on Build GRCh38
rs755579192 Chr.6: 33279825-33279825 on Build GRCh38
rs759888424 Chr.6: 33279767-33279767 on Build GRCh38
rs775633418 Chr.6: 33279846-33279846 on Build GRCh38
rs761743806 Chr.6: 33279752-33279752 on Build GRCh38
rs142850284 Chr.6: 33279829-33279829 on Build GRCh38
rs768450555 Chr.6: 33279742-33279742 on Build GRCh38
rs72654497 Chr.6: 33279818-33279818 on Build GRCh38
rs752068623 Chr.6: 33279817-33279817 on Build GRCh38
rs772459571 Chr.6: 33279828-33279828 on Build GRCh38
rs201671590 Chr.6: 33279850-33279850 on Build GRCh38
rs549701826 Chr.6: 33279688-33279688 on Build GRCh38
rs752915965 Chr.6: 33279784-33279784 on Build GRCh38
rs765528166 Chr.6: 33279782-33279782 on Build GRCh38
rs748615780 Chr.6: 33279761-33279761 on Build GRCh38
rs1002222812 Chr.6: 33279683-33279683 on Build GRCh38
rs764448741 Chr.6: 33279806-33279806 on Build GRCh38
rs528507375 Chr.6: 33279733-33279733 on Build GRCh38
rs749180414 Chr.6: 33279809-33279809 on Build GRCh38
rs779705594 Chr.6: 33279826-33279826 on Build GRCh38
rs960934342 Chr.6: 33279692-33279692 on Build GRCh38
rs774193035 Chr.6: 33279840-33279840 on Build GRCh38

View Full Product Details