Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4818123-4818350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TICAM1
Entrez Gene ID
148022
Gene Name
toll like receptor adaptor molecule 1
Gene Aliases
IIAE6, MyD88-3, PRVTIRB, TICAM-1, TRIF
UniGene
Hs.29344
Gene Chromosome Location
Chr.19: 4815924-4831742 on Build GRCh38
Transcripts
NM_182919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995940 c.38A>G p.D13G Chr.19: 4818340-4818340 on Build GRCh38
4284926 c.116G>C p.G39A Chr.19: 4818262-4818262 on Build GRCh38
4424311 c.230G>A p.W77* Chr.19: 4818148-4818148 on Build GRCh38
6801743 c.133G>T p.A45S Chr.19: 4818245-4818245 on Build GRCh38
1680532 c.173G>T p.R58M Chr.19: 4818205-4818205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758613862 Chr.19: 4818168-4818168 on Build GRCh38
rs765301499 Chr.19: 4818256-4818256 on Build GRCh38
rs746146801 Chr.19: 4818167-4818167 on Build GRCh38
rs768957889 Chr.19: 4818327-4818327 on Build GRCh38
rs147427321 Chr.19: 4818176-4818176 on Build GRCh38
rs764485662 Chr.19: 4818234-4818234 on Build GRCh38
rs749244272 Chr.19: 4818306-4818306 on Build GRCh38
rs113493978 Chr.19: 4818321-4818321 on Build GRCh38
rs769794539 Chr.19: 4818175-4818175 on Build GRCh38
rs11466719 Chr.19: 4818155-4818155 on Build GRCh38
rs370301428 Chr.19: 4818141-4818141 on Build GRCh38
rs747970296 Chr.19: 4818280-4818280 on Build GRCh38
rs201616584 Chr.19: 4818288-4818288 on Build GRCh38
rs139810575 Chr.19: 4818195-4818195 on Build GRCh38
rs760228606 Chr.19: 4818225-4818225 on Build GRCh38
rs753040804 Chr.19: 4818147-4818147 on Build GRCh38
rs199816697 Chr.19: 4818140-4818140 on Build GRCh38
rs948956807 Chr.19: 4818224-4818225 on Build GRCh38
rs376605000 Chr.19: 4818174-4818174 on Build GRCh38
rs372818181 Chr.19: 4818166-4818166 on Build GRCh38
rs878979221 Chr.19: 4818207-4818207 on Build GRCh38
rs182370758 Chr.19: 4818188-4818188 on Build GRCh38
rs996988678 Chr.19: 4818227-4818228 on Build GRCh38
rs79591246 Chr.19: 4818283-4818283 on Build GRCh38
rs935231020 Chr.19: 4818143-4818143 on Build GRCh38
rs745665530 Chr.19: 4818222-4818222 on Build GRCh38
rs761078466 Chr.19: 4818227-4818227 on Build GRCh38
rs745319253 Chr.19: 4818316-4818316 on Build GRCh38
rs754350473 Chr.19: 4818243-4818243 on Build GRCh38
rs756698093 Chr.19: 4818151-4818151 on Build GRCh38
rs371101914 Chr.19: 4818180-4818180 on Build GRCh38
rs760351659 Chr.19: 4818325-4818325 on Build GRCh38
rs780135844 Chr.19: 4818154-4818165 on Build GRCh38
rs142713853 Chr.19: 4818165-4818165 on Build GRCh38
rs780201394 Chr.19: 4818278-4818278 on Build GRCh38
rs375571285 Chr.19: 4818244-4818244 on Build GRCh38
rs770902111 Chr.19: 4818308-4818308 on Build GRCh38
rs539505019 Chr.19: 4818270-4818270 on Build GRCh38
rs201782115 Chr.19: 4818199-4818199 on Build GRCh38
rs758634091 Chr.19: 4818277-4818277 on Build GRCh38
rs149930656 Chr.19: 4818197-4818197 on Build GRCh38
rs774006618 Chr.19: 4818309-4818309 on Build GRCh38

View Full Product Details