Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28655252-28655525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDF2
Entrez Gene ID
6388
Gene Name
stromal cell derived factor 2
Gene Aliases
-
UniGene
Hs.514036
Gene Chromosome Location
Chr.17: 28648356-28662189 on Build GRCh38
Transcripts
NM_006923.3 NR_045585.1 XM_011525106.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2800643 c.278G>A p.R93Q Chr.17: 28655357-28655357 on Build GRCh38
5352088 c.152-6C>G p.? Chr.17: 28655489-28655489 on Build GRCh38
6773729 c.217C>T p.R73W Chr.17: 28655418-28655418 on Build GRCh38
4169195 c.218G>A p.R73Q Chr.17: 28655417-28655417 on Build GRCh38
5963988 c.152-7A>G p.? Chr.17: 28655490-28655490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767924184 Chr.17: 28655503-28655503 on Build GRCh38
rs972676207 Chr.17: 28655365-28655365 on Build GRCh38
rs942284192 Chr.17: 28655347-28655347 on Build GRCh38
rs929731792 Chr.17: 28655390-28655391 on Build GRCh38
rs778547628 Chr.17: 28655358-28655358 on Build GRCh38
rs769231523 Chr.17: 28655438-28655438 on Build GRCh38
rs776810897 Chr.17: 28655283-28655283 on Build GRCh38
rs979873048 Chr.17: 28655272-28655272 on Build GRCh38
rs746830583 Chr.17: 28655386-28655386 on Build GRCh38
rs979926878 Chr.17: 28655324-28655324 on Build GRCh38
rs11542345 Chr.17: 28655313-28655313 on Build GRCh38
rs752542351 Chr.17: 28655359-28655359 on Build GRCh38
rs762833961 Chr.17: 28655464-28655476 on Build GRCh38
rs770279086 Chr.17: 28655315-28655315 on Build GRCh38
rs746151655 Chr.17: 28655401-28655401 on Build GRCh38
rs1050769710 Chr.17: 28655418-28655418 on Build GRCh38
rs368337665 Chr.17: 28655495-28655495 on Build GRCh38
rs34038517 Chr.17: 28655336-28655336 on Build GRCh38
rs62066799 Chr.17: 28655390-28655390 on Build GRCh38
rs758409844 Chr.17: 28655369-28655369 on Build GRCh38
rs775680467 Chr.17: 28655404-28655404 on Build GRCh38
rs376355455 Chr.17: 28655353-28655353 on Build GRCh38
rs775848807 Chr.17: 28655317-28655317 on Build GRCh38
rs769518864 Chr.17: 28655304-28655308 on Build GRCh38
rs773151905 Chr.17: 28655456-28655457 on Build GRCh38
rs766054367 Chr.17: 28655348-28655348 on Build GRCh38
rs967488606 Chr.17: 28655286-28655286 on Build GRCh38
rs752008136 Chr.17: 28655338-28655338 on Build GRCh38
rs956898560 Chr.17: 28655293-28655293 on Build GRCh38
rs919423952 Chr.17: 28655366-28655366 on Build GRCh38
rs763351641 Chr.17: 28655417-28655417 on Build GRCh38
rs547468803 Chr.17: 28655375-28655375 on Build GRCh38
rs771299392 Chr.17: 28655273-28655273 on Build GRCh38
rs777381395 Chr.17: 28655379-28655379 on Build GRCh38
rs760275449 Chr.17: 28655341-28655341 on Build GRCh38
rs371703183 Chr.17: 28655284-28655284 on Build GRCh38
rs143512042 Chr.17: 28655303-28655303 on Build GRCh38
rs774661692 Chr.17: 28655451-28655451 on Build GRCh38
rs754468031 Chr.17: 28655357-28655357 on Build GRCh38
rs781581029 Chr.17: 28655398-28655398 on Build GRCh38
rs35579310 Chr.17: 28655323-28655323 on Build GRCh38
rs200127951 Chr.17: 28655442-28655442 on Build GRCh38

View Full Product Details