Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 147204638-147204871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STXBP5
Entrez Gene ID
134957
Gene Name
syntaxin binding protein 5
Gene Aliases
LGL3, LLGL3, Nbla04300
UniGene
Hs.736439
Gene Chromosome Location
Chr.6: 147204184-147390476 on Build GRCh38
Transcripts
NM_001127715.2 NM_139244.4 XM_006715339.2 XM_017010268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5582662 c.115G>A p.E39K Chr.6: 147204647-147204647 on Build GRCh38
6305496 c.113A>G p.Q38R Chr.6: 147204645-147204645 on Build GRCh38
3829261 c.144C>T p.L48L Chr.6: 147204676-147204676 on Build GRCh38
1074333 c.147C>T p.C49C Chr.6: 147204679-147204679 on Build GRCh38
3829262 c.144C>T p.L48L Chr.6: 147204676-147204676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045199674 Chr.6: 147204731-147204731 on Build GRCh38
rs757159776 Chr.6: 147204667-147204667 on Build GRCh38
rs1054527815 Chr.6: 147204818-147204818 on Build GRCh38
rs374854337 Chr.6: 147204804-147204804 on Build GRCh38
rs756151736 Chr.6: 147204701-147204701 on Build GRCh38
rs553098369 Chr.6: 147204795-147204795 on Build GRCh38
rs201371480 Chr.6: 147204703-147204703 on Build GRCh38
rs763941350 Chr.6: 147204664-147204664 on Build GRCh38
rs894250236 Chr.6: 147204825-147204825 on Build GRCh38
rs750438948 Chr.6: 147204697-147204697 on Build GRCh38
rs999899454 Chr.6: 147204810-147204810 on Build GRCh38
rs540128146 Chr.6: 147204840-147204840 on Build GRCh38
rs932708618 Chr.6: 147204721-147204721 on Build GRCh38
rs760465930 Chr.6: 147204729-147204731 on Build GRCh38
rs890299176 Chr.6: 147204758-147204758 on Build GRCh38
rs944518513 Chr.6: 147204773-147204777 on Build GRCh38
rs760123944 Chr.6: 147204724-147204724 on Build GRCh38
rs779184684 Chr.6: 147204732-147204732 on Build GRCh38
rs942044741 Chr.6: 147204831-147204831 on Build GRCh38
rs1050249719 Chr.6: 147204728-147204728 on Build GRCh38
rs41311404 Chr.6: 147204710-147204710 on Build GRCh38
rs767340630 Chr.6: 147204676-147204676 on Build GRCh38
rs751314989 Chr.6: 147204666-147204666 on Build GRCh38
rs752203626 Chr.6: 147204661-147204661 on Build GRCh38
rs956651147 Chr.6: 147204688-147204688 on Build GRCh38
rs926597091 Chr.6: 147204786-147204786 on Build GRCh38
rs770894319 Chr.6: 147204706-147204706 on Build GRCh38
rs1041618786 Chr.6: 147204785-147204790 on Build GRCh38
rs867715866 Chr.6: 147204798-147204798 on Build GRCh38
rs749466897 Chr.6: 147204709-147204709 on Build GRCh38
rs903065711 Chr.6: 147204802-147204803 on Build GRCh38

View Full Product Details