Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108862261-108862532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL7A
Entrez Gene ID
10881
Gene Name
actin like 7A
Gene Aliases
-
UniGene
Hs.123530
Gene Chromosome Location
Chr.9: 108862228-108863755 on Build GRCh38
Transcripts
NM_006687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242796 c.132C>A p.V44V Chr.9: 108862454-108862454 on Build GRCh38
364837 c.45G>C p.K15N Chr.9: 108862367-108862367 on Build GRCh38
5766647 c.14C>A p.P5Q Chr.9: 108862336-108862336 on Build GRCh38
3652699 c.28G>A p.G10R Chr.9: 108862350-108862350 on Build GRCh38
271086 c.191C>A p.P64H Chr.9: 108862513-108862513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766550736 Chr.9: 108862456-108862456 on Build GRCh38
rs149755351 Chr.9: 108862432-108862432 on Build GRCh38
rs377067877 Chr.9: 108862501-108862501 on Build GRCh38
rs748934579 Chr.9: 108862441-108862441 on Build GRCh38
rs753663933 Chr.9: 108862452-108862452 on Build GRCh38
rs533177437 Chr.9: 108862410-108862410 on Build GRCh38
rs779143807 Chr.9: 108862309-108862309 on Build GRCh38
rs780011699 Chr.9: 108862314-108862314 on Build GRCh38
rs752508657 Chr.9: 108862304-108862304 on Build GRCh38
rs781438053 Chr.9: 108862463-108862463 on Build GRCh38
rs751656468 Chr.9: 108862459-108862459 on Build GRCh38
rs773885762 Chr.9: 108862443-108862443 on Build GRCh38
rs769876931 Chr.9: 108862404-108862404 on Build GRCh38
rs541324563 Chr.9: 108862384-108862384 on Build GRCh38
rs150570578 Chr.9: 108862363-108862363 on Build GRCh38
rs757444638 Chr.9: 108862306-108862306 on Build GRCh38
rs1052161431 Chr.9: 108862488-108862488 on Build GRCh38
rs778239046 Chr.9: 108862491-108862491 on Build GRCh38
rs867346152 Chr.9: 108862350-108862350 on Build GRCh38
rs745940963 Chr.9: 108862310-108862310 on Build GRCh38
rs746883505 Chr.9: 108862365-108862367 on Build GRCh38
rs748602710 Chr.9: 108862464-108862464 on Build GRCh38
rs958184341 Chr.9: 108862390-108862390 on Build GRCh38
rs769066418 Chr.9: 108862320-108862320 on Build GRCh38
rs749366196 Chr.9: 108862436-108862436 on Build GRCh38
rs772249486 Chr.9: 108862313-108862313 on Build GRCh38
rs756723476 Chr.9: 108862487-108862487 on Build GRCh38
rs768983062 Chr.9: 108862512-108862512 on Build GRCh38
rs933614719 Chr.9: 108862478-108862478 on Build GRCh38
rs770515602 Chr.9: 108862442-108862442 on Build GRCh38
rs528800850 Chr.9: 108862282-108862282 on Build GRCh38
rs745719906 Chr.9: 108862469-108862470 on Build GRCh38
rs562009267 Chr.9: 108862315-108862315 on Build GRCh38
rs763809988 Chr.9: 108862451-108862451 on Build GRCh38
rs752347038 Chr.9: 108862373-108862373 on Build GRCh38
rs559552010 Chr.9: 108862402-108862402 on Build GRCh38
rs759465196 Chr.9: 108862385-108862385 on Build GRCh38
rs771019158 Chr.9: 108862383-108862384 on Build GRCh38
rs770228436 Chr.9: 108862433-108862433 on Build GRCh38
rs201993006 Chr.9: 108862446-108862446 on Build GRCh38
rs929626036 Chr.9: 108862508-108862508 on Build GRCh38
rs755967608 Chr.9: 108862393-108862393 on Build GRCh38
rs747108147 Chr.9: 108862424-108862424 on Build GRCh38
rs780141230 Chr.9: 108862420-108862420 on Build GRCh38
rs368653764 Chr.9: 108862455-108862455 on Build GRCh38
rs11539309 Chr.9: 108862510-108862510 on Build GRCh38
rs777032610 Chr.9: 108862324-108862324 on Build GRCh38
rs781080517 Chr.9: 108862415-108862416 on Build GRCh38
rs752455696 Chr.9: 108862389-108862389 on Build GRCh38
rs529223031 Chr.9: 108862353-108862353 on Build GRCh38
rs767399859 Chr.9: 108862388-108862388 on Build GRCh38
rs139566194 Chr.9: 108862425-108862425 on Build GRCh38
rs762310749 Chr.9: 108862347-108862347 on Build GRCh38
rs755073101 Chr.9: 108862462-108862462 on Build GRCh38
rs781003916 Chr.9: 108862286-108862286 on Build GRCh38
rs570985989 Chr.9: 108862445-108862445 on Build GRCh38
rs758552714 Chr.9: 108862411-108862411 on Build GRCh38
rs201549336 Chr.9: 108862440-108862440 on Build GRCh38
rs773060237 Chr.9: 108862370-108862370 on Build GRCh38

View Full Product Details