Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170235107-170235378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCI
Entrez Gene ID
5584
Gene Name
protein kinase C iota
Gene Aliases
DXS1179E, PKCI, nPKC-iota
UniGene
Hs.478199
Gene Chromosome Location
Chr.3: 170222432-170305982 on Build GRCh38
Transcripts
NM_002740.5 XM_017006860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5962620 c.220G>A p.E74K Chr.3: 170235348-170235348 on Build GRCh38
7233087 c.219G>A p.E73E Chr.3: 170235347-170235347 on Build GRCh38
4467428 c.148C>T p.L50F Chr.3: 170235276-170235276 on Build GRCh38
4847613 c.157G>C p.E53Q Chr.3: 170235285-170235285 on Build GRCh38
3590027 c.141T>C p.F47F Chr.3: 170235269-170235269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370076127 Chr.3: 170235181-170235181 on Build GRCh38
rs772402293 Chr.3: 170235319-170235319 on Build GRCh38
rs755677529 Chr.3: 170235291-170235291 on Build GRCh38
rs747020297 Chr.3: 170235323-170235323 on Build GRCh38
rs757899632 Chr.3: 170235315-170235315 on Build GRCh38
rs527299746 Chr.3: 170235134-170235134 on Build GRCh38
rs1032101463 Chr.3: 170235301-170235301 on Build GRCh38
rs200321008 Chr.3: 170235354-170235354 on Build GRCh38
rs762597865 Chr.3: 170235236-170235236 on Build GRCh38
rs1029403354 Chr.3: 170235132-170235132 on Build GRCh38
rs776632465 Chr.3: 170235339-170235339 on Build GRCh38
rs762941664 Chr.3: 170235347-170235347 on Build GRCh38
rs772491628 Chr.3: 170235188-170235188 on Build GRCh38
rs751420634 Chr.3: 170235247-170235247 on Build GRCh38
rs768794063 Chr.3: 170235327-170235327 on Build GRCh38
rs775805215 Chr.3: 170235322-170235322 on Build GRCh38
rs760859573 Chr.3: 170235202-170235202 on Build GRCh38
rs774122349 Chr.3: 170235190-170235196 on Build GRCh38
rs759436279 Chr.3: 170235216-170235217 on Build GRCh38
rs376325909 Chr.3: 170235200-170235200 on Build GRCh38
rs202068509 Chr.3: 170235277-170235277 on Build GRCh38
rs191543472 Chr.3: 170235152-170235152 on Build GRCh38
rs201089076 Chr.3: 170235324-170235324 on Build GRCh38
rs759821509 Chr.3: 170235186-170235186 on Build GRCh38
rs767213844 Chr.3: 170235297-170235298 on Build GRCh38
rs140807328 Chr.3: 170235314-170235314 on Build GRCh38
rs766148364 Chr.3: 170235246-170235246 on Build GRCh38
rs773232919 Chr.3: 170235216-170235216 on Build GRCh38
rs369333148 Chr.3: 170235283-170235283 on Build GRCh38
rs765036171 Chr.3: 170235212-170235212 on Build GRCh38
rs961288557 Chr.3: 170235255-170235255 on Build GRCh38
rs749381509 Chr.3: 170235317-170235317 on Build GRCh38
rs867093541 Chr.3: 170235281-170235281 on Build GRCh38
rs767204074 Chr.3: 170235282-170235282 on Build GRCh38

View Full Product Details