Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87233380-87233621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF292
Entrez Gene ID
23036
Gene Name
zinc finger protein 292
Gene Aliases
Nbla00365, ZFP292, ZN-16, Zn-15, bA393I2.3
UniGene
Hs.485892
Gene Chromosome Location
Chr.6: 87155551-87264172 on Build GRCh38
Transcripts
NM_015021.1 XM_005248697.4 XM_011535625.2 XM_017010575.1 XM_017010576.1 XM_017010577.1 XM_017010578.1 XM_017010579.1 XM_017010580.1 XM_017010581.1 XM_017010582.1 XM_017010583.1 XM_017010584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442366 c.190A>G p.T64A Chr.6: 87233411-87233411 on Build GRCh38
7124545 c.649G>C p.D217H Chr.6: 87233435-87233435 on Build GRCh38
6471426 c.259C>A p.L87I Chr.6: 87233480-87233480 on Build GRCh38
7418296 c.250C>A p.Q84K Chr.6: 87233471-87233471 on Build GRCh38
1082212 c.629C>T p.A210V Chr.6: 87233415-87233415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201983558 Chr.6: 87233434-87233434 on Build GRCh38
rs750581478 Chr.6: 87233440-87233440 on Build GRCh38
rs747332293 Chr.6: 87233521-87233521 on Build GRCh38
rs752527655 Chr.6: 87233461-87233461 on Build GRCh38
rs765427153 Chr.6: 87233435-87233435 on Build GRCh38
rs372606820 Chr.6: 87233412-87233412 on Build GRCh38
rs755878554 Chr.6: 87233462-87233462 on Build GRCh38
rs754572936 Chr.6: 87233503-87233503 on Build GRCh38
rs370598235 Chr.6: 87233514-87233514 on Build GRCh38
rs776722461 Chr.6: 87233414-87233414 on Build GRCh38
rs775986532 Chr.6: 87233519-87233519 on Build GRCh38
rs201229625 Chr.6: 87233504-87233504 on Build GRCh38
rs745446754 Chr.6: 87233505-87233505 on Build GRCh38
rs1026546193 Chr.6: 87233581-87233581 on Build GRCh38
rs917526936 Chr.6: 87233413-87233413 on Build GRCh38
rs754936498 Chr.6: 87233448-87233448 on Build GRCh38
rs143331467 Chr.6: 87233580-87233580 on Build GRCh38
rs761920173 Chr.6: 87233547-87233547 on Build GRCh38
rs16878734 Chr.6: 87233563-87233563 on Build GRCh38
rs200257228 Chr.6: 87233545-87233545 on Build GRCh38
rs772551631 Chr.6: 87233511-87233511 on Build GRCh38
rs748927338 Chr.6: 87233497-87233497 on Build GRCh38
rs768933206 Chr.6: 87233539-87233539 on Build GRCh38
rs377445408 Chr.6: 87233419-87233419 on Build GRCh38
rs777421481 Chr.6: 87233473-87233473 on Build GRCh38
rs770183088 Chr.6: 87233558-87233558 on Build GRCh38
rs767629944 Chr.6: 87233453-87233453 on Build GRCh38
rs1016673659 Chr.6: 87233551-87233551 on Build GRCh38

View Full Product Details