Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31671497-31671764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
UniGene
Hs.17255
Gene Chromosome Location
Chr.21: 31671000-31732118 on Build GRCh38
Transcripts
NM_001145444.1 NM_001145445.1 NM_020706.2 XM_005261017.2 XM_006724035.2 XM_006724036.2 XM_017028415.1 XM_017028416.1 XM_017028417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481626 c.3185A>G p.D1062G Chr.21: 31671658-31671658 on Build GRCh38
1483903 c.3081C>T p.N1027N Chr.21: 31671762-31671762 on Build GRCh38
2842787 c.3087C>A p.D1029E Chr.21: 31671756-31671756 on Build GRCh38
5366192 c.3156C>T p.R1052R Chr.21: 31671687-31671687 on Build GRCh38
2842785 c.3105G>A p.R1035R Chr.21: 31671738-31671738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761459493 Chr.21: 31671551-31671551 on Build GRCh38
rs750620228 Chr.21: 31671687-31671687 on Build GRCh38
rs202128594 Chr.21: 31671725-31671725 on Build GRCh38
rs758295971 Chr.21: 31671689-31671689 on Build GRCh38
rs776175845 Chr.21: 31671541-31671541 on Build GRCh38
rs150267574 Chr.21: 31671566-31671566 on Build GRCh38
rs757252416 Chr.21: 31671683-31671683 on Build GRCh38
rs142270458 Chr.21: 31671645-31671645 on Build GRCh38
rs778996761 Chr.21: 31671597-31671597 on Build GRCh38
rs963666002 Chr.21: 31671616-31671616 on Build GRCh38
rs528190794 Chr.21: 31671530-31671530 on Build GRCh38
rs1011234438 Chr.21: 31671575-31671575 on Build GRCh38
rs200713906 Chr.21: 31671649-31671649 on Build GRCh38
rs769168666 Chr.21: 31671745-31671745 on Build GRCh38
rs768052599 Chr.21: 31671638-31671638 on Build GRCh38
rs761218828 Chr.21: 31671648-31671648 on Build GRCh38
rs779925064 Chr.21: 31671715-31671715 on Build GRCh38
rs771969961 Chr.21: 31671617-31671617 on Build GRCh38
rs998830801 Chr.21: 31671611-31671611 on Build GRCh38
rs904527622 Chr.21: 31671602-31671602 on Build GRCh38
rs780990870 Chr.21: 31671736-31671736 on Build GRCh38
rs754014862 Chr.21: 31671676-31671676 on Build GRCh38
rs751982837 Chr.21: 31671560-31671560 on Build GRCh38
rs772823824 Chr.21: 31671653-31671653 on Build GRCh38
rs546757948 Chr.21: 31671556-31671556 on Build GRCh38
rs370864418 Chr.21: 31671528-31671528 on Build GRCh38
rs759942580 Chr.21: 31671655-31671655 on Build GRCh38
rs763606869 Chr.21: 31671539-31671539 on Build GRCh38
rs745697205 Chr.21: 31671609-31671609 on Build GRCh38
rs750971849 Chr.21: 31671586-31671589 on Build GRCh38
rs1034774977 Chr.21: 31671640-31671640 on Build GRCh38
rs779243584 Chr.21: 31671685-31671685 on Build GRCh38
rs767821506 Chr.21: 31671657-31671657 on Build GRCh38
rs138418942 Chr.21: 31671738-31671738 on Build GRCh38
rs775587465 Chr.21: 31671620-31671620 on Build GRCh38
rs374026931 Chr.21: 31671582-31671582 on Build GRCh38
rs201240032 Chr.21: 31671573-31671573 on Build GRCh38
rs746900042 Chr.21: 31671637-31671637 on Build GRCh38
rs757555139 Chr.21: 31671586-31671586 on Build GRCh38
rs138926969 Chr.21: 31671571-31671571 on Build GRCh38
rs73903115 Chr.21: 31671669-31671669 on Build GRCh38
rs1017713649 Chr.21: 31671659-31671659 on Build GRCh38
rs753234671 Chr.21: 31671667-31671667 on Build GRCh38
rs746953503 Chr.21: 31671724-31671724 on Build GRCh38
rs144640840 Chr.21: 31671578-31671578 on Build GRCh38
rs367603854 Chr.21: 31671618-31671618 on Build GRCh38

View Full Product Details