Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 29465520-29465787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SVIL
Entrez Gene ID
6840
Gene Name
supervillin
Gene Aliases
-
UniGene
Hs.499209
Gene Chromosome Location
Chr.10: 29457338-29737001 on Build GRCh38
Transcripts
NM_001323599.1 NM_001323600.1 NM_003174.3 NM_021738.2 XM_005252570.2 XM_005252571.3 XM_005252573.2 XM_011519630.2 XM_011519631.2 XM_011519632.2 XM_011519633.2 XM_011519634.1 XM_011519635.1 XM_011519636.2 XM_011519637.2 XM_011519638.2 XM_017016575.1 XM_017016576.1 XM_017016577.1 XM_017016578.1 XM_017016579.1 XM_017016580.1 XM_017016581.1 XM_017016582.1 NR_110920.1 NR_110921.1 NR_110922.1 NR_110925.1 NR_110926.1 NR_110927.1 NR_110928.1 NR_110929.1

Gene Symbol
SVIL-AS1
Entrez Gene ID
102724316
Gene Name
SVIL antisense RNA 1
Gene Aliases
-
UniGene
Hs.499209
Gene Chromosome Location
Chr.10: 29409534-29487856 on Build GRCh38
Transcripts
NM_001323599.1 NM_001323600.1 NM_003174.3 NM_021738.2 XM_005252570.2 XM_005252571.3 XM_005252573.2 XM_011519630.2 XM_011519631.2 XM_011519632.2 XM_011519633.2 XM_011519634.1 XM_011519635.1 XM_011519636.2 XM_011519637.2 XM_011519638.2 XM_017016575.1 XM_017016576.1 XM_017016577.1 XM_017016578.1 XM_017016579.1 XM_017016580.1 XM_017016581.1 XM_017016582.1 NR_110920.1 NR_110921.1 NR_110922.1 NR_110925.1 NR_110926.1 NR_110927.1 NR_110928.1 NR_110929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5747120 c.6059A>G p.Y2020C Chr.10: 29465669-29465669 on Build GRCh38
6364117 c.6041C>A p.A2014E Chr.10: 29465687-29465687 on Build GRCh38
684556 c.5987G>A p.S1996N Chr.10: 29465741-29465741 on Build GRCh38
343841 c.6029C>G p.S2010C Chr.10: 29465699-29465699 on Build GRCh38
3368008 c.6086G>T p.S2029I Chr.10: 29465642-29465642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766222089 Chr.10: 29465696-29465696 on Build GRCh38
rs145284035 Chr.10: 29465673-29465673 on Build GRCh38
rs912997814 Chr.10: 29465643-29465643 on Build GRCh38
rs759298298 Chr.10: 29465708-29465708 on Build GRCh38
rs17694739 Chr.10: 29465606-29465606 on Build GRCh38
rs532348488 Chr.10: 29465594-29465594 on Build GRCh38
rs151196345 Chr.10: 29465692-29465692 on Build GRCh38
rs200107973 Chr.10: 29465608-29465608 on Build GRCh38
rs376172339 Chr.10: 29465694-29465694 on Build GRCh38
rs749465942 Chr.10: 29465665-29465665 on Build GRCh38
rs765018549 Chr.10: 29465589-29465589 on Build GRCh38
rs771185982 Chr.10: 29465553-29465553 on Build GRCh38
rs149615507 Chr.10: 29465674-29465674 on Build GRCh38
rs746255809 Chr.10: 29465741-29465741 on Build GRCh38
rs773978944 Chr.10: 29465698-29465698 on Build GRCh38
rs771331394 Chr.10: 29465670-29465670 on Build GRCh38
rs1057952 Chr.10: 29465680-29465680 on Build GRCh38
rs747705163 Chr.10: 29465684-29465684 on Build GRCh38
rs778027430 Chr.10: 29465663-29465663 on Build GRCh38
rs368340289 Chr.10: 29465571-29465571 on Build GRCh38
rs772663424 Chr.10: 29465679-29465679 on Build GRCh38
rs111478482 Chr.10: 29465689-29465689 on Build GRCh38
rs768292889 Chr.10: 29465564-29465564 on Build GRCh38
rs375330240 Chr.10: 29465562-29465562 on Build GRCh38
rs1019247954 Chr.10: 29465569-29465569 on Build GRCh38
rs4469788 Chr.10: 29465612-29465612 on Build GRCh38
rs145077377 Chr.10: 29465723-29465723 on Build GRCh38
rs752815649 Chr.10: 29465635-29465635 on Build GRCh38
rs150327954 Chr.10: 29465597-29465597 on Build GRCh38
rs138805266 Chr.10: 29465730-29465730 on Build GRCh38
rs994339440 Chr.10: 29465600-29465600 on Build GRCh38
rs866212672 Chr.10: 29465745-29465745 on Build GRCh38
rs183121122 Chr.10: 29465561-29465561 on Build GRCh38
rs777940872 Chr.10: 29465549-29465549 on Build GRCh38
rs201067657 Chr.10: 29465550-29465550 on Build GRCh38
rs754824969 Chr.10: 29465609-29465609 on Build GRCh38
rs56248456 Chr.10: 29465746-29465746 on Build GRCh38
rs187952659 Chr.10: 29465580-29465580 on Build GRCh38
rs111548204 Chr.10: 29465614-29465614 on Build GRCh38
rs764013430 Chr.10: 29465728-29465728 on Build GRCh38
rs7921306 Chr.10: 29465715-29465715 on Build GRCh38
rs762876101 Chr.10: 29465603-29465603 on Build GRCh38
rs377177601 Chr.10: 29465729-29465729 on Build GRCh38
rs756164209 Chr.10: 29465636-29465636 on Build GRCh38
rs140166967 Chr.10: 29465555-29465555 on Build GRCh38

View Full Product Details