Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86605165-86605390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374845
Entrez Gene ID
105374845
Gene Name
uncharacterized LOC105374845
Gene Aliases
-
UniGene
Hs.741633
Gene Chromosome Location
Chr.2: 86562345-86618766 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103
Entrez Gene ID
7844
Gene Name
ring finger protein 103
Gene Aliases
HKF-1, KF-1, KF1, ZFP-103, ZFP103
UniGene
Hs.469199
Gene Chromosome Location
Chr.2: 86603393-86623877 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103-CHMP3
Entrez Gene ID
100526767
Gene Name
RNF103-CHMP3 readthrough
Gene Aliases
RNF103-VPS24
UniGene
Hs.591582
Gene Chromosome Location
Chr.2: 86503430-86721122 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6767148 c.637C>T p.R213W Chr.2: 86605264-86605264 on Build GRCh38
1409854 c.609delT p.F203fs*4 Chr.2: 86605292-86605292 on Build GRCh38
3673756 c.533C>A p.P178Q Chr.2: 86605368-86605368 on Build GRCh38
5708351 c.594A>C p.E198D Chr.2: 86605307-86605307 on Build GRCh38
1023524 c.678A>T p.E226D Chr.2: 86605223-86605223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763576107 Chr.2: 86605352-86605352 on Build GRCh38
rs767159180 Chr.2: 86605265-86605265 on Build GRCh38
rs201226159 Chr.2: 86605313-86605313 on Build GRCh38
rs764506626 Chr.2: 86605363-86605363 on Build GRCh38
rs966706490 Chr.2: 86605320-86605320 on Build GRCh38
rs1561253 Chr.2: 86605271-86605271 on Build GRCh38
rs771078408 Chr.2: 86605317-86605317 on Build GRCh38
rs763712968 Chr.2: 86605231-86605231 on Build GRCh38
rs367619971 Chr.2: 86605209-86605209 on Build GRCh38
rs750016390 Chr.2: 86605270-86605270 on Build GRCh38
rs752277962 Chr.2: 86605318-86605318 on Build GRCh38
rs746047010 Chr.2: 86605324-86605324 on Build GRCh38
rs759994300 Chr.2: 86605214-86605214 on Build GRCh38
rs149860510 Chr.2: 86605254-86605254 on Build GRCh38
rs200619349 Chr.2: 86605339-86605339 on Build GRCh38
rs770238112 Chr.2: 86605328-86605328 on Build GRCh38
rs771262664 Chr.2: 86605191-86605191 on Build GRCh38
rs755321549 Chr.2: 86605263-86605263 on Build GRCh38
rs368892771 Chr.2: 86605285-86605285 on Build GRCh38
rs754847352 Chr.2: 86605302-86605302 on Build GRCh38
rs765470440 Chr.2: 86605215-86605215 on Build GRCh38
rs35921467 Chr.2: 86605362-86605363 on Build GRCh38
rs371846352 Chr.2: 86605217-86605217 on Build GRCh38
rs747092202 Chr.2: 86605314-86605314 on Build GRCh38
rs763341633 Chr.2: 86605228-86605228 on Build GRCh38

View Full Product Details