Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89873605-89873856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF25
Entrez Gene ID
22980
Gene Name
transcription factor 25
Gene Aliases
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049
UniGene
Hs.415342
Gene Chromosome Location
Chr.16: 89873583-89911384 on Build GRCh38
Transcripts
NM_014972.2 XM_005256297.2 XM_005256298.2 XM_005256299.2 XM_011522956.2 XM_011522957.2 XM_011522958.2 XM_011522959.2 XM_011522960.2 XM_011522961.2 XM_017023047.1 XM_017023048.1 XM_017023049.1 XM_017023050.1 XM_017023051.1 XM_017023052.1 XM_017023054.1 XM_017023055.1 XM_017023056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6145733 c.27G>T p.L9L Chr.16: 89873694-89873694 on Build GRCh38
4590323 c.35A>G p.E12G Chr.16: 89873702-89873702 on Build GRCh38
4129528 c.186C>T p.F62F Chr.16: 89873853-89873853 on Build GRCh38
4732683 c.1A>G p.M1V Chr.16: 89873668-89873668 on Build GRCh38
4333294 c.25C>G p.L9V Chr.16: 89873692-89873692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751220185 Chr.16: 89873623-89873623 on Build GRCh38
rs780368173 Chr.16: 89873725-89873725 on Build GRCh38
rs998701264 Chr.16: 89873783-89873783 on Build GRCh38
rs755109638 Chr.16: 89873789-89873789 on Build GRCh38
rs751824206 Chr.16: 89873784-89873784 on Build GRCh38
rs542920247 Chr.16: 89873807-89873807 on Build GRCh38
rs773679581 Chr.16: 89873741-89873741 on Build GRCh38
rs984291141 Chr.16: 89873817-89873817 on Build GRCh38
rs760178755 Chr.16: 89873754-89873754 on Build GRCh38
rs930791953 Chr.16: 89873732-89873732 on Build GRCh38
rs377564706 Chr.16: 89873651-89873651 on Build GRCh38
rs777265841 Chr.16: 89873663-89873663 on Build GRCh38
rs758559258 Chr.16: 89873779-89873779 on Build GRCh38
rs755835505 Chr.16: 89873636-89873636 on Build GRCh38
rs150672592 Chr.16: 89873736-89873736 on Build GRCh38
rs201905874 Chr.16: 89873639-89873639 on Build GRCh38
rs560994628 Chr.16: 89873820-89873820 on Build GRCh38
rs766306758 Chr.16: 89873780-89873780 on Build GRCh38
rs774983711 Chr.16: 89873696-89873696 on Build GRCh38
rs920778543 Chr.16: 89873713-89873713 on Build GRCh38
rs545118216 Chr.16: 89873708-89873708 on Build GRCh38
rs771470053 Chr.16: 89873753-89873753 on Build GRCh38
rs757200510 Chr.16: 89873711-89873711 on Build GRCh38
rs759133077 Chr.16: 89873626-89873626 on Build GRCh38
rs748552916 Chr.16: 89873794-89873794 on Build GRCh38
rs369986838 Chr.16: 89873637-89873637 on Build GRCh38
rs779067324 Chr.16: 89873643-89873643 on Build GRCh38
rs13338056 Chr.16: 89873824-89873824 on Build GRCh38
rs773804363 Chr.16: 89873673-89873673 on Build GRCh38
rs992967709 Chr.16: 89873811-89873811 on Build GRCh38
rs767240026 Chr.16: 89873683-89873683 on Build GRCh38
rs755378511 Chr.16: 89873727-89873727 on Build GRCh38
rs778311342 Chr.16: 89873800-89873800 on Build GRCh38
rs761869554 Chr.16: 89873761-89873761 on Build GRCh38
rs373492730 Chr.16: 89873834-89873834 on Build GRCh38
rs970096636 Chr.16: 89873815-89873815 on Build GRCh38
rs768876241 Chr.16: 89873662-89873662 on Build GRCh38
rs373068160 Chr.16: 89873760-89873760 on Build GRCh38
rs746696128 Chr.16: 89873809-89873809 on Build GRCh38
rs748679373 Chr.16: 89873731-89873731 on Build GRCh38
rs957802340 Chr.16: 89873624-89873624 on Build GRCh38
rs760576413 Chr.16: 89873701-89873701 on Build GRCh38
rs866363973 Chr.16: 89873680-89873680 on Build GRCh38
rs989242855 Chr.16: 89873627-89873627 on Build GRCh38
rs781481877 Chr.16: 89873730-89873730 on Build GRCh38
rs940267951 Chr.16: 89873822-89873822 on Build GRCh38
rs574192477 Chr.16: 89873629-89873629 on Build GRCh38
rs762505967 Chr.16: 89873770-89873772 on Build GRCh38
rs115517842 Chr.16: 89873676-89873676 on Build GRCh38
rs769694962 Chr.16: 89873837-89873837 on Build GRCh38
rs772368625 Chr.16: 89873650-89873650 on Build GRCh38
rs768150645 Chr.16: 89873819-89873819 on Build GRCh38
rs1036047523 Chr.16: 89873682-89873682 on Build GRCh38
rs750487313 Chr.16: 89873718-89873718 on Build GRCh38
rs749754028 Chr.16: 89873806-89873806 on Build GRCh38
rs762244532 Chr.16: 89873665-89873665 on Build GRCh38
rs139741086 Chr.16: 89873758-89873758 on Build GRCh38
rs765427008 Chr.16: 89873715-89873715 on Build GRCh38
rs1040774929 Chr.16: 89873766-89873766 on Build GRCh38
rs750358832 Chr.16: 89873775-89873775 on Build GRCh38
rs1023462901 Chr.16: 89873812-89873812 on Build GRCh38
rs960296287 Chr.16: 89873802-89873802 on Build GRCh38
rs758375927 Chr.16: 89873724-89873724 on Build GRCh38
rs543072960 Chr.16: 89873637-89873638 on Build GRCh38
rs757478007 Chr.16: 89873642-89873642 on Build GRCh38
rs747195994 Chr.16: 89873726-89873726 on Build GRCh38
rs879655516 Chr.16: 89873705-89873705 on Build GRCh38
rs575295320 Chr.16: 89873764-89873764 on Build GRCh38
rs745432572 Chr.16: 89873745-89873745 on Build GRCh38

View Full Product Details