Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149070777-149071042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF786
Entrez Gene ID
136051
Gene Name
zinc finger protein 786
Gene Aliases
-
UniGene
Hs.729572
Gene Chromosome Location
Chr.7: 149069641-149090777 on Build GRCh38
Transcripts
NM_152411.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238502 c.1818G>T p.G606G Chr.7: 149070954-149070954 on Build GRCh38
6502829 c.1812G>A p.L604L Chr.7: 149070960-149070960 on Build GRCh38
3879139 c.1749C>A p.H583Q Chr.7: 149071023-149071023 on Build GRCh38
3635765 c.1779C>T p.F593F Chr.7: 149070993-149070993 on Build GRCh38
4744973 c.1743G>A p.T581T Chr.7: 149071029-149071029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749904470 Chr.7: 149070982-149070982 on Build GRCh38
rs775537628 Chr.7: 149070831-149070831 on Build GRCh38
rs755423563 Chr.7: 149070864-149070864 on Build GRCh38
rs571736582 Chr.7: 149070801-149070801 on Build GRCh38
rs780694969 Chr.7: 149070967-149070967 on Build GRCh38
rs749070939 Chr.7: 149070993-149070993 on Build GRCh38
rs752971225 Chr.7: 149070993-149070994 on Build GRCh38
rs201581301 Chr.7: 149070894-149070894 on Build GRCh38
rs200634145 Chr.7: 149070909-149070909 on Build GRCh38
rs774481144 Chr.7: 149070927-149070927 on Build GRCh38
rs767760536 Chr.7: 149070833-149070833 on Build GRCh38
rs746060036 Chr.7: 149070942-149070942 on Build GRCh38
rs771610664 Chr.7: 149070803-149070803 on Build GRCh38
rs368418768 Chr.7: 149070893-149070893 on Build GRCh38
rs752122751 Chr.7: 149070951-149070951 on Build GRCh38
rs200913705 Chr.7: 149070880-149070880 on Build GRCh38
rs756828290 Chr.7: 149070913-149070913 on Build GRCh38
rs757892586 Chr.7: 149070867-149070867 on Build GRCh38
rs191711263 Chr.7: 149070870-149070870 on Build GRCh38
rs775551028 Chr.7: 149070881-149070881 on Build GRCh38
rs765062697 Chr.7: 149070829-149070829 on Build GRCh38
rs760640981 Chr.7: 149070928-149070928 on Build GRCh38
rs760012696 Chr.7: 149070915-149070916 on Build GRCh38
rs766491480 Chr.7: 149070899-149070899 on Build GRCh38
rs765692939 Chr.7: 149070901-149070901 on Build GRCh38
rs74782558 Chr.7: 149070813-149070813 on Build GRCh38
rs779877843 Chr.7: 149070989-149070989 on Build GRCh38
rs775726975 Chr.7: 149071019-149071019 on Build GRCh38
rs776755256 Chr.7: 149070869-149070869 on Build GRCh38
rs758838611 Chr.7: 149070905-149070905 on Build GRCh38
rs765637485 Chr.7: 149070992-149070995 on Build GRCh38
rs769978952 Chr.7: 149071018-149071018 on Build GRCh38
rs779927007 Chr.7: 149070923-149070923 on Build GRCh38
rs958359135 Chr.7: 149070841-149070841 on Build GRCh38
rs768717144 Chr.7: 149070885-149070885 on Build GRCh38
rs766657533 Chr.7: 149070859-149070859 on Build GRCh38
rs772848173 Chr.7: 149070809-149070809 on Build GRCh38
rs769344146 Chr.7: 149070922-149070922 on Build GRCh38
rs371048918 Chr.7: 149070823-149070823 on Build GRCh38
rs776709318 Chr.7: 149071007-149071007 on Build GRCh38
rs754133533 Chr.7: 149070900-149070900 on Build GRCh38
rs371831859 Chr.7: 149070916-149070916 on Build GRCh38
rs745968865 Chr.7: 149071011-149071011 on Build GRCh38
rs762922242 Chr.7: 149070883-149070883 on Build GRCh38
rs750540733 Chr.7: 149070834-149070834 on Build GRCh38
rs753007851 Chr.7: 149070945-149070945 on Build GRCh38
rs375572534 Chr.7: 149070821-149070821 on Build GRCh38
rs779256801 Chr.7: 149070866-149070866 on Build GRCh38
rs755750935 Chr.7: 149070987-149070987 on Build GRCh38
rs367578528 Chr.7: 149070965-149070965 on Build GRCh38
rs921270553 Chr.7: 149070936-149070936 on Build GRCh38
rs945563044 Chr.7: 149070964-149070964 on Build GRCh38
rs771108934 Chr.7: 149070929-149070929 on Build GRCh38
rs759748899 Chr.7: 149070940-149070940 on Build GRCh38
rs777209058 Chr.7: 149070868-149070868 on Build GRCh38
rs375415453 Chr.7: 149070889-149070889 on Build GRCh38
rs778989581 Chr.7: 149070996-149070996 on Build GRCh38
rs776708319 Chr.7: 149070934-149070934 on Build GRCh38
rs572331975 Chr.7: 149070924-149070924 on Build GRCh38
rs973965497 Chr.7: 149070932-149070932 on Build GRCh38
rs745618544 Chr.7: 149070877-149070877 on Build GRCh38
rs573027357 Chr.7: 149070840-149070840 on Build GRCh38
rs199965509 Chr.7: 149070949-149070949 on Build GRCh38
rs762850364 Chr.7: 149070997-149070997 on Build GRCh38
rs374085431 Chr.7: 149070912-149070912 on Build GRCh38
rs768668002 Chr.7: 149070926-149070926 on Build GRCh38
rs895062832 Chr.7: 149070985-149070985 on Build GRCh38
rs564354984 Chr.7: 149070950-149070950 on Build GRCh38
rs754105396 Chr.7: 149070862-149070862 on Build GRCh38
rs370259254 Chr.7: 149070812-149070812 on Build GRCh38
rs367975181 Chr.7: 149070915-149070915 on Build GRCh38

View Full Product Details