Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168541942-168542167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XCL2
Entrez Gene ID
6846
Gene Name
X-C motif chemokine ligand 2
Gene Aliases
SCM-1b, SCM1B, SCYC2
UniGene
Hs.458346
Gene Chromosome Location
Chr.1: 168540765-168543997 on Build GRCh38
Transcripts
NM_003175.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024885 c.78C>G p.V26V Chr.1: 168542091-168542091 on Build GRCh38
3997056 c.62-4T>C p.? Chr.1: 168542111-168542111 on Build GRCh38
7145640 c.73G>A p.E25K Chr.1: 168542096-168542096 on Build GRCh38
3746152 c.82C>G p.H28D Chr.1: 168542087-168542087 on Build GRCh38
6541212 c.163T>A p.L55M Chr.1: 168542006-168542006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4478789 Chr.1: 168542111-168542111 on Build GRCh38
rs370342820 Chr.1: 168541968-168541968 on Build GRCh38
rs116168093 Chr.1: 168541967-168541967 on Build GRCh38
rs755652691 Chr.1: 168541979-168541979 on Build GRCh38
rs140039166 Chr.1: 168542099-168542099 on Build GRCh38
rs375960253 Chr.1: 168541987-168541987 on Build GRCh38
rs754751363 Chr.1: 168542017-168542017 on Build GRCh38
rs768975796 Chr.1: 168541993-168541993 on Build GRCh38
rs773490264 Chr.1: 168542076-168542076 on Build GRCh38
rs767305180 Chr.1: 168542030-168542030 on Build GRCh38
rs770920437 Chr.1: 168542135-168542135 on Build GRCh38
rs200963789 Chr.1: 168542108-168542108 on Build GRCh38
rs146927911 Chr.1: 168542054-168542054 on Build GRCh38
rs746389958 Chr.1: 168541984-168541984 on Build GRCh38
rs762052390 Chr.1: 168542006-168542006 on Build GRCh38
rs778118723 Chr.1: 168542128-168542128 on Build GRCh38
rs759451456 Chr.1: 168542138-168542138 on Build GRCh38
rs770179447 Chr.1: 168542124-168542124 on Build GRCh38
rs777200720 Chr.1: 168541983-168541983 on Build GRCh38
rs145525833 Chr.1: 168542031-168542031 on Build GRCh38
rs773306206 Chr.1: 168542089-168542089 on Build GRCh38
rs749482486 Chr.1: 168542131-168542131 on Build GRCh38
rs201016321 Chr.1: 168541986-168541986 on Build GRCh38
rs763023609 Chr.1: 168542091-168542091 on Build GRCh38
rs752010542 Chr.1: 168542112-168542112 on Build GRCh38
rs142896007 Chr.1: 168542093-168542093 on Build GRCh38
rs1045395856 Chr.1: 168542056-168542056 on Build GRCh38
rs770125968 Chr.1: 168542065-168542065 on Build GRCh38
rs374310854 Chr.1: 168542016-168542016 on Build GRCh38
rs541486301 Chr.1: 168542113-168542113 on Build GRCh38
rs753398601 Chr.1: 168542105-168542105 on Build GRCh38
rs765409091 Chr.1: 168542010-168542010 on Build GRCh38
rs748517105 Chr.1: 168542123-168542123 on Build GRCh38
rs773365565 Chr.1: 168542011-168542011 on Build GRCh38
rs4301615 Chr.1: 168542087-168542087 on Build GRCh38
rs755707749 Chr.1: 168542039-168542039 on Build GRCh38
rs748392312 Chr.1: 168542062-168542062 on Build GRCh38
rs1012865054 Chr.1: 168542047-168542047 on Build GRCh38
rs558497766 Chr.1: 168542043-168542043 on Build GRCh38
rs780406644 Chr.1: 168542048-168542048 on Build GRCh38
rs747497420 Chr.1: 168542053-168542053 on Build GRCh38
rs746563888 Chr.1: 168542117-168542117 on Build GRCh38
rs370855359 Chr.1: 168542012-168542012 on Build GRCh38
rs4282797 Chr.1: 168542046-168542046 on Build GRCh38
rs768152986 Chr.1: 168542126-168542126 on Build GRCh38
rs774433599 Chr.1: 168542136-168542136 on Build GRCh38
rs1022490882 Chr.1: 168542134-168542134 on Build GRCh38
rs777325495 Chr.1: 168542040-168542040 on Build GRCh38
rs776983840 Chr.1: 168542058-168542058 on Build GRCh38
rs779658973 Chr.1: 168542116-168542117 on Build GRCh38
rs751296820 Chr.1: 168541966-168541966 on Build GRCh38
rs758020940 Chr.1: 168542094-168542095 on Build GRCh38
rs774517244 Chr.1: 168542121-168542121 on Build GRCh38
rs772557907 Chr.1: 168541985-168541985 on Build GRCh38
rs763093707 Chr.1: 168542077-168542077 on Build GRCh38
rs776737025 Chr.1: 168542000-168542000 on Build GRCh38
rs771041566 Chr.1: 168542079-168542079 on Build GRCh38
rs781466138 Chr.1: 168542116-168542116 on Build GRCh38
rs537245270 Chr.1: 168542036-168542036 on Build GRCh38
rs1047139325 Chr.1: 168541981-168541981 on Build GRCh38
rs4501820 Chr.1: 168542083-168542083 on Build GRCh38
rs879218300 Chr.1: 168542130-168542130 on Build GRCh38
rs750075246 Chr.1: 168541977-168541979 on Build GRCh38
rs747789111 Chr.1: 168541975-168541975 on Build GRCh38

View Full Product Details