Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57661089-57661362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B1
Entrez Gene ID
5205
Gene Name
ATPase phospholipid transporting 8B1
Gene Aliases
ATPIC, BRIC, FIC1, ICP1, PFIC, PFIC1
UniGene
Hs.216623
Gene Chromosome Location
Chr.18: 57646426-57803828 on Build GRCh38
Transcripts
NM_005603.4 XM_006722481.3 XM_011526022.2 XM_011526023.2 NM_001242804.1

Gene Symbol
LOC100505549
Entrez Gene ID
100505549
Gene Name
uncharacterized LOC100505549
Gene Aliases
-
UniGene
Hs.660645
Gene Chromosome Location
Chr.18: 57630302-57669276 on Build GRCh38
Transcripts
NM_005603.4 XM_006722481.3 XM_011526022.2 XM_011526023.2 NM_001242804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1184167 c.2645G>T p.R882M Chr.18: 57661236-57661236 on Build GRCh38
5625938 c.2546G>A p.R849Q Chr.18: 57661335-57661335 on Build GRCh38
438244 c.2584G>T p.A862S Chr.18: 57661297-57661297 on Build GRCh38
6608301 c.2689G>T p.D897Y Chr.18: 57661192-57661192 on Build GRCh38
6608300 c.2602G>A p.V868I Chr.18: 57661279-57661279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563512282 Chr.18: 57661139-57661139 on Build GRCh38
rs963708713 Chr.18: 57661308-57661308 on Build GRCh38
rs34105567 Chr.18: 57661328-57661328 on Build GRCh38
rs958793199 Chr.18: 57661289-57661289 on Build GRCh38
rs758888388 Chr.18: 57661165-57661165 on Build GRCh38
rs767398921 Chr.18: 57661224-57661224 on Build GRCh38
rs747330169 Chr.18: 57661184-57661184 on Build GRCh38
rs761625705 Chr.18: 57661306-57661306 on Build GRCh38
rs35757087 Chr.18: 57661217-57661217 on Build GRCh38
rs751994606 Chr.18: 57661274-57661274 on Build GRCh38
rs147024196 Chr.18: 57661307-57661307 on Build GRCh38
rs121909103 Chr.18: 57661282-57661282 on Build GRCh38
rs2271770 Chr.18: 57661140-57661140 on Build GRCh38
rs747209505 Chr.18: 57661129-57661130 on Build GRCh38
rs121909098 Chr.18: 57661207-57661207 on Build GRCh38
rs35455826 Chr.18: 57661281-57661282 on Build GRCh38
rs752763738 Chr.18: 57661257-57661257 on Build GRCh38
rs748897543 Chr.18: 57661292-57661292 on Build GRCh38
rs778408511 Chr.18: 57661273-57661273 on Build GRCh38
rs755157556 Chr.18: 57661152-57661152 on Build GRCh38
rs762815189 Chr.18: 57661208-57661208 on Build GRCh38
rs368471389 Chr.18: 57661301-57661301 on Build GRCh38
rs774396862 Chr.18: 57661297-57661297 on Build GRCh38
rs762368766 Chr.18: 57661131-57661131 on Build GRCh38
rs765429033 Chr.18: 57661256-57661256 on Build GRCh38
rs764028689 Chr.18: 57661211-57661211 on Build GRCh38
rs545885891 Chr.18: 57661206-57661206 on Build GRCh38
rs572394080 Chr.18: 57661247-57661247 on Build GRCh38
rs372904950 Chr.18: 57661160-57661160 on Build GRCh38
rs769329072 Chr.18: 57661201-57661201 on Build GRCh38
rs754099805 Chr.18: 57661149-57661149 on Build GRCh38
rs771192142 Chr.18: 57661187-57661187 on Build GRCh38
rs773092889 Chr.18: 57661323-57661323 on Build GRCh38
rs754154585 Chr.18: 57661229-57661229 on Build GRCh38
rs199588645 Chr.18: 57661134-57661134 on Build GRCh38
rs532483640 Chr.18: 57661280-57661280 on Build GRCh38
rs886043906 Chr.18: 57661200-57661200 on Build GRCh38
rs34681681 Chr.18: 57661326-57661326 on Build GRCh38
rs776319141 Chr.18: 57661189-57661189 on Build GRCh38
rs540027832 Chr.18: 57661218-57661218 on Build GRCh38
rs144656719 Chr.18: 57661335-57661335 on Build GRCh38
rs748633006 Chr.18: 57661163-57661163 on Build GRCh38
rs758831980 Chr.18: 57661269-57661269 on Build GRCh38
rs750735500 Chr.18: 57661279-57661279 on Build GRCh38
rs199716374 Chr.18: 57661244-57661244 on Build GRCh38
rs201885528 Chr.18: 57661286-57661286 on Build GRCh38
rs745546266 Chr.18: 57661281-57661281 on Build GRCh38
rs113626153 Chr.18: 57661190-57661190 on Build GRCh38
rs768040256 Chr.18: 57661295-57661295 on Build GRCh38

View Full Product Details