Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4759164-4759423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
UniGene
Hs.487393
Gene Chromosome Location
Chr.7: 4644691-4771443 on Build GRCh38
Transcripts
NM_001037165.1 XM_011515191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6666349 c.1325G>A p.R442Q Chr.7: 4759197-4759197 on Build GRCh38
4765024 c.1415_1416insC p.V474fs*78 Chr.7: 4759314-4759315 on Build GRCh38
1622904 c.1345+9C>T p.? Chr.7: 4759226-4759226 on Build GRCh38
5766739 c.1470C>T p.L490L Chr.7: 4759369-4759369 on Build GRCh38
6666304 c.1390C>T p.R464W Chr.7: 4759196-4759196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372195376 Chr.7: 4759246-4759246 on Build GRCh38
rs983882306 Chr.7: 4759212-4759212 on Build GRCh38
rs766226492 Chr.7: 4759263-4759263 on Build GRCh38
rs763625523 Chr.7: 4759297-4759297 on Build GRCh38
rs746221966 Chr.7: 4759257-4759257 on Build GRCh38
rs763536098 Chr.7: 4759242-4759242 on Build GRCh38
rs148694575 Chr.7: 4759369-4759369 on Build GRCh38
rs755214829 Chr.7: 4759339-4759339 on Build GRCh38
rs752269513 Chr.7: 4759237-4759237 on Build GRCh38
rs368436854 Chr.7: 4759289-4759289 on Build GRCh38
rs368553969 Chr.7: 4759197-4759197 on Build GRCh38
rs374000244 Chr.7: 4759235-4759235 on Build GRCh38
rs749458058 Chr.7: 4759264-4759264 on Build GRCh38
rs780420365 Chr.7: 4759261-4759261 on Build GRCh38
rs746977867 Chr.7: 4759271-4759271 on Build GRCh38
rs1033406050 Chr.7: 4759360-4759360 on Build GRCh38
rs1052347166 Chr.7: 4759270-4759270 on Build GRCh38
rs531929514 Chr.7: 4759345-4759345 on Build GRCh38
rs762977413 Chr.7: 4759239-4759240 on Build GRCh38
rs756351396 Chr.7: 4759260-4759260 on Build GRCh38
rs759517208 Chr.7: 4759383-4759384 on Build GRCh38
rs750822067 Chr.7: 4759307-4759307 on Build GRCh38
rs754228342 Chr.7: 4759399-4759399 on Build GRCh38
rs746107121 Chr.7: 4759203-4759203 on Build GRCh38
rs775135870 Chr.7: 4759291-4759291 on Build GRCh38
rs757574740 Chr.7: 4759198-4759198 on Build GRCh38
rs925611580 Chr.7: 4759282-4759282 on Build GRCh38
rs369230415 Chr.7: 4759318-4759318 on Build GRCh38
rs919493975 Chr.7: 4759259-4759259 on Build GRCh38
rs759368909 Chr.7: 4759286-4759286 on Build GRCh38
rs143714848 Chr.7: 4759216-4759216 on Build GRCh38
rs983830013 Chr.7: 4759384-4759384 on Build GRCh38
rs768275843 Chr.7: 4759370-4759370 on Build GRCh38
rs761383854 Chr.7: 4759373-4759373 on Build GRCh38
rs1037868541 Chr.7: 4759288-4759288 on Build GRCh38
rs868350010 Chr.7: 4759187-4759187 on Build GRCh38
rs889319853 Chr.7: 4759277-4759277 on Build GRCh38
rs943025937 Chr.7: 4759273-4759273 on Build GRCh38
rs776517689 Chr.7: 4759277-4759278 on Build GRCh38
rs754138347 Chr.7: 4759333-4759333 on Build GRCh38
rs773910421 Chr.7: 4759372-4759372 on Build GRCh38
rs772204773 Chr.7: 4759223-4759223 on Build GRCh38
rs757905106 Chr.7: 4759241-4759241 on Build GRCh38
rs768055959 Chr.7: 4759305-4759305 on Build GRCh38
rs749368186 Chr.7: 4759219-4759219 on Build GRCh38
rs756506614 Chr.7: 4759308-4759308 on Build GRCh38
rs770875716 Chr.7: 4759272-4759272 on Build GRCh38
rs550442414 Chr.7: 4759348-4759348 on Build GRCh38
rs779177576 Chr.7: 4759341-4759341 on Build GRCh38
rs905875128 Chr.7: 4759330-4759330 on Build GRCh38
rs374666686 Chr.7: 4759405-4759405 on Build GRCh38
rs527250909 Chr.7: 4759226-4759226 on Build GRCh38
rs774445243 Chr.7: 4759240-4759244 on Build GRCh38
rs770027556 Chr.7: 4759210-4759210 on Build GRCh38
rs375036990 Chr.7: 4759304-4759304 on Build GRCh38
rs775223866 Chr.7: 4759361-4759361 on Build GRCh38
rs372541725 Chr.7: 4759396-4759396 on Build GRCh38
rs776343559 Chr.7: 4759275-4759275 on Build GRCh38
rs774528295 Chr.7: 4759265-4759265 on Build GRCh38
rs766872799 Chr.7: 4759377-4759377 on Build GRCh38
rs527754335 Chr.7: 4759247-4759247 on Build GRCh38
rs766885716 Chr.7: 4759397-4759397 on Build GRCh38
rs776450295 Chr.7: 4759230-4759230 on Build GRCh38
rs201286259 Chr.7: 4759294-4759294 on Build GRCh38
rs773289863 Chr.7: 4759227-4759227 on Build GRCh38
rs376544531 Chr.7: 4759251-4759251 on Build GRCh38
rs745732765 Chr.7: 4759350-4759350 on Build GRCh38
rs769597835 Chr.7: 4759356-4759356 on Build GRCh38
rs760502960 Chr.7: 4759260-4759263 on Build GRCh38
rs751038104 Chr.7: 4759301-4759301 on Build GRCh38
rs770836439 Chr.7: 4759346-4759346 on Build GRCh38
rs766114576 Chr.7: 4759229-4759229 on Build GRCh38
rs1047589308 Chr.7: 4759299-4759299 on Build GRCh38
rs368950661 Chr.7: 4759200-4759200 on Build GRCh38
rs760634384 Chr.7: 4759228-4759228 on Build GRCh38
rs750956393 Chr.7: 4759395-4759395 on Build GRCh38
rs929055838 Chr.7: 4759296-4759296 on Build GRCh38

View Full Product Details