Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1004753-1004994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN3B
Entrez Gene ID
116444
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Gene Aliases
GluN3B, NR3B
UniGene
Hs.660378
Gene Chromosome Location
Chr.19: 1000419-1009814 on Build GRCh38
Transcripts
NM_138690.2 XM_017026243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4688804 c.1457G>A p.R486Q Chr.19: 1004958-1004958 on Build GRCh38
6675626 c.1262C>T p.T421M Chr.19: 1004763-1004763 on Build GRCh38
5429972 c.1358C>T p.S453L Chr.19: 1004859-1004859 on Build GRCh38
6675623 c.1348A>G p.T450A Chr.19: 1004849-1004849 on Build GRCh38
7154523 c.1366C>G p.L456V Chr.19: 1004867-1004867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139557341 Chr.19: 1004803-1004803 on Build GRCh38
rs766382202 Chr.19: 1004963-1004963 on Build GRCh38
rs964135310 Chr.19: 1004897-1004897 on Build GRCh38
rs763353254 Chr.19: 1004851-1004851 on Build GRCh38
rs746593296 Chr.19: 1004973-1004973 on Build GRCh38
rs147521452 Chr.19: 1004774-1004774 on Build GRCh38
rs201601935 Chr.19: 1004855-1004855 on Build GRCh38
rs368049150 Chr.19: 1004860-1004860 on Build GRCh38
rs761357444 Chr.19: 1004783-1004783 on Build GRCh38
rs878889783 Chr.19: 1004970-1004970 on Build GRCh38
rs753149188 Chr.19: 1004849-1004849 on Build GRCh38
rs767617201 Chr.19: 1004902-1004902 on Build GRCh38
rs771611326 Chr.19: 1004842-1004842 on Build GRCh38
rs777543158 Chr.19: 1004930-1004930 on Build GRCh38
rs758507260 Chr.19: 1004800-1004800 on Build GRCh38
rs933848477 Chr.19: 1004906-1004906 on Build GRCh38
rs776145597 Chr.19: 1004938-1004938 on Build GRCh38
rs774035095 Chr.19: 1004834-1004834 on Build GRCh38
rs754737058 Chr.19: 1004918-1004918 on Build GRCh38
rs199811690 Chr.19: 1004964-1004964 on Build GRCh38
rs371910067 Chr.19: 1004867-1004867 on Build GRCh38
rs748647527 Chr.19: 1004898-1004899 on Build GRCh38
rs745740525 Chr.19: 1004820-1004820 on Build GRCh38
rs762054649 Chr.19: 1004957-1004957 on Build GRCh38
rs201453184 Chr.19: 1004967-1004967 on Build GRCh38
rs201400509 Chr.19: 1004960-1004960 on Build GRCh38
rs754387721 Chr.19: 1004792-1004792 on Build GRCh38
rs995765219 Chr.19: 1004866-1004866 on Build GRCh38
rs753813570 Chr.19: 1004913-1004913 on Build GRCh38
rs756841039 Chr.19: 1004974-1004974 on Build GRCh38
rs751621630 Chr.19: 1004840-1004840 on Build GRCh38
rs766363768 Chr.19: 1004911-1004911 on Build GRCh38
rs1045561835 Chr.19: 1004939-1004939 on Build GRCh38
rs757287420 Chr.19: 1004859-1004859 on Build GRCh38
rs771500715 Chr.19: 1004876-1004876 on Build GRCh38
rs372774829 Chr.19: 1004965-1004965 on Build GRCh38
rs201092021 Chr.19: 1004793-1004793 on Build GRCh38
rs377106421 Chr.19: 1004886-1004886 on Build GRCh38
rs558873297 Chr.19: 1004954-1004954 on Build GRCh38
rs764451151 Chr.19: 1004893-1004893 on Build GRCh38
rs1001941197 Chr.19: 1004968-1004968 on Build GRCh38
rs766848212 Chr.19: 1004790-1004790 on Build GRCh38
rs778500000 Chr.19: 1004874-1004874 on Build GRCh38
rs563617329 Chr.19: 1004798-1004798 on Build GRCh38
rs61750461 Chr.19: 1004873-1004873 on Build GRCh38
rs892110155 Chr.19: 1004828-1004828 on Build GRCh38
rs568781579 Chr.19: 1004882-1004882 on Build GRCh38
rs758162921 Chr.19: 1004928-1004928 on Build GRCh38
rs145632166 Chr.19: 1004872-1004872 on Build GRCh38
rs764279599 Chr.19: 1004854-1004854 on Build GRCh38
rs763293595 Chr.19: 1004888-1004888 on Build GRCh38
rs781058218 Chr.19: 1004810-1004810 on Build GRCh38
rs533006807 Chr.19: 1004909-1004909 on Build GRCh38
rs768236351 Chr.19: 1004829-1004829 on Build GRCh38
rs140599178 Chr.19: 1004937-1004937 on Build GRCh38
rs774451027 Chr.19: 1004896-1004896 on Build GRCh38
rs749744571 Chr.19: 1004948-1004948 on Build GRCh38
rs746574979 Chr.19: 1004932-1004932 on Build GRCh38
rs10666583 Chr.19: 1004897-1004898 on Build GRCh38
rs144095033 Chr.19: 1004875-1004875 on Build GRCh38
rs4806909 Chr.19: 1004845-1004845 on Build GRCh38
rs765635549 Chr.19: 1004797-1004797 on Build GRCh38
rs780253391 Chr.19: 1004851-1004853 on Build GRCh38
rs61736783 Chr.19: 1004809-1004809 on Build GRCh38
rs760067939 Chr.19: 1004844-1004844 on Build GRCh38
rs201638380 Chr.19: 1004804-1004804 on Build GRCh38
rs76859376 Chr.19: 1004881-1004881 on Build GRCh38
rs767778650 Chr.19: 1004958-1004958 on Build GRCh38
rs761292242 Chr.19: 1004838-1004838 on Build GRCh38
rs752584001 Chr.19: 1004966-1004966 on Build GRCh38
rs749482351 Chr.19: 1004885-1004885 on Build GRCh38
rs374546657 Chr.19: 1004827-1004827 on Build GRCh38
rs750543592 Chr.19: 1004904-1004904 on Build GRCh38
rs770471437 Chr.19: 1004885-1004885 on Build GRCh38
rs149732272 Chr.19: 1004865-1004865 on Build GRCh38
rs4806908 Chr.19: 1004824-1004824 on Build GRCh38
rs769589780 Chr.19: 1004821-1004821 on Build GRCh38
rs747783907 Chr.19: 1004919-1004919 on Build GRCh38
rs200904237 Chr.19: 1004905-1004905 on Build GRCh38
rs945871154 Chr.19: 1004933-1004933 on Build GRCh38
rs76549448 Chr.19: 1004884-1004884 on Build GRCh38
rs80202383 Chr.19: 1004951-1004951 on Build GRCh38

View Full Product Details