Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23563421-23563666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
UniGene
Hs.343244
Gene Chromosome Location
Chr.14: 23559563-23569056 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021743.1 XM_017021744.1 XM_017021745.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Gene Symbol
LOC102724814
Entrez Gene ID
102724814
Gene Name
uncharacterized LOC102724814
Gene Aliases
-
UniGene
Hs.569375
Gene Chromosome Location
Chr.14: 23561097-23568076 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021743.1 XM_017021744.1 XM_017021745.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6367976 c.1306G>T p.D436Y Chr.14: 23563484-23563484 on Build GRCh38
6601532 c.1346C>G p.A449G Chr.14: 23563444-23563444 on Build GRCh38
4464118 c.1319C>T p.A440V Chr.14: 23563471-23563471 on Build GRCh38
3987591 c.798-1G>A p.? Chr.14: 23563639-23563639 on Build GRCh38
1317875 c.1339G>T p.G447W Chr.14: 23563451-23563451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150451648 Chr.14: 23563453-23563453 on Build GRCh38
rs759093157 Chr.14: 23563446-23563446 on Build GRCh38
rs370083184 Chr.14: 23563585-23563585 on Build GRCh38
rs146957849 Chr.14: 23563613-23563613 on Build GRCh38
rs776340168 Chr.14: 23563568-23563568 on Build GRCh38
rs372112838 Chr.14: 23563634-23563634 on Build GRCh38
rs778617867 Chr.14: 23563509-23563509 on Build GRCh38
rs755447663 Chr.14: 23563582-23563597 on Build GRCh38
rs866292267 Chr.14: 23563555-23563555 on Build GRCh38
rs926994757 Chr.14: 23563506-23563506 on Build GRCh38
rs746634190 Chr.14: 23563564-23563567 on Build GRCh38
rs140628752 Chr.14: 23563451-23563451 on Build GRCh38
rs749982998 Chr.14: 23563444-23563445 on Build GRCh38
rs781231680 Chr.14: 23563640-23563640 on Build GRCh38
rs145360826 Chr.14: 23563466-23563466 on Build GRCh38
rs779809117 Chr.14: 23563452-23563452 on Build GRCh38
rs377233339 Chr.14: 23563518-23563518 on Build GRCh38
rs750830947 Chr.14: 23563484-23563484 on Build GRCh38
rs751773168 Chr.14: 23563622-23563622 on Build GRCh38
rs754369597 Chr.14: 23563470-23563470 on Build GRCh38
rs751542196 Chr.14: 23563488-23563488 on Build GRCh38
rs754721186 Chr.14: 23563445-23563445 on Build GRCh38
rs572740402 Chr.14: 23563444-23563444 on Build GRCh38
rs771582263 Chr.14: 23563526-23563526 on Build GRCh38
rs376640065 Chr.14: 23563462-23563462 on Build GRCh38
rs760289303 Chr.14: 23563545-23563545 on Build GRCh38
rs558439142 Chr.14: 23563468-23563468 on Build GRCh38
rs765684529 Chr.14: 23563482-23563482 on Build GRCh38
rs148842762 Chr.14: 23563481-23563481 on Build GRCh38
rs200923646 Chr.14: 23563486-23563486 on Build GRCh38
rs768282221 Chr.14: 23563551-23563551 on Build GRCh38
rs775189531 Chr.14: 23563528-23563528 on Build GRCh38
rs1054381923 Chr.14: 23563547-23563547 on Build GRCh38
rs752351764 Chr.14: 23563501-23563501 on Build GRCh38
rs867073333 Chr.14: 23563443-23563444 on Build GRCh38
rs184979172 Chr.14: 23563641-23563641 on Build GRCh38
rs757801608 Chr.14: 23563445-23563445 on Build GRCh38
rs543928791 Chr.14: 23563487-23563487 on Build GRCh38
rs574899141 Chr.14: 23563584-23563584 on Build GRCh38
rs201169755 Chr.14: 23563604-23563604 on Build GRCh38
rs770909327 Chr.14: 23563505-23563505 on Build GRCh38
rs201144415 Chr.14: 23563596-23563596 on Build GRCh38
rs373872679 Chr.14: 23563500-23563500 on Build GRCh38

View Full Product Details