Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55280214-55280487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCRTR2
Entrez Gene ID
3062
Gene Name
hypocretin receptor 2
Gene Aliases
OX2R
UniGene
Hs.151624
Gene Chromosome Location
Chr.6: 55174273-55284698 on Build GRCh38
Transcripts
NM_001526.3 XM_017010798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625821 c.984A>C p.R328S Chr.6: 55280323-55280323 on Build GRCh38
1643116 c.1071T>C p.S357S Chr.6: 55280410-55280410 on Build GRCh38
5724794 c.1016G>A p.R339K Chr.6: 55280355-55280355 on Build GRCh38
5601595 c.1041C>T p.T347T Chr.6: 55280380-55280380 on Build GRCh38
1080419 c.1073C>T p.A358V Chr.6: 55280412-55280412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199661865 Chr.6: 55280343-55280343 on Build GRCh38
rs765787442 Chr.6: 55280398-55280398 on Build GRCh38
rs907949384 Chr.6: 55280340-55280340 on Build GRCh38
rs763062065 Chr.6: 55280346-55280346 on Build GRCh38
rs922534637 Chr.6: 55280309-55280309 on Build GRCh38
rs201021664 Chr.6: 55280429-55280429 on Build GRCh38
rs777251944 Chr.6: 55280459-55280459 on Build GRCh38
rs565871240 Chr.6: 55280255-55280255 on Build GRCh38
rs777303200 Chr.6: 55280311-55280311 on Build GRCh38
rs200649013 Chr.6: 55280280-55280280 on Build GRCh38
rs754093211 Chr.6: 55280357-55280357 on Build GRCh38
rs528553321 Chr.6: 55280270-55280270 on Build GRCh38
rs188098116 Chr.6: 55280456-55280456 on Build GRCh38
rs771167608 Chr.6: 55280454-55280457 on Build GRCh38
rs747618725 Chr.6: 55280305-55280305 on Build GRCh38
rs201320094 Chr.6: 55280415-55280415 on Build GRCh38
rs200521452 Chr.6: 55280380-55280380 on Build GRCh38
rs376929252 Chr.6: 55280260-55280260 on Build GRCh38
rs368871388 Chr.6: 55280432-55280432 on Build GRCh38
rs74438514 Chr.6: 55280402-55280402 on Build GRCh38
rs749218027 Chr.6: 55280313-55280313 on Build GRCh38
rs750585608 Chr.6: 55280286-55280286 on Build GRCh38
rs200028165 Chr.6: 55280458-55280458 on Build GRCh38
rs750858233 Chr.6: 55280279-55280279 on Build GRCh38
rs562543892 Chr.6: 55280301-55280301 on Build GRCh38
rs548946088 Chr.6: 55280235-55280235 on Build GRCh38
rs780903591 Chr.6: 55280304-55280304 on Build GRCh38
rs974707486 Chr.6: 55280247-55280247 on Build GRCh38
rs868517395 Chr.6: 55280377-55280377 on Build GRCh38
rs199929361 Chr.6: 55280423-55280423 on Build GRCh38
rs772142372 Chr.6: 55280345-55280345 on Build GRCh38
rs773954688 Chr.6: 55280322-55280322 on Build GRCh38
rs775874081 Chr.6: 55280347-55280347 on Build GRCh38
rs761029342 Chr.6: 55280362-55280362 on Build GRCh38
rs755597307 Chr.6: 55280310-55280310 on Build GRCh38
rs983053652 Chr.6: 55280320-55280320 on Build GRCh38
rs758823526 Chr.6: 55280416-55280416 on Build GRCh38
rs202070686 Chr.6: 55280314-55280314 on Build GRCh38
rs965953397 Chr.6: 55280242-55280242 on Build GRCh38
rs764241978 Chr.6: 55280389-55280389 on Build GRCh38
rs761148778 Chr.6: 55280297-55280297 on Build GRCh38
rs76232306 Chr.6: 55280396-55280396 on Build GRCh38
rs1037631866 Chr.6: 55280360-55280360 on Build GRCh38

View Full Product Details