Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 65963134-65963405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGA2
Entrez Gene ID
8091
Gene Name
high mobility group AT-hook 2
Gene Aliases
BABL, HMGI-C, HMGIC, LIPO, STQTL9
UniGene
Hs.505924
Gene Chromosome Location
Chr.12: 65824460-65966291 on Build GRCh38
Transcripts
NM_003483.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1559406 c.323A>T p.E108V Chr.12: 65963285-65963285 on Build GRCh38
1299780 c.292G>A p.E98K Chr.12: 65963254-65963254 on Build GRCh38
3812785 c.319G>A p.E107K Chr.12: 65963281-65963281 on Build GRCh38
1242348 c.330G>T p.*110Y Chr.12: 65963292-65963292 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750164743 Chr.12: 65963335-65963335 on Build GRCh38
rs1052803161 Chr.12: 65963352-65963352 on Build GRCh38
rs377439189 Chr.12: 65963308-65963308 on Build GRCh38
rs966134592 Chr.12: 65963181-65963181 on Build GRCh38
rs764303427 Chr.12: 65963202-65963202 on Build GRCh38
rs767097693 Chr.12: 65963223-65963223 on Build GRCh38
rs781333561 Chr.12: 65963187-65963187 on Build GRCh38
rs57800850 Chr.12: 65963220-65963220 on Build GRCh38
rs199886793 Chr.12: 65963326-65963326 on Build GRCh38
rs954891299 Chr.12: 65963240-65963240 on Build GRCh38
rs778563109 Chr.12: 65963280-65963280 on Build GRCh38
rs143880692 Chr.12: 65963262-65963262 on Build GRCh38
rs745605177 Chr.12: 65963281-65963281 on Build GRCh38
rs752349495 Chr.12: 65963226-65963226 on Build GRCh38
rs201860923 Chr.12: 65963277-65963277 on Build GRCh38
rs563544853 Chr.12: 65963203-65963203 on Build GRCh38
rs772000269 Chr.12: 65963293-65963293 on Build GRCh38
rs374602952 Chr.12: 65963296-65963296 on Build GRCh38
rs765484518 Chr.12: 65963210-65963210 on Build GRCh38
rs768594093 Chr.12: 65963307-65963307 on Build GRCh38
rs200916173 Chr.12: 65963294-65963294 on Build GRCh38
rs750986770 Chr.12: 65963377-65963377 on Build GRCh38
rs755573046 Chr.12: 65963245-65963245 on Build GRCh38
rs936462782 Chr.12: 65963343-65963343 on Build GRCh38
rs762427793 Chr.12: 65963207-65963207 on Build GRCh38
rs972296499 Chr.12: 65963205-65963205 on Build GRCh38
rs754611627 Chr.12: 65963221-65963221 on Build GRCh38
rs1043157437 Chr.12: 65963297-65963297 on Build GRCh38
rs528046066 Chr.12: 65963279-65963279 on Build GRCh38

View Full Product Details