Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2859974-2860207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG2
Entrez Gene ID
8908
Gene Name
glycogenin 2
Gene Aliases
GN-2, GN2
UniGene
Hs.567381
Gene Chromosome Location
Chr.X: 2828712-2882820 on Build GRCh38
Transcripts
NM_001079855.1 NM_001184702.1 NM_001184703.1 NM_001184704.1 NM_003918.2 XM_011545600.2 XM_017029927.1 XM_017029928.1 XM_017029929.1 XM_017029930.1 XM_017029931.1

SNP Detail

SNP ID Chromosome Location
rs764375681 Chr.X: 2860049-2860049 on Build GRCh38
rs779792037 Chr.X: 2860082-2860082 on Build GRCh38
rs775430734 Chr.X: 2860035-2860035 on Build GRCh38
rs768493526 Chr.X: 2860099-2860099 on Build GRCh38
rs149379913 Chr.X: 2860050-2860050 on Build GRCh38
rs1018419757 Chr.X: 2859995-2859995 on Build GRCh38
rs889121132 Chr.X: 2860174-2860174 on Build GRCh38
rs750861457 Chr.X: 2860127-2860127 on Build GRCh38
rs200995683 Chr.X: 2860048-2860048 on Build GRCh38
rs183122891 Chr.X: 2860171-2860171 on Build GRCh38
rs768348721 Chr.X: 2860034-2860034 on Build GRCh38
rs759725646 Chr.X: 2860045-2860045 on Build GRCh38
rs138758732 Chr.X: 2860047-2860047 on Build GRCh38
rs190852517 Chr.X: 2860005-2860005 on Build GRCh38
rs746663164 Chr.X: 2860092-2860092 on Build GRCh38
rs766467407 Chr.X: 2860041-2860041 on Build GRCh38
rs777010939 Chr.X: 2860016-2860016 on Build GRCh38
rs758001369 Chr.X: 2860070-2860070 on Build GRCh38
rs906299690 Chr.X: 2860177-2860177 on Build GRCh38
rs763534786 Chr.X: 2860135-2860135 on Build GRCh38
rs369239504 Chr.X: 2860006-2860006 on Build GRCh38
rs923383271 Chr.X: 2860091-2860091 on Build GRCh38
rs778453159 Chr.X: 2860065-2860065 on Build GRCh38
rs771888233 Chr.X: 2860013-2860013 on Build GRCh38
rs201622542 Chr.X: 2860035-2860035 on Build GRCh38
rs767686645 Chr.X: 2860046-2860046 on Build GRCh38
rs774198371 Chr.X: 2860001-2860001 on Build GRCh38
rs201970747 Chr.X: 2860032-2860032 on Build GRCh38
rs776345214 Chr.X: 2860103-2860103 on Build GRCh38
rs377738756 Chr.X: 2860067-2860067 on Build GRCh38
rs765696744 Chr.X: 2860056-2860056 on Build GRCh38
rs146066080 Chr.X: 2860027-2860027 on Build GRCh38
rs774463697 Chr.X: 2860042-2860042 on Build GRCh38
rs746181691 Chr.X: 2860021-2860021 on Build GRCh38
rs964177599 Chr.X: 2860009-2860009 on Build GRCh38

View Full Product Details