Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42127388-42127659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKH
Entrez Gene ID
160851
Gene Name
diacylglycerol kinase eta
Gene Aliases
DGKeta
UniGene
Hs.630380
Gene Chromosome Location
Chr.13: 42040036-42256584 on Build GRCh38
Transcripts
NM_001204504.2 NM_152910.5 NM_178009.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4136224 c.200T>A p.I67N Chr.13: 42127470-42127470 on Build GRCh38
147677 c.303+8A>G p.? Chr.13: 42127581-42127581 on Build GRCh38
1940989 c.274C>T p.R92C Chr.13: 42127544-42127544 on Build GRCh38
4047659 c.276C>T p.R92R Chr.13: 42127546-42127546 on Build GRCh38
5376757 c.268C>T p.R90* Chr.13: 42127538-42127538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762884362 Chr.13: 42127611-42127612 on Build GRCh38
rs760083087 Chr.13: 42127572-42127572 on Build GRCh38
rs750570863 Chr.13: 42127610-42127610 on Build GRCh38
rs749533854 Chr.13: 42127620-42127620 on Build GRCh38
rs765186276 Chr.13: 42127440-42127442 on Build GRCh38
rs758216608 Chr.13: 42127464-42127464 on Build GRCh38
rs772820872 Chr.13: 42127557-42127557 on Build GRCh38
rs181300382 Chr.13: 42127533-42127533 on Build GRCh38
rs200957514 Chr.13: 42127582-42127582 on Build GRCh38
rs140398882 Chr.13: 42127531-42127531 on Build GRCh38
rs754758594 Chr.13: 42127436-42127436 on Build GRCh38
rs763913209 Chr.13: 42127608-42127608 on Build GRCh38
rs670676 Chr.13: 42127603-42127603 on Build GRCh38
rs766115476 Chr.13: 42127577-42127577 on Build GRCh38
rs558834369 Chr.13: 42127442-42127442 on Build GRCh38
rs760406979 Chr.13: 42127426-42127426 on Build GRCh38
rs370756160 Chr.13: 42127592-42127592 on Build GRCh38
rs370461472 Chr.13: 42127418-42127418 on Build GRCh38
rs745902521 Chr.13: 42127490-42127490 on Build GRCh38
rs1344286 Chr.13: 42127463-42127463 on Build GRCh38
rs9566925 Chr.13: 42127538-42127538 on Build GRCh38
rs796380805 Chr.13: 42127476-42127476 on Build GRCh38
rs752639807 Chr.13: 42127457-42127457 on Build GRCh38
rs976454082 Chr.13: 42127453-42127453 on Build GRCh38
rs749854131 Chr.13: 42127414-42127414 on Build GRCh38
rs780215186 Chr.13: 42127619-42127619 on Build GRCh38
rs781360779 Chr.13: 42127486-42127486 on Build GRCh38
rs773281658 Chr.13: 42127547-42127547 on Build GRCh38
rs772419889 Chr.13: 42127550-42127550 on Build GRCh38
rs756406141 Chr.13: 42127500-42127500 on Build GRCh38
rs767066460 Chr.13: 42127413-42127413 on Build GRCh38
rs920914397 Chr.13: 42127455-42127455 on Build GRCh38
rs143084777 Chr.13: 42127613-42127613 on Build GRCh38
rs201753254 Chr.13: 42127524-42127524 on Build GRCh38
rs769146516 Chr.13: 42127542-42127542 on Build GRCh38
rs150403121 Chr.13: 42127545-42127545 on Build GRCh38
rs540298428 Chr.13: 42127434-42127434 on Build GRCh38
rs605371 Chr.13: 42127581-42127581 on Build GRCh38
rs774798310 Chr.13: 42127544-42127544 on Build GRCh38
rs753684104 Chr.13: 42127435-42127435 on Build GRCh38
rs1046602603 Chr.13: 42127623-42127623 on Build GRCh38
rs200027507 Chr.13: 42127583-42127583 on Build GRCh38

View Full Product Details