Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144085428-144085683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAA1
Entrez Gene ID
8733
Gene Name
glycosylphosphatidylinositol anchor attachment 1
Gene Aliases
GAA1, hGAA1
UniGene
Hs.627962
Gene Chromosome Location
Chr.8: 144082621-144086216 on Build GRCh38
Transcripts
NM_003801.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2870775 c.1409C>T p.A470V Chr.8: 144085437-144085437 on Build GRCh38
1187666 c.1451G>A p.R484Q Chr.8: 144085479-144085479 on Build GRCh38
5984030 c.1539C>T p.I513I Chr.8: 144085660-144085660 on Build GRCh38
6502911 c.1550A>C p.N517T Chr.8: 144085671-144085671 on Build GRCh38
6112532 c.1545C>T p.L515L Chr.8: 144085666-144085666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373779637 Chr.8: 144085510-144085510 on Build GRCh38
rs369984599 Chr.8: 144085607-144085607 on Build GRCh38
rs552750856 Chr.8: 144085523-144085523 on Build GRCh38
rs367939672 Chr.8: 144085522-144085522 on Build GRCh38
rs782494128 Chr.8: 144085577-144085577 on Build GRCh38
rs782071116 Chr.8: 144085564-144085564 on Build GRCh38
rs73379193 Chr.8: 144085598-144085598 on Build GRCh38
rs782144477 Chr.8: 144085575-144085575 on Build GRCh38
rs782642507 Chr.8: 144085513-144085513 on Build GRCh38
rs782357196 Chr.8: 144085562-144085562 on Build GRCh38
rs561318989 Chr.8: 144085457-144085457 on Build GRCh38
rs202106798 Chr.8: 144085593-144085593 on Build GRCh38
rs782658003 Chr.8: 144085627-144085627 on Build GRCh38
rs1021718622 Chr.8: 144085592-144085592 on Build GRCh38
rs374430537 Chr.8: 144085479-144085479 on Build GRCh38
rs782625698 Chr.8: 144085656-144085656 on Build GRCh38
rs782259698 Chr.8: 144085553-144085554 on Build GRCh38
rs782252524 Chr.8: 144085643-144085643 on Build GRCh38
rs1127096 Chr.8: 144085569-144085569 on Build GRCh38
rs1031476698 Chr.8: 144085599-144085599 on Build GRCh38
rs781809982 Chr.8: 144085492-144085492 on Build GRCh38
rs782478182 Chr.8: 144085520-144085520 on Build GRCh38
rs782519746 Chr.8: 144085512-144085512 on Build GRCh38
rs112521600 Chr.8: 144085661-144085661 on Build GRCh38
rs968626124 Chr.8: 144085594-144085594 on Build GRCh38
rs782138044 Chr.8: 144085459-144085459 on Build GRCh38
rs782728894 Chr.8: 144085485-144085485 on Build GRCh38
rs368192144 Chr.8: 144085586-144085586 on Build GRCh38
rs575210002 Chr.8: 144085526-144085526 on Build GRCh38
rs781946943 Chr.8: 144085563-144085563 on Build GRCh38
rs782453969 Chr.8: 144085499-144085499 on Build GRCh38
rs781892219 Chr.8: 144085660-144085660 on Build GRCh38
rs782571129 Chr.8: 144085662-144085662 on Build GRCh38
rs371849501 Chr.8: 144085557-144085557 on Build GRCh38
rs376474105 Chr.8: 144085484-144085484 on Build GRCh38
rs920181678 Chr.8: 144085468-144085468 on Build GRCh38
rs782402057 Chr.8: 144085525-144085525 on Build GRCh38
rs782494799 Chr.8: 144085645-144085645 on Build GRCh38
rs781982804 Chr.8: 144085483-144085483 on Build GRCh38
rs370035088 Chr.8: 144085506-144085506 on Build GRCh38

View Full Product Details