Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10957511-10957784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf127
Entrez Gene ID
148345
Gene Name
chromosome 1 open reading frame 127
Gene Aliases
-
UniGene
Hs.127026
Gene Chromosome Location
Chr.1: 10946473-10982037 on Build GRCh38
Transcripts
NM_001170754.1 XM_017000373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667299 c.577C>G p.R193G Chr.1: 10957732-10957732 on Build GRCh38
174201 c.130C>T p.R44* Chr.1: 10957732-10957732 on Build GRCh38
382738 c.247G>C p.V83L Chr.1: 10957615-10957615 on Build GRCh38
893558 c.179A>G p.Y60C Chr.1: 10957683-10957683 on Build GRCh38
6581377 c.633C>T p.D211D Chr.1: 10957676-10957676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762445663 Chr.1: 10957547-10957547 on Build GRCh38
rs748521272 Chr.1: 10957695-10957695 on Build GRCh38
rs972557027 Chr.1: 10957626-10957626 on Build GRCh38
rs370007917 Chr.1: 10957633-10957633 on Build GRCh38
rs7511959 Chr.1: 10957552-10957552 on Build GRCh38
rs539069352 Chr.1: 10957606-10957606 on Build GRCh38
rs34691090 Chr.1: 10957560-10957560 on Build GRCh38
rs1008246466 Chr.1: 10957639-10957639 on Build GRCh38
rs779082540 Chr.1: 10957683-10957683 on Build GRCh38
rs947716138 Chr.1: 10957660-10957660 on Build GRCh38
rs765490325 Chr.1: 10957723-10957723 on Build GRCh38
rs759826302 Chr.1: 10957622-10957622 on Build GRCh38
rs770877171 Chr.1: 10957703-10957703 on Build GRCh38
rs763913947 Chr.1: 10957749-10957749 on Build GRCh38
rs769110588 Chr.1: 10957543-10957543 on Build GRCh38
rs772624436 Chr.1: 10957697-10957697 on Build GRCh38
rs372213743 Chr.1: 10957569-10957569 on Build GRCh38
rs780684223 Chr.1: 10957576-10957576 on Build GRCh38
rs752415081 Chr.1: 10957589-10957589 on Build GRCh38
rs770482435 Chr.1: 10957575-10957575 on Build GRCh38
rs763210466 Chr.1: 10957538-10957538 on Build GRCh38
rs200159296 Chr.1: 10957737-10957737 on Build GRCh38
rs762355962 Chr.1: 10957591-10957591 on Build GRCh38
rs745911890 Chr.1: 10957702-10957702 on Build GRCh38
rs769505025 Chr.1: 10957666-10957666 on Build GRCh38
rs142917171 Chr.1: 10957615-10957615 on Build GRCh38
rs372321055 Chr.1: 10957643-10957643 on Build GRCh38
rs917833959 Chr.1: 10957707-10957707 on Build GRCh38
rs760826441 Chr.1: 10957598-10957598 on Build GRCh38
rs753977796 Chr.1: 10957621-10957621 on Build GRCh38
rs547465775 Chr.1: 10957619-10957619 on Build GRCh38
rs12567143 Chr.1: 10957618-10957618 on Build GRCh38
rs571824426 Chr.1: 10957580-10957580 on Build GRCh38
rs751454965 Chr.1: 10957758-10957758 on Build GRCh38
rs778809021 Chr.1: 10957565-10957565 on Build GRCh38
rs916333997 Chr.1: 10957657-10957657 on Build GRCh38
rs780781215 Chr.1: 10957664-10957664 on Build GRCh38
rs752492379 Chr.1: 10957640-10957640 on Build GRCh38
rs750546827 Chr.1: 10957550-10957550 on Build GRCh38
rs949282026 Chr.1: 10957760-10957760 on Build GRCh38
rs774805100 Chr.1: 10957584-10957584 on Build GRCh38
rs997024707 Chr.1: 10957586-10957586 on Build GRCh38
rs35950437 Chr.1: 10957577-10957577 on Build GRCh38
rs774685320 Chr.1: 10957546-10957546 on Build GRCh38
rs766706826 Chr.1: 10957555-10957555 on Build GRCh38
rs772712478 Chr.1: 10957597-10957597 on Build GRCh38
rs565842231 Chr.1: 10957720-10957720 on Build GRCh38
rs752528228 Chr.1: 10957568-10957568 on Build GRCh38
rs761338661 Chr.1: 10957605-10957605 on Build GRCh38
rs377153016 Chr.1: 10957654-10957654 on Build GRCh38
rs138123175 Chr.1: 10957655-10957655 on Build GRCh38
rs766815981 Chr.1: 10957644-10957644 on Build GRCh38
rs767660308 Chr.1: 10957548-10957548 on Build GRCh38
rs765006781 Chr.1: 10957549-10957560 on Build GRCh38
rs762711419 Chr.1: 10957738-10957738 on Build GRCh38

View Full Product Details