Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53306813-53307058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf10
Entrez Gene ID
60314
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
Gamm1, MST024, MSTP024, MYG, MYG1
UniGene
Hs.655988
Gene Chromosome Location
Chr.12: 53299686-53307181 on Build GRCh38
Transcripts
NM_021640.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468583 c.1019_1020GC>AA p.C340>? Chr.12: 53307037-53307038 on Build GRCh38
468582 c.1019G>A p.C340Y Chr.12: 53307037-53307037 on Build GRCh38
6007862 c.997C>T p.Q333* Chr.12: 53307015-53307015 on Build GRCh38
4666691 c.905G>A p.R302Q Chr.12: 53306819-53306819 on Build GRCh38
459460 c.1008G>C p.G336G Chr.12: 53307026-53307026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770202277 Chr.12: 53306955-53306955 on Build GRCh38
rs751595894 Chr.12: 53306835-53306835 on Build GRCh38
rs746731363 Chr.12: 53307026-53307026 on Build GRCh38
rs774443884 Chr.12: 53306981-53306981 on Build GRCh38
rs1033498484 Chr.12: 53306836-53306836 on Build GRCh38
rs56900621 Chr.12: 53306838-53306838 on Build GRCh38
rs779838328 Chr.12: 53306887-53306887 on Build GRCh38
rs754873691 Chr.12: 53306911-53306911 on Build GRCh38
rs756016518 Chr.12: 53306882-53306882 on Build GRCh38
rs367601830 Chr.12: 53307017-53307017 on Build GRCh38
rs749310832 Chr.12: 53306896-53306896 on Build GRCh38
rs370681256 Chr.12: 53306892-53306892 on Build GRCh38
rs764301739 Chr.12: 53306833-53306833 on Build GRCh38
rs142952424 Chr.12: 53306998-53306998 on Build GRCh38
rs1045181 Chr.12: 53306922-53306922 on Build GRCh38
rs767879540 Chr.12: 53306860-53306860 on Build GRCh38
rs201220548 Chr.12: 53306918-53306918 on Build GRCh38
rs369983586 Chr.12: 53306912-53306914 on Build GRCh38
rs775866911 Chr.12: 53307030-53307030 on Build GRCh38
rs989405842 Chr.12: 53306867-53306867 on Build GRCh38
rs764525199 Chr.12: 53306972-53306972 on Build GRCh38
rs747626617 Chr.12: 53307003-53307003 on Build GRCh38
rs775858998 Chr.12: 53306965-53306965 on Build GRCh38
rs757951996 Chr.12: 53307007-53307007 on Build GRCh38
rs763451616 Chr.12: 53306971-53306971 on Build GRCh38
rs750207325 Chr.12: 53306877-53306877 on Build GRCh38
rs374635731 Chr.12: 53306991-53306991 on Build GRCh38
rs753251164 Chr.12: 53306873-53306873 on Build GRCh38
rs372883926 Chr.12: 53306989-53306989 on Build GRCh38
rs770496861 Chr.12: 53307029-53307029 on Build GRCh38
rs148585651 Chr.12: 53306997-53306997 on Build GRCh38
rs771670017 Chr.12: 53306929-53306929 on Build GRCh38
rs762020423 Chr.12: 53306984-53306984 on Build GRCh38
rs771273317 Chr.12: 53306916-53306916 on Build GRCh38
rs778745212 Chr.12: 53307002-53307002 on Build GRCh38
rs766129380 Chr.12: 53306992-53306992 on Build GRCh38
rs111308539 Chr.12: 53306889-53306889 on Build GRCh38
rs568308688 Chr.12: 53306927-53306927 on Build GRCh38
rs750530813 Chr.12: 53306935-53306935 on Build GRCh38
rs202213306 Chr.12: 53306988-53306988 on Build GRCh38
rs746208275 Chr.12: 53306946-53306946 on Build GRCh38
rs772891157 Chr.12: 53306939-53306939 on Build GRCh38

View Full Product Details