Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24225000-24225273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT5
Entrez Gene ID
2687
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGL, GGT 5, GGT-REL, GGTLA1
UniGene
Hs.437156
Gene Chromosome Location
Chr.22: 24219654-24245142 on Build GRCh38
Transcripts
NM_001099781.2 NM_001099782.2 NM_001302464.1 NM_001302465.1 NM_004121.3 XM_005261557.2 XM_005261558.2 XM_011530133.1 XM_011530134.1 XM_011530135.1 XM_011530136.1 XM_011530137.2 XM_017028768.1 XM_017028769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3708176 c.1600C>A p.P534T Chr.22: 24225010-24225010 on Build GRCh38
1032678 c.1578C>T p.S526S Chr.22: 24225032-24225032 on Build GRCh38
3964143 c.1544C>A p.A515E Chr.22: 24225066-24225066 on Build GRCh38
726142 c.1596C>T p.Y532Y Chr.22: 24225014-24225014 on Build GRCh38
6209035 c.1547C>A p.A516D Chr.22: 24225063-24225063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777378390 Chr.22: 24225119-24225119 on Build GRCh38
rs758005500 Chr.22: 24225106-24225106 on Build GRCh38
rs1049186603 Chr.22: 24225030-24225030 on Build GRCh38
rs779874745 Chr.22: 24225206-24225206 on Build GRCh38
rs1036547052 Chr.22: 24225092-24225092 on Build GRCh38
rs759800101 Chr.22: 24225234-24225234 on Build GRCh38
rs182867617 Chr.22: 24225132-24225132 on Build GRCh38
rs755578145 Chr.22: 24225082-24225082 on Build GRCh38
rs746671447 Chr.22: 24225129-24225129 on Build GRCh38
rs767531469 Chr.22: 24225054-24225054 on Build GRCh38
rs749181027 Chr.22: 24225212-24225212 on Build GRCh38
rs1056441583 Chr.22: 24225162-24225162 on Build GRCh38
rs781106041 Chr.22: 24225135-24225135 on Build GRCh38
rs892435899 Chr.22: 24225247-24225247 on Build GRCh38
rs369915142 Chr.22: 24225204-24225204 on Build GRCh38
rs112092021 Chr.22: 24225190-24225190 on Build GRCh38
rs199510644 Chr.22: 24225214-24225214 on Build GRCh38
rs777095964 Chr.22: 24225223-24225223 on Build GRCh38
rs200684960 Chr.22: 24225222-24225222 on Build GRCh38
rs894850599 Chr.22: 24225188-24225188 on Build GRCh38
rs755159959 Chr.22: 24225138-24225138 on Build GRCh38
rs765632350 Chr.22: 24225086-24225086 on Build GRCh38
rs773450659 Chr.22: 24225055-24225055 on Build GRCh38
rs754142074 Chr.22: 24225074-24225074 on Build GRCh38
rs745618291 Chr.22: 24225198-24225198 on Build GRCh38
rs760823413 Chr.22: 24225065-24225065 on Build GRCh38
rs1003134270 Chr.22: 24225159-24225159 on Build GRCh38
rs113295783 Chr.22: 24225243-24225243 on Build GRCh38
rs369769344 Chr.22: 24225124-24225124 on Build GRCh38
rs761855801 Chr.22: 24225027-24225027 on Build GRCh38
rs574777300 Chr.22: 24225101-24225101 on Build GRCh38
rs201616574 Chr.22: 24225219-24225219 on Build GRCh38
rs371500873 Chr.22: 24225066-24225066 on Build GRCh38
rs375273797 Chr.22: 24225123-24225123 on Build GRCh38
rs952661223 Chr.22: 24225022-24225022 on Build GRCh38

View Full Product Details