Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27488387-27488622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
UniGene
Hs.282121
Gene Chromosome Location
Chr.12: 27446482-27506911 on Build GRCh38
Transcripts
NM_001145010.1 XM_011520634.2 XM_011520635.2 XM_011520636.2 XM_017019254.1 XM_017019255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
401331 c.363C>A p.L121L Chr.12: 27488460-27488460 on Build GRCh38
3753134 c.593G>A p.S198N Chr.12: 27488540-27488540 on Build GRCh38
1361088 c.419G>A p.G140D Chr.12: 27488516-27488516 on Build GRCh38
1361087 c.569G>A p.G190D Chr.12: 27488516-27488516 on Build GRCh38
3753136 c.443G>A p.S148N Chr.12: 27488540-27488540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs967140263 Chr.12: 27488505-27488505 on Build GRCh38
rs572537159 Chr.12: 27488560-27488560 on Build GRCh38
rs1054342378 Chr.12: 27488461-27488461 on Build GRCh38
rs921200523 Chr.12: 27488481-27488481 on Build GRCh38
rs148846851 Chr.12: 27488512-27488512 on Build GRCh38
rs766850054 Chr.12: 27488445-27488445 on Build GRCh38
rs890995265 Chr.12: 27488523-27488523 on Build GRCh38
rs448088 Chr.12: 27488442-27488442 on Build GRCh38
rs186554621 Chr.12: 27488534-27488534 on Build GRCh38
rs953015419 Chr.12: 27488521-27488521 on Build GRCh38
rs412978 Chr.12: 27488540-27488540 on Build GRCh38
rs959193956 Chr.12: 27488570-27488570 on Build GRCh38
rs145820348 Chr.12: 27488413-27488413 on Build GRCh38
rs985762867 Chr.12: 27488530-27488530 on Build GRCh38
rs773980437 Chr.12: 27488589-27488589 on Build GRCh38
rs758607397 Chr.12: 27488415-27488415 on Build GRCh38
rs930745266 Chr.12: 27488593-27488593 on Build GRCh38
rs76288777 Chr.12: 27488532-27488532 on Build GRCh38
rs919444816 Chr.12: 27488559-27488559 on Build GRCh38
rs1034231736 Chr.12: 27488421-27488421 on Build GRCh38
rs925618863 Chr.12: 27488515-27488515 on Build GRCh38
rs1020549126 Chr.12: 27488495-27488495 on Build GRCh38
rs924599814 Chr.12: 27488430-27488430 on Build GRCh38

View Full Product Details