Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108907987-108908258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCA1C
Entrez Gene ID
9626
Gene Name
guanylate cyclase activator 1C
Gene Aliases
GCAP3
UniGene
Hs.233363
Gene Chromosome Location
Chr.3: 108907644-108955195 on Build GRCh38
Transcripts
NM_005459.3 XM_005247910.3 XM_011513334.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3585050 c.615G>A p.K205K Chr.3: 108908037-108908037 on Build GRCh38
3585051 c.577G>A p.D193N Chr.3: 108908075-108908075 on Build GRCh38
3585052 c.498G>A p.L166L Chr.3: 108908154-108908154 on Build GRCh38
3845772 c.560G>A p.R187Q Chr.3: 108908132-108908132 on Build GRCh38
3585054 c.468C>T p.I156I Chr.3: 108908184-108908184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776975372 Chr.3: 108908207-108908207 on Build GRCh38
rs200165076 Chr.3: 108908111-108908111 on Build GRCh38
rs953822004 Chr.3: 108908058-108908058 on Build GRCh38
rs1028693252 Chr.3: 108908133-108908133 on Build GRCh38
rs370259737 Chr.3: 108908162-108908162 on Build GRCh38
rs764004495 Chr.3: 108908019-108908019 on Build GRCh38
rs749320902 Chr.3: 108908114-108908115 on Build GRCh38
rs202047632 Chr.3: 108908105-108908105 on Build GRCh38
rs1020871668 Chr.3: 108908024-108908024 on Build GRCh38
rs866962064 Chr.3: 108908165-108908165 on Build GRCh38
rs16854916 Chr.3: 108908177-108908177 on Build GRCh38
rs377468133 Chr.3: 108908142-108908142 on Build GRCh38
rs966485382 Chr.3: 108908042-108908042 on Build GRCh38
rs778238802 Chr.3: 108908084-108908084 on Build GRCh38
rs774254004 Chr.3: 108908148-108908148 on Build GRCh38
rs757566635 Chr.3: 108908201-108908201 on Build GRCh38
rs761853844 Chr.3: 108908034-108908034 on Build GRCh38
rs767452738 Chr.3: 108908166-108908166 on Build GRCh38
rs985131659 Chr.3: 108908085-108908085 on Build GRCh38
rs781686520 Chr.3: 108908121-108908121 on Build GRCh38
rs751443999 Chr.3: 108908029-108908029 on Build GRCh38
rs140784316 Chr.3: 108908051-108908051 on Build GRCh38
rs944300868 Chr.3: 108908131-108908131 on Build GRCh38
rs752983518 Chr.3: 108908077-108908077 on Build GRCh38
rs762730549 Chr.3: 108908138-108908138 on Build GRCh38
rs140624957 Chr.3: 108908009-108908009 on Build GRCh38
rs759659775 Chr.3: 108908208-108908208 on Build GRCh38
rs775107558 Chr.3: 108908132-108908132 on Build GRCh38
rs977702220 Chr.3: 108908193-108908193 on Build GRCh38
rs146701862 Chr.3: 108908075-108908075 on Build GRCh38
rs565596041 Chr.3: 108908122-108908122 on Build GRCh38
rs144704413 Chr.3: 108908053-108908053 on Build GRCh38
rs750391393 Chr.3: 108908167-108908167 on Build GRCh38
rs200874514 Chr.3: 108908010-108908010 on Build GRCh38
rs771419612 Chr.3: 108908109-108908109 on Build GRCh38
rs769302584 Chr.3: 108908125-108908125 on Build GRCh38
rs775125499 Chr.3: 108908017-108908017 on Build GRCh38
rs140265807 Chr.3: 108908098-108908098 on Build GRCh38
rs374759057 Chr.3: 108908115-108908115 on Build GRCh38
rs777879332 Chr.3: 108908187-108908187 on Build GRCh38
rs752965142 Chr.3: 108908179-108908179 on Build GRCh38
rs373644613 Chr.3: 108908195-108908195 on Build GRCh38
rs762674581 Chr.3: 108908018-108908018 on Build GRCh38
rs201373110 Chr.3: 108908033-108908033 on Build GRCh38
rs557545795 Chr.3: 108908041-108908041 on Build GRCh38
rs367948714 Chr.3: 108908178-108908178 on Build GRCh38
rs368031353 Chr.3: 108908231-108908231 on Build GRCh38
rs755141026 Chr.3: 108908052-108908052 on Build GRCh38

View Full Product Details