Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 827788-828045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZU1
Entrez Gene ID
566
Gene Name
azurocidin 1
Gene Aliases
AZAMP, AZU, CAP37, HBP, HUMAZUR, NAZC, hHBP
UniGene
Hs.72885
Gene Chromosome Location
Chr.19: 827826-832018 on Build GRCh38
Transcripts
NM_001700.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1003496 c.23C>T p.A8V Chr.19: 827869-827869 on Build GRCh38
3693237 c.50C>A p.S17* Chr.19: 827896-827896 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs909815375 Chr.19: 827989-827989 on Build GRCh38
rs750078815 Chr.19: 827941-827941 on Build GRCh38
rs1052363793 Chr.19: 827896-827896 on Build GRCh38
rs943819284 Chr.19: 827985-827985 on Build GRCh38
rs942537431 Chr.19: 828023-828023 on Build GRCh38
rs1007846453 Chr.19: 827853-827853 on Build GRCh38
rs1046919524 Chr.19: 827970-827970 on Build GRCh38
rs572420584 Chr.19: 827924-827924 on Build GRCh38
rs771209379 Chr.19: 827864-827864 on Build GRCh38
rs760328681 Chr.19: 827923-827924 on Build GRCh38
rs201295101 Chr.19: 827935-827935 on Build GRCh38
rs1040785457 Chr.19: 828008-828008 on Build GRCh38
rs918269095 Chr.19: 827818-827818 on Build GRCh38
rs919846579 Chr.19: 827950-827950 on Build GRCh38
rs367776900 Chr.19: 827923-827923 on Build GRCh38
rs747245762 Chr.19: 827863-827863 on Build GRCh38
rs146182220 Chr.19: 827897-827897 on Build GRCh38
rs375208692 Chr.19: 827903-827903 on Build GRCh38
rs778232796 Chr.19: 828024-828024 on Build GRCh38
rs951125091 Chr.19: 827980-827980 on Build GRCh38
rs649411 Chr.19: 827975-827975 on Build GRCh38
rs768465758 Chr.19: 827913-827913 on Build GRCh38
rs556429462 Chr.19: 827819-827819 on Build GRCh38
rs886893547 Chr.19: 827846-827846 on Build GRCh38
rs758875960 Chr.19: 827844-827844 on Build GRCh38
rs900081651 Chr.19: 827886-827886 on Build GRCh38
rs982097798 Chr.19: 827922-827922 on Build GRCh38
rs755648949 Chr.19: 827821-827821 on Build GRCh38
rs139128406 Chr.19: 827901-827901 on Build GRCh38
rs34124897 Chr.19: 827891-827891 on Build GRCh38
rs201600404 Chr.19: 827904-827904 on Build GRCh38
rs926584370 Chr.19: 827933-827933 on Build GRCh38
rs998239872 Chr.19: 827894-827894 on Build GRCh38
rs760152527 Chr.19: 827947-827947 on Build GRCh38
rs564111589 Chr.19: 827948-827948 on Build GRCh38
rs970839244 Chr.19: 827911-827911 on Build GRCh38
rs113981649 Chr.19: 827957-827957 on Build GRCh38
rs753290003 Chr.19: 827953-827953 on Build GRCh38
rs779346784 Chr.19: 827822-827822 on Build GRCh38
rs748666639 Chr.19: 827843-827843 on Build GRCh38
rs991383517 Chr.19: 827946-827946 on Build GRCh38

View Full Product Details