Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5015056-5015329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTL1
Entrez Gene ID
54360
Gene Name
cytokine like 1
Gene Aliases
C17, C4orf4
UniGene
Hs.13872
Gene Chromosome Location
Chr.4: 5014586-5019470 on Build GRCh38
Transcripts
NM_018659.2 XM_017008299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
179110 c.386C>T p.T129M Chr.4: 5015176-5015176 on Build GRCh38
4408315 c.387G>A p.T129T Chr.4: 5015175-5015175 on Build GRCh38
1055880 c.375C>T p.I125I Chr.4: 5015187-5015187 on Build GRCh38
4545919 c.388G>A p.V130I Chr.4: 5015174-5015174 on Build GRCh38
1055881 c.336A>G p.V112V Chr.4: 5015226-5015226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761108071 Chr.4: 5015262-5015262 on Build GRCh38
rs768797314 Chr.4: 5015167-5015167 on Build GRCh38
rs1010640036 Chr.4: 5015087-5015087 on Build GRCh38
rs764581502 Chr.4: 5015192-5015193 on Build GRCh38
rs758050570 Chr.4: 5015194-5015194 on Build GRCh38
rs6847131 Chr.4: 5015114-5015114 on Build GRCh38
rs773394467 Chr.4: 5015261-5015261 on Build GRCh38
rs1030014834 Chr.4: 5015271-5015271 on Build GRCh38
rs749759953 Chr.4: 5015161-5015161 on Build GRCh38
rs757725243 Chr.4: 5015206-5015206 on Build GRCh38
rs375564556 Chr.4: 5015136-5015136 on Build GRCh38
rs754484807 Chr.4: 5015109-5015109 on Build GRCh38
rs754455404 Chr.4: 5015273-5015273 on Build GRCh38
rs780809052 Chr.4: 5015232-5015232 on Build GRCh38
rs952911381 Chr.4: 5015222-5015222 on Build GRCh38
rs772365669 Chr.4: 5015256-5015256 on Build GRCh38
rs774618923 Chr.4: 5015175-5015175 on Build GRCh38
rs752190395 Chr.4: 5015192-5015192 on Build GRCh38
rs563528458 Chr.4: 5015092-5015092 on Build GRCh38
rs181452793 Chr.4: 5015112-5015112 on Build GRCh38
rs137887145 Chr.4: 5015162-5015162 on Build GRCh38
rs370399082 Chr.4: 5015163-5015163 on Build GRCh38
rs11722554 Chr.4: 5015156-5015156 on Build GRCh38
rs368311678 Chr.4: 5015155-5015155 on Build GRCh38
rs758069053 Chr.4: 5015120-5015120 on Build GRCh38
rs532688534 Chr.4: 5015280-5015280 on Build GRCh38
rs73198523 Chr.4: 5015245-5015245 on Build GRCh38
rs749362670 Chr.4: 5015246-5015246 on Build GRCh38
rs34323389 Chr.4: 5015223-5015224 on Build GRCh38
rs751219293 Chr.4: 5015219-5015219 on Build GRCh38
rs976522565 Chr.4: 5015220-5015220 on Build GRCh38
rs185753377 Chr.4: 5015268-5015268 on Build GRCh38
rs746816904 Chr.4: 5015139-5015139 on Build GRCh38
rs376640064 Chr.4: 5015242-5015242 on Build GRCh38
rs778529679 Chr.4: 5015111-5015111 on Build GRCh38
rs544477622 Chr.4: 5015298-5015298 on Build GRCh38
rs767816726 Chr.4: 5015188-5015188 on Build GRCh38
rs374634052 Chr.4: 5015142-5015142 on Build GRCh38
rs762531488 Chr.4: 5015281-5015281 on Build GRCh38
rs778419855 Chr.4: 5015236-5015236 on Build GRCh38
rs985901835 Chr.4: 5015303-5015303 on Build GRCh38
rs138467520 Chr.4: 5015223-5015223 on Build GRCh38
rs558515410 Chr.4: 5015251-5015251 on Build GRCh38
rs969089428 Chr.4: 5015094-5015094 on Build GRCh38
rs761905601 Chr.4: 5015255-5015255 on Build GRCh38
rs761041037 Chr.4: 5015189-5015189 on Build GRCh38
rs933106031 Chr.4: 5015079-5015079 on Build GRCh38
rs777555516 Chr.4: 5015196-5015196 on Build GRCh38
rs750945407 Chr.4: 5015108-5015108 on Build GRCh38
rs538800764 Chr.4: 5015248-5015248 on Build GRCh38
rs201652130 Chr.4: 5015226-5015226 on Build GRCh38
rs200085707 Chr.4: 5015176-5015176 on Build GRCh38

View Full Product Details