Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57172651-57172924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM192A
Entrez Gene ID
80011
Gene Name
family with sequence similarity 192 member A
Gene Aliases
C16orf94, CDA018, CDA10, NIP30
UniGene
Hs.396740
Gene Chromosome Location
Chr.16: 57152466-57186242 on Build GRCh38
Transcripts
NM_024946.2 XM_005256156.4 XM_005256158.3 XM_005256160.2 XM_005256163.2 XM_005256164.3 XM_005256165.2 XM_006721275.3 XM_011523343.2 XM_017023681.1 XM_017023682.1 XM_017023683.1 XM_017023684.1 XM_017023685.1 XM_017023686.1 XM_017023687.1 XM_017023688.1 XM_017023689.1 XM_017023690.1 XM_017023691.1 XM_017023692.1 XM_017023693.1 XM_017023694.1 XM_017023695.1 XM_017023696.1 XM_017023697.1 XM_017023698.1 XM_017023699.1 XM_017023700.1 XM_017023701.1 XM_017023702.1 XM_017023703.1 XM_017023704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2836307 c.226+1G>A p.? Chr.16: 57172775-57172775 on Build GRCh38
4682837 c.192G>T p.R64S Chr.16: 57172810-57172810 on Build GRCh38
3969604 c.207C>T p.Y69Y Chr.16: 57172795-57172795 on Build GRCh38
6328201 c.226+5G>A p.? Chr.16: 57172771-57172771 on Build GRCh38
3510469 c.142G>A p.V48I Chr.16: 57172860-57172860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7192556 Chr.16: 57172687-57172687 on Build GRCh38
rs765715075 Chr.16: 57172878-57172878 on Build GRCh38
rs755335453 Chr.16: 57172871-57172871 on Build GRCh38
rs986876953 Chr.16: 57172776-57172776 on Build GRCh38
rs780543235 Chr.16: 57172897-57172897 on Build GRCh38
rs775256303 Chr.16: 57172787-57172787 on Build GRCh38
rs767648815 Chr.16: 57172722-57172722 on Build GRCh38
rs756043615 Chr.16: 57172902-57172902 on Build GRCh38
rs748128025 Chr.16: 57172776-57172781 on Build GRCh38
rs566394282 Chr.16: 57172807-57172815 on Build GRCh38
rs369877166 Chr.16: 57172783-57172783 on Build GRCh38
rs749152481 Chr.16: 57172794-57172794 on Build GRCh38
rs3751702 Chr.16: 57172725-57172725 on Build GRCh38
rs778975665 Chr.16: 57172789-57172789 on Build GRCh38
rs754310197 Chr.16: 57172732-57172732 on Build GRCh38
rs773384495 Chr.16: 57172807-57172807 on Build GRCh38
rs768449295 Chr.16: 57172795-57172795 on Build GRCh38
rs924117889 Chr.16: 57172733-57172733 on Build GRCh38
rs755127738 Chr.16: 57172754-57172757 on Build GRCh38
rs575837295 Chr.16: 57172721-57172721 on Build GRCh38
rs756676664 Chr.16: 57172724-57172724 on Build GRCh38
rs745443818 Chr.16: 57172899-57172899 on Build GRCh38
rs777500500 Chr.16: 57172749-57172749 on Build GRCh38
rs1039747090 Chr.16: 57172818-57172818 on Build GRCh38
rs372393113 Chr.16: 57172742-57172742 on Build GRCh38
rs745759613 Chr.16: 57172760-57172760 on Build GRCh38
rs11555755 Chr.16: 57172805-57172805 on Build GRCh38
rs886943674 Chr.16: 57172874-57172874 on Build GRCh38
rs761248202 Chr.16: 57172828-57172828 on Build GRCh38
rs774379565 Chr.16: 57172796-57172796 on Build GRCh38
rs375146943 Chr.16: 57172887-57172887 on Build GRCh38
rs942639690 Chr.16: 57172817-57172817 on Build GRCh38
rs769898845 Chr.16: 57172801-57172803 on Build GRCh38
rs767059423 Chr.16: 57172840-57172840 on Build GRCh38
rs750846774 Chr.16: 57172723-57172723 on Build GRCh38
rs751129045 Chr.16: 57172890-57172890 on Build GRCh38
rs546658164 Chr.16: 57172851-57172851 on Build GRCh38
rs55680142 Chr.16: 57172750-57172750 on Build GRCh38
rs762119081 Chr.16: 57172804-57172804 on Build GRCh38
rs756971349 Chr.16: 57172896-57172896 on Build GRCh38
rs931233633 Chr.16: 57172841-57172841 on Build GRCh38
rs528576294 Chr.16: 57172816-57172816 on Build GRCh38
rs781365232 Chr.16: 57172754-57172755 on Build GRCh38
rs764360207 Chr.16: 57172720-57172720 on Build GRCh38
rs780782917 Chr.16: 57172759-57172759 on Build GRCh38

View Full Product Details