Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102716596-102716833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

Gene Symbol
GPRASP2
Entrez Gene ID
114928
Gene Name
G protein-coupled receptor associated sorting protein 2
Gene Aliases
GASP2
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102712176-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

SNP Detail

SNP ID Chromosome Location
rs912019428 Chr.X: 102716689-102716689 on Build GRCh38
rs769510587 Chr.X: 102716726-102716726 on Build GRCh38
rs1036348853 Chr.X: 102716753-102716753 on Build GRCh38
rs766463571 Chr.X: 102716622-102716622 on Build GRCh38
rs748760002 Chr.X: 102716760-102716760 on Build GRCh38
rs375571887 Chr.X: 102716694-102716694 on Build GRCh38
rs372332241 Chr.X: 102716683-102716683 on Build GRCh38
rs769537243 Chr.X: 102716623-102716623 on Build GRCh38
rs768764070 Chr.X: 102716796-102716796 on Build GRCh38
rs747996065 Chr.X: 102716621-102716621 on Build GRCh38
rs745501234 Chr.X: 102716702-102716702 on Build GRCh38
rs755507410 Chr.X: 102716660-102716660 on Build GRCh38
rs138144817 Chr.X: 102716618-102716618 on Build GRCh38
rs779714407 Chr.X: 102716746-102716746 on Build GRCh38
rs753150242 Chr.X: 102716687-102716687 on Build GRCh38
rs972451861 Chr.X: 102716624-102716624 on Build GRCh38

View Full Product Details