Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2236756-2237005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L2
Entrez Gene ID
1775
Gene Name
deoxyribonuclease I-like 2
Gene Aliases
DNAS1L2
UniGene
Hs.103503
Gene Chromosome Location
Chr.16: 2236252-2238711 on Build GRCh38
Transcripts
NM_001301680.1 NM_001374.2 XM_011522399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3690854 c.28G>A p.A10T Chr.16: 2236844-2236844 on Build GRCh38
1204228 c.122C>A p.A41D Chr.16: 2236938-2236938 on Build GRCh38
3794692 c.12C>T p.P4P Chr.16: 2236828-2236828 on Build GRCh38
4678789 c.87T>A p.I29I Chr.16: 2236903-2236903 on Build GRCh38
4430946 c.106A>G p.K36E Chr.16: 2236922-2236922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766007984 Chr.16: 2236974-2236978 on Build GRCh38
rs907194521 Chr.16: 2236958-2236958 on Build GRCh38
rs745485688 Chr.16: 2236950-2236950 on Build GRCh38
rs374157200 Chr.16: 2236934-2236934 on Build GRCh38
rs774053884 Chr.16: 2236937-2236937 on Build GRCh38
rs772725165 Chr.16: 2236865-2236865 on Build GRCh38
rs771884051 Chr.16: 2236959-2236959 on Build GRCh38
rs950706463 Chr.16: 2236868-2236868 on Build GRCh38
rs191517595 Chr.16: 2236875-2236875 on Build GRCh38
rs759409448 Chr.16: 2236817-2236817 on Build GRCh38
rs780442279 Chr.16: 2236853-2236853 on Build GRCh38
rs770641064 Chr.16: 2236870-2236870 on Build GRCh38
rs975824420 Chr.16: 2236801-2236801 on Build GRCh38
rs748729518 Chr.16: 2236932-2236932 on Build GRCh38
rs1019563043 Chr.16: 2236876-2236876 on Build GRCh38
rs757426045 Chr.16: 2236842-2236842 on Build GRCh38
rs767484858 Chr.16: 2236818-2236818 on Build GRCh38
rs779124859 Chr.16: 2236843-2236843 on Build GRCh38
rs931672724 Chr.16: 2236844-2236844 on Build GRCh38
rs747517722 Chr.16: 2236855-2236855 on Build GRCh38
rs761800411 Chr.16: 2236984-2236984 on Build GRCh38
rs776432899 Chr.16: 2236980-2236980 on Build GRCh38
rs538519818 Chr.16: 2236978-2236978 on Build GRCh38
rs367638880 Chr.16: 2236813-2236813 on Build GRCh38
rs774139745 Chr.16: 2236873-2236873 on Build GRCh38
rs762665213 Chr.16: 2236850-2236851 on Build GRCh38
rs760623413 Chr.16: 2236890-2236890 on Build GRCh38
rs781553956 Chr.16: 2236930-2236930 on Build GRCh38
rs200032282 Chr.16: 2236969-2236969 on Build GRCh38
rs746257430 Chr.16: 2236785-2236785 on Build GRCh38
rs777878114 Chr.16: 2236829-2236829 on Build GRCh38
rs753899553 Chr.16: 2236892-2236892 on Build GRCh38
rs376704000 Chr.16: 2236891-2236891 on Build GRCh38
rs767398904 Chr.16: 2236884-2236884 on Build GRCh38
rs760403854 Chr.16: 2236973-2236973 on Build GRCh38
rs747353441 Chr.16: 2236929-2236929 on Build GRCh38
rs772514934 Chr.16: 2236792-2236792 on Build GRCh38
rs775982907 Chr.16: 2236797-2236797 on Build GRCh38
rs750633649 Chr.16: 2236895-2236895 on Build GRCh38
rs748761472 Chr.16: 2236867-2236867 on Build GRCh38
rs772425363 Chr.16: 2236852-2236852 on Build GRCh38
rs772635994 Chr.16: 2236810-2236810 on Build GRCh38
rs752667826 Chr.16: 2236822-2236822 on Build GRCh38
rs769323139 Chr.16: 2236807-2236807 on Build GRCh38
rs761155611 Chr.16: 2236806-2236806 on Build GRCh38
rs975295199 Chr.16: 2236951-2236951 on Build GRCh38
rs756056800 Chr.16: 2236828-2236828 on Build GRCh38
rs775189893 Chr.16: 2236888-2236888 on Build GRCh38
rs762622221 Chr.16: 2236811-2236811 on Build GRCh38
rs369778646 Chr.16: 2236898-2236898 on Build GRCh38
rs968529557 Chr.16: 2236974-2236974 on Build GRCh38
rs769070425 Chr.16: 2236859-2236859 on Build GRCh38
rs780421376 Chr.16: 2236908-2236908 on Build GRCh38
rs753895862 Chr.16: 2236830-2236830 on Build GRCh38
rs757336279 Chr.16: 2236893-2236893 on Build GRCh38
rs746167259 Chr.16: 2236851-2236851 on Build GRCh38
rs536434345 Chr.16: 2236814-2236814 on Build GRCh38

View Full Product Details