Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 79201535-79201808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGN3
Entrez Gene ID
9324
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
PNAS-24, PNAS-25, TRIP7
UniGene
Hs.77558
Gene Chromosome Location
Chr.6: 79201245-79234738 on Build GRCh38
Transcripts
NM_001201362.1 NM_001201363.1 NM_001318884.1 NM_001318885.1 NM_001318886.1 NM_001318887.1 NM_001318888.1 NM_004242.3 NM_138730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681039 c.289G>A p.E97K Chr.6: 79201699-79201699 on Build GRCh38
1081879 c.295G>T p.E99* Chr.6: 79201693-79201693 on Build GRCh38
5772806 c.291G>C p.E97D Chr.6: 79201697-79201697 on Build GRCh38
4545389 c.283G>C p.D95H Chr.6: 79201705-79201705 on Build GRCh38
6335919 c.262-3C>A p.? Chr.6: 79201729-79201729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs560675825 Chr.6: 79201591-79201591 on Build GRCh38
rs758271519 Chr.6: 79201668-79201668 on Build GRCh38
rs762495208 Chr.6: 79201777-79201777 on Build GRCh38
rs747543374 Chr.6: 79201762-79201762 on Build GRCh38
rs758142127 Chr.6: 79201736-79201736 on Build GRCh38
rs762106155 Chr.6: 79201656-79201657 on Build GRCh38
rs764858714 Chr.6: 79201752-79201752 on Build GRCh38
rs775973553 Chr.6: 79201644-79201644 on Build GRCh38
rs761584191 Chr.6: 79201654-79201654 on Build GRCh38
rs750056069 Chr.6: 79201653-79201653 on Build GRCh38
rs755506120 Chr.6: 79201693-79201693 on Build GRCh38
rs530700035 Chr.6: 79201751-79201751 on Build GRCh38
rs776218235 Chr.6: 79201755-79201755 on Build GRCh38
rs9448658 Chr.6: 79201764-79201764 on Build GRCh38
rs950535803 Chr.6: 79201566-79201566 on Build GRCh38
rs182681016 Chr.6: 79201697-79201697 on Build GRCh38
rs9448657 Chr.6: 79201573-79201573 on Build GRCh38
rs532506985 Chr.6: 79201669-79201669 on Build GRCh38
rs752710119 Chr.6: 79201699-79201699 on Build GRCh38
rs750803070 Chr.6: 79201656-79201656 on Build GRCh38
rs1039423124 Chr.6: 79201787-79201787 on Build GRCh38
rs762670071 Chr.6: 79201774-79201774 on Build GRCh38
rs772687462 Chr.6: 79201768-79201768 on Build GRCh38
rs777793703 Chr.6: 79201710-79201710 on Build GRCh38
rs765239771 Chr.6: 79201593-79201593 on Build GRCh38
rs944692373 Chr.6: 79201780-79201780 on Build GRCh38
rs772384598 Chr.6: 79201660-79201660 on Build GRCh38
rs761383067 Chr.6: 79201564-79201564 on Build GRCh38
rs766885887 Chr.6: 79201777-79201777 on Build GRCh38
rs971047695 Chr.6: 79201590-79201590 on Build GRCh38
rs1027931363 Chr.6: 79201565-79201565 on Build GRCh38
rs199768088 Chr.6: 79201588-79201588 on Build GRCh38
rs745710812 Chr.6: 79201728-79201728 on Build GRCh38
rs533445936 Chr.6: 79201753-79201754 on Build GRCh38
rs947549722 Chr.6: 79201719-79201719 on Build GRCh38
rs752158058 Chr.6: 79201684-79201684 on Build GRCh38
rs370315630 Chr.6: 79201700-79201700 on Build GRCh38
rs768576918 Chr.6: 79201753-79201753 on Build GRCh38
rs374045554 Chr.6: 79201752-79201752 on Build GRCh38
rs779804073 Chr.6: 79201741-79201741 on Build GRCh38
rs71547744 Chr.6: 79201743-79201743 on Build GRCh38
rs541460283 Chr.6: 79201632-79201632 on Build GRCh38
rs776269640 Chr.6: 79201742-79201743 on Build GRCh38
rs540368467 Chr.6: 79201576-79201576 on Build GRCh38
rs1024002247 Chr.6: 79201586-79201586 on Build GRCh38

View Full Product Details